Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 159257352-159257849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDSUB1
Entrez Gene ID
151525
Gene Name
WD repeat, sterile alpha motif and U-box domain containing 1
Gene Aliases
UBOX6, WDSAM1
UniGene
Hs.20848
Gene Chromosome Location
Chr.2: 159235793-159286799 on Build GRCh38
Transcripts
NM_001128212.2 NM_001128213.1 NM_001307994.1 NM_152528.2 XM_005246355.2 XM_006712341.3 XM_011510753.2 XM_011510754.2 XM_017003481.1 XM_017003482.1 XM_017003483.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5506806 c.931G>C p.D311H Chr.2: 159257779-159257779 on Build GRCh38
148964 c.906C>T p.D302D Chr.2: 159257804-159257804 on Build GRCh38
7347469 c.898T>C p.S300P Chr.2: 159257812-159257812 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs749703767 Chr.2: 159257751-159257751 on Build GRCh38
rs547256031 Chr.2: 159257592-159257592 on Build GRCh38
rs571780796 Chr.2: 159257533-159257533 on Build GRCh38
rs780770631 Chr.2: 159257708-159257708 on Build GRCh38
rs928193903 Chr.2: 159257645-159257645 on Build GRCh38
rs189633231 Chr.2: 159257674-159257674 on Build GRCh38
rs540135465 Chr.2: 159257450-159257450 on Build GRCh38
rs967674481 Chr.2: 159257746-159257746 on Build GRCh38
rs940973285 Chr.2: 159257648-159257648 on Build GRCh38
rs139910347 Chr.2: 159257797-159257797 on Build GRCh38
rs927950196 Chr.2: 159257636-159257636 on Build GRCh38
rs772634704 Chr.2: 159257543-159257543 on Build GRCh38
rs767575875 Chr.2: 159257776-159257776 on Build GRCh38
rs377087274 Chr.2: 159257576-159257576 on Build GRCh38
rs1005261383 Chr.2: 159257429-159257429 on Build GRCh38
rs556590414 Chr.2: 159257532-159257532 on Build GRCh38
rs899153011 Chr.2: 159257670-159257670 on Build GRCh38
rs774542067 Chr.2: 159257764-159257764 on Build GRCh38
rs551421686 Chr.2: 159257433-159257433 on Build GRCh38
rs967296075 Chr.2: 159257494-159257494 on Build GRCh38
rs185746496 Chr.2: 159257447-159257447 on Build GRCh38
rs897023152 Chr.2: 159257477-159257477 on Build GRCh38
rs939392659 Chr.2: 159257672-159257672 on Build GRCh38
rs554949824 Chr.2: 159257387-159257388 on Build GRCh38
rs977462589 Chr.2: 159257512-159257512 on Build GRCh38
rs994480297 Chr.2: 159257714-159257714 on Build GRCh38
rs906798694 Chr.2: 159257426-159257426 on Build GRCh38
rs895298335 Chr.2: 159257382-159257382 on Build GRCh38
rs773681711 Chr.2: 159257721-159257721 on Build GRCh38
rs771349884 Chr.2: 159257758-159257758 on Build GRCh38
rs368712484 Chr.2: 159257785-159257785 on Build GRCh38
rs759810220 Chr.2: 159257767-159257767 on Build GRCh38
rs912777868 Chr.2: 159257411-159257411 on Build GRCh38
rs1029306154 Chr.2: 159257490-159257490 on Build GRCh38
rs114630061 Chr.2: 159257379-159257379 on Build GRCh38
rs770357374 Chr.2: 159257716-159257716 on Build GRCh38
rs143101860 Chr.2: 159257595-159257595 on Build GRCh38
rs749288955 Chr.2: 159257472-159257472 on Build GRCh38
rs538885380 Chr.2: 159257540-159257540 on Build GRCh38
rs6746645 Chr.2: 159257489-159257489 on Build GRCh38
rs11324351 Chr.2: 159257395-159257395 on Build GRCh38
rs867429782 Chr.2: 159257724-159257724 on Build GRCh38
rs538486161 Chr.2: 159257521-159257521 on Build GRCh38
rs553459700 Chr.2: 159257388-159257388 on Build GRCh38
rs6746843 Chr.2: 159257473-159257473 on Build GRCh38
rs751243617 Chr.2: 159257818-159257818 on Build GRCh38
rs760832652 Chr.2: 159257791-159257791 on Build GRCh38
rs941455447 Chr.2: 159257677-159257677 on Build GRCh38
rs754721313 Chr.2: 159257824-159257824 on Build GRCh38
rs6746547 Chr.2: 159257425-159257425 on Build GRCh38
rs899222546 Chr.2: 159257437-159257437 on Build GRCh38
rs566972139 Chr.2: 159257492-159257492 on Build GRCh38
rs148220102 Chr.2: 159257684-159257684 on Build GRCh38
rs945532727 Chr.2: 159257423-159257423 on Build GRCh38
rs948250486 Chr.2: 159257422-159257422 on Build GRCh38
rs945590825 Chr.2: 159257596-159257596 on Build GRCh38
rs3815475 Chr.2: 159257804-159257804 on Build GRCh38
rs144861177 Chr.2: 159257798-159257798 on Build GRCh38
rs747645391 Chr.2: 159257712-159257712 on Build GRCh38
rs140416254 Chr.2: 159257809-159257809 on Build GRCh38
rs897001797 Chr.2: 159257686-159257686 on Build GRCh38
rs762114699 Chr.2: 159257518-159257518 on Build GRCh38
rs13395766 Chr.2: 159257675-159257675 on Build GRCh38
rs376285387 Chr.2: 159257455-159257455 on Build GRCh38
rs554997624 Chr.2: 159257661-159257661 on Build GRCh38
rs748717473 Chr.2: 159257568-159257568 on Build GRCh38
rs1055160329 Chr.2: 159257385-159257385 on Build GRCh38
rs543181768 Chr.2: 159257633-159257633 on Build GRCh38

View Full Product Details