Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 169544757-169545304 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FASTKD1
Entrez Gene ID
79675
Gene Name
FAST kinase domains 1
Gene Aliases
-
UniGene
Hs.529276
Gene Chromosome Location
Chr.2: 169527879-169573921 on Build GRCh38
Transcripts
NM_001281476.2 NM_001322046.1 NM_001322048.1 NM_001322049.1 NM_024622.5 NR_104020.2 XM_006712750.3 XM_006712751.3 XM_017004911.1 XM_017004912.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6660696 c.1776A>T p.E592D Chr.2: 169544761-169544761 on Build GRCh38
1291351 c.1710T>C p.P570P Chr.2: 169544827-169544827 on Build GRCh38
4428246 c.1774G>T p.E592* Chr.2: 169544763-169544763 on Build GRCh38
3425437 c.1717A>G p.I573V Chr.2: 169544820-169544820 on Build GRCh38
244419 c.1706A>G p.H569R Chr.2: 169544831-169544831 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs988219520 Chr.2: 169545086-169545086 on Build GRCh38
rs543988629 Chr.2: 169544988-169544988 on Build GRCh38
rs548738013 Chr.2: 169545115-169545115 on Build GRCh38
rs1000969162 Chr.2: 169545037-169545037 on Build GRCh38
rs973328717 Chr.2: 169544904-169544904 on Build GRCh38
rs1053209936 Chr.2: 169545161-169545161 on Build GRCh38
rs1013151191 Chr.2: 169544782-169544782 on Build GRCh38
rs541992657 Chr.2: 169544800-169544800 on Build GRCh38
rs568580925 Chr.2: 169545129-169545129 on Build GRCh38
rs950555423 Chr.2: 169544933-169544933 on Build GRCh38
rs904777901 Chr.2: 169545139-169545139 on Build GRCh38
rs777323445 Chr.2: 169545184-169545184 on Build GRCh38
rs765570133 Chr.2: 169544832-169544832 on Build GRCh38
rs78491846 Chr.2: 169545210-169545210 on Build GRCh38
rs756397050 Chr.2: 169544848-169544848 on Build GRCh38
rs929052928 Chr.2: 169545215-169545215 on Build GRCh38
rs927663325 Chr.2: 169544961-169544961 on Build GRCh38
rs139546135 Chr.2: 169544797-169544797 on Build GRCh38
rs144290701 Chr.2: 169544832-169544832 on Build GRCh38
rs761596946 Chr.2: 169544821-169544821 on Build GRCh38
rs779873542 Chr.2: 169544845-169544845 on Build GRCh38
rs758211801 Chr.2: 169544841-169544841 on Build GRCh38
rs767292918 Chr.2: 169544976-169544976 on Build GRCh38
rs779510022 Chr.2: 169544857-169544857 on Build GRCh38
rs771797962 Chr.2: 169544804-169544804 on Build GRCh38
rs986086123 Chr.2: 169545157-169545157 on Build GRCh38
rs1032416453 Chr.2: 169545038-169545038 on Build GRCh38
rs771279778 Chr.2: 169544855-169544855 on Build GRCh38
rs12470809 Chr.2: 169545163-169545163 on Build GRCh38
rs749757992 Chr.2: 169544794-169544794 on Build GRCh38
rs777819670 Chr.2: 169544849-169544849 on Build GRCh38
rs763221593 Chr.2: 169544887-169544888 on Build GRCh38
rs530602208 Chr.2: 169544949-169544949 on Build GRCh38
rs999001128 Chr.2: 169545145-169545145 on Build GRCh38
rs746407002 Chr.2: 169544861-169544861 on Build GRCh38
rs749402081 Chr.2: 169544854-169544854 on Build GRCh38
rs372186353 Chr.2: 169544903-169544903 on Build GRCh38
rs946388039 Chr.2: 169545088-169545088 on Build GRCh38
rs1008916481 Chr.2: 169545183-169545183 on Build GRCh38
rs748178523 Chr.2: 169544791-169544791 on Build GRCh38
rs187841192 Chr.2: 169545062-169545062 on Build GRCh38
rs561762592 Chr.2: 169544805-169544805 on Build GRCh38
rs182548470 Chr.2: 169545008-169545008 on Build GRCh38
rs759954911 Chr.2: 169544801-169544801 on Build GRCh38
rs371143972 Chr.2: 169544839-169544839 on Build GRCh38
rs750676911 Chr.2: 169544860-169544860 on Build GRCh38
rs532958380 Chr.2: 169545006-169545006 on Build GRCh38
rs965095567 Chr.2: 169544895-169544895 on Build GRCh38
rs760388786 Chr.2: 169544873-169544873 on Build GRCh38
rs767567370 Chr.2: 169545071-169545071 on Build GRCh38
rs1023833633 Chr.2: 169545019-169545019 on Build GRCh38
rs765224579 Chr.2: 169544835-169544835 on Build GRCh38
rs954725757 Chr.2: 169545133-169545133 on Build GRCh38
rs537214154 Chr.2: 169544982-169544982 on Build GRCh38
rs772530384 Chr.2: 169544863-169544863 on Build GRCh38
rs781422946 Chr.2: 169544790-169544790 on Build GRCh38
rs753444362 Chr.2: 169544816-169544816 on Build GRCh38
rs199963186 Chr.2: 169545202-169545203 on Build GRCh38
rs1056846738 Chr.2: 169545219-169545219 on Build GRCh38
rs769844413 Chr.2: 169544793-169544793 on Build GRCh38
rs564029447 Chr.2: 169545002-169545002 on Build GRCh38
rs750603867 Chr.2: 169544910-169544910 on Build GRCh38
rs1043730999 Chr.2: 169544785-169544785 on Build GRCh38
rs537516575 Chr.2: 169545151-169545151 on Build GRCh38
rs776061701 Chr.2: 169544870-169544870 on Build GRCh38
rs765567122 Chr.2: 169544902-169544902 on Build GRCh38
rs754759283 Chr.2: 169544934-169544934 on Build GRCh38
rs916237750 Chr.2: 169545026-169545026 on Build GRCh38
rs528814867 Chr.2: 169545085-169545085 on Build GRCh38

View Full Product Details