Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 172088327-172088821 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLX1
Entrez Gene ID
1745
Gene Name
distal-less homeobox 1
Gene Aliases
-
UniGene
Hs.407015
Gene Chromosome Location
Chr.2: 172085480-172089674 on Build GRCh38
Transcripts
NM_001038493.1 NM_178120.4

SNP Detail

SNP ID Chromosome Location
rs80173687 Chr.2: 172088754-172088754 on Build GRCh38
rs929538309 Chr.2: 172088444-172088444 on Build GRCh38
rs555718509 Chr.2: 172088746-172088746 on Build GRCh38
rs1024711545 Chr.2: 172088761-172088761 on Build GRCh38
rs181665527 Chr.2: 172088693-172088693 on Build GRCh38
rs1013326043 Chr.2: 172088542-172088542 on Build GRCh38
rs1032278028 Chr.2: 172088677-172088677 on Build GRCh38
rs788173 Chr.2: 172088732-172088732 on Build GRCh38
rs537456161 Chr.2: 172088737-172088737 on Build GRCh38
rs775819773 Chr.2: 172088795-172088795 on Build GRCh38
rs1049327718 Chr.2: 172088497-172088497 on Build GRCh38
rs1023380288 Chr.2: 172088530-172088530 on Build GRCh38
rs912424203 Chr.2: 172088712-172088712 on Build GRCh38
rs896219906 Chr.2: 172088534-172088534 on Build GRCh38
rs788172 Chr.2: 172088710-172088710 on Build GRCh38
rs935835990 Chr.2: 172088465-172088465 on Build GRCh38
rs966115503 Chr.2: 172088731-172088731 on Build GRCh38
rs995032862 Chr.2: 172088370-172088370 on Build GRCh38
rs36106394 Chr.2: 172088553-172088553 on Build GRCh38
rs531211683 Chr.2: 172088369-172088369 on Build GRCh38
rs910649773 Chr.2: 172088391-172088391 on Build GRCh38
rs117299827 Chr.2: 172088750-172088750 on Build GRCh38
rs966355073 Chr.2: 172088608-172088608 on Build GRCh38
rs558285040 Chr.2: 172088716-172088716 on Build GRCh38
rs572454360 Chr.2: 172088779-172088779 on Build GRCh38
rs1024356787 Chr.2: 172088402-172088402 on Build GRCh38
rs568296029 Chr.2: 172088513-172088513 on Build GRCh38
rs971888291 Chr.2: 172088414-172088414 on Build GRCh38
rs375887391 Chr.2: 172088461-172088461 on Build GRCh38
rs986596825 Chr.2: 172088380-172088380 on Build GRCh38
rs913158991 Chr.2: 172088619-172088619 on Build GRCh38
rs1040380934 Chr.2: 172088430-172088430 on Build GRCh38
rs559737120 Chr.2: 172088774-172088774 on Build GRCh38
rs535716353 Chr.2: 172088641-172088641 on Build GRCh38
rs970503755 Chr.2: 172088539-172088539 on Build GRCh38
rs945655045 Chr.2: 172088626-172088626 on Build GRCh38
rs944801802 Chr.2: 172088427-172088427 on Build GRCh38
rs1020567832 Chr.2: 172088549-172088549 on Build GRCh38
rs927099229 Chr.2: 172088459-172088459 on Build GRCh38
rs1027881660 Chr.2: 172088384-172088384 on Build GRCh38
rs909482321 Chr.2: 172088771-172088771 on Build GRCh38
rs990037511 Chr.2: 172088600-172088600 on Build GRCh38
rs1001150305 Chr.2: 172088665-172088665 on Build GRCh38
rs547320111 Chr.2: 172088687-172088687 on Build GRCh38
rs1001998116 Chr.2: 172088770-172088770 on Build GRCh38
rs950692693 Chr.2: 172088389-172088389 on Build GRCh38
rs887905581 Chr.2: 172088509-172088509 on Build GRCh38
rs186854626 Chr.2: 172088797-172088797 on Build GRCh38
rs1055256762 Chr.2: 172088532-172088532 on Build GRCh38
rs3821186 Chr.2: 172088439-172088439 on Build GRCh38
rs971452847 Chr.2: 172088769-172088769 on Build GRCh38

View Full Product Details