Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 201299116-201299555 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALS2CR12
Entrez Gene ID
130540
Gene Name
amyotrophic lateral sclerosis 2 chromosome region candidate 12
Gene Aliases
-
UniGene
Hs.107944
Gene Chromosome Location
Chr.2: 201288271-201357398 on Build GRCh38
Transcripts
NM_001127391.2 NM_001289993.1 NM_139163.3 NR_110620.1 XM_011510606.2 XM_011510610.2 XM_011510612.2 XM_017003360.1 XM_017003361.1 XM_017003362.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3798505 c.922G>A p.E308K Chr.2: 201299258-201299258 on Build GRCh38
7276898 c.880-3_880-2delTA p.? Chr.2: 201299302-201299303 on Build GRCh38
1014890 c.935C>T p.T312I Chr.2: 201299245-201299245 on Build GRCh38
5829363 c.880-3delT p.? Chr.2: 201299303-201299303 on Build GRCh38
5354937 c.919G>C p.E307Q Chr.2: 201299261-201299261 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1033654060 Chr.2: 201299317-201299317 on Build GRCh38
rs752858823 Chr.2: 201299224-201299224 on Build GRCh38
rs763058005 Chr.2: 201299337-201299337 on Build GRCh38
rs765343679 Chr.2: 201299218-201299218 on Build GRCh38
rs376686644 Chr.2: 201299172-201299172 on Build GRCh38
rs752548515 Chr.2: 201299393-201299393 on Build GRCh38
rs200864880 Chr.2: 201299244-201299244 on Build GRCh38
rs1010853294 Chr.2: 201299314-201299314 on Build GRCh38
rs756109843 Chr.2: 201299515-201299515 on Build GRCh38
rs1024259910 Chr.2: 201299374-201299374 on Build GRCh38
rs767397212 Chr.2: 201299316-201299316 on Build GRCh38
rs770790240 Chr.2: 201299326-201299326 on Build GRCh38
rs954752329 Chr.2: 201299375-201299375 on Build GRCh38
rs924449752 Chr.2: 201299500-201299500 on Build GRCh38
rs754718609 Chr.2: 201299311-201299311 on Build GRCh38
rs959382431 Chr.2: 201299338-201299338 on Build GRCh38
rs942010160 Chr.2: 201299394-201299394 on Build GRCh38
rs551563665 Chr.2: 201299158-201299158 on Build GRCh38
rs369249173 Chr.2: 201299292-201299292 on Build GRCh38
rs778324754 Chr.2: 201299245-201299245 on Build GRCh38
rs971219165 Chr.2: 201299376-201299376 on Build GRCh38
rs753022256 Chr.2: 201299335-201299335 on Build GRCh38
rs75012247 Chr.2: 201299400-201299400 on Build GRCh38
rs760482730 Chr.2: 201299187-201299187 on Build GRCh38
rs1004423229 Chr.2: 201299238-201299238 on Build GRCh38
rs766717160 Chr.2: 201299176-201299176 on Build GRCh38
rs749173863 Chr.2: 201299290-201299290 on Build GRCh38
rs377614873 Chr.2: 201299365-201299365 on Build GRCh38
rs778344249 Chr.2: 201299297-201299297 on Build GRCh38
rs752046128 Chr.2: 201299248-201299248 on Build GRCh38
rs760160965 Chr.2: 201299195-201299195 on Build GRCh38
rs771518956 Chr.2: 201299315-201299315 on Build GRCh38
rs759794883 Chr.2: 201299334-201299334 on Build GRCh38
rs781596416 Chr.2: 201299272-201299272 on Build GRCh38
rs776574543 Chr.2: 201299333-201299333 on Build GRCh38
rs150075649 Chr.2: 201299258-201299258 on Build GRCh38
rs963885715 Chr.2: 201299364-201299364 on Build GRCh38
rs983874712 Chr.2: 201299386-201299386 on Build GRCh38
rs138471870 Chr.2: 201299280-201299280 on Build GRCh38
rs200651165 Chr.2: 201299192-201299192 on Build GRCh38
rs766149742 Chr.2: 201299190-201299190 on Build GRCh38
rs891281755 Chr.2: 201299178-201299178 on Build GRCh38

View Full Product Details