Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 233742635-233743180 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJB3
Entrez Gene ID
414061
Gene Name
DnaJ heat shock protein family (Hsp40) member B3
Gene Aliases
HCG3
UniGene
Hs.725533
Gene Chromosome Location
Chr.2: 233742750-233744015 on Build GRCh38
Transcripts
NM_001001394.3 NM_019075.2 NM_019093.2 NM_007120.2 NM_019078.1 NM_001072.3 NM_205862.1 NM_019077.2 NM_019076.4 NM_021027.2

Gene Symbol
UGT1A10
Entrez Gene ID
54575
Gene Name
UDP glucuronosyltransferase family 1 member A10
Gene Aliases
UDPGT, UGT-1J, UGT1-10, UGT1.10, UGT1J
UniGene
Hs.554822
Gene Chromosome Location
Chr.2: 233636477-233773305 on Build GRCh38
Transcripts
NM_001001394.3 NM_019075.2 NM_019093.2 NM_007120.2 NM_019078.1 NM_001072.3 NM_205862.1 NM_019077.2 NM_019076.4 NM_021027.2

Gene Symbol
UGT1A3
Entrez Gene ID
54659
Gene Name
UDP glucuronosyltransferase family 1 member A3
Gene Aliases
UDPGT, UDPGT 1-3, UGT-1C, UGT1-03, UGT1.3, UGT1A3S, UGT1C
UniGene
Hs.554822
Gene Chromosome Location
Chr.2: 233729127-233773299 on Build GRCh38
Transcripts
NM_001001394.3 NM_019075.2 NM_019093.2 NM_007120.2 NM_019078.1 NM_001072.3 NM_205862.1 NM_019077.2 NM_019076.4 NM_021027.2

Gene Symbol
UGT1A4
Entrez Gene ID
54657
Gene Name
UDP glucuronosyltransferase family 1 member A4
Gene Aliases
HUG-BR2, UDPGT, UDPGT 1-4, UGT-1D, UGT1-04, UGT1.4, UGT1A4S, UGT1D
UniGene
Hs.554822
Gene Chromosome Location
Chr.2: 233718792-233773299 on Build GRCh38
Transcripts
NM_001001394.3 NM_019075.2 NM_019093.2 NM_007120.2 NM_019078.1 NM_001072.3 NM_205862.1 NM_019077.2 NM_019076.4 NM_021027.2

Gene Symbol
UGT1A5
Entrez Gene ID
54579
Gene Name
UDP glucuronosyltransferase family 1 member A5
Gene Aliases
UDPGT, UDPGT 1-5, UGT1E
UniGene
Hs.554822
Gene Chromosome Location
Chr.2: 233712992-233773299 on Build GRCh38
Transcripts
NM_001001394.3 NM_019075.2 NM_019093.2 NM_007120.2 NM_019078.1 NM_001072.3 NM_205862.1 NM_019077.2 NM_019076.4 NM_021027.2

Gene Symbol
UGT1A6
Entrez Gene ID
54578
Gene Name
UDP glucuronosyltransferase family 1 member A6
Gene Aliases
GNT1, HLUGP, HLUGP1, UDPGT, UDPGT 1-6, UGT1, UGT1A6S, UGT1F
UniGene
Hs.554822
Gene Chromosome Location
Chr.2: 233691675-233773305 on Build GRCh38
Transcripts
NM_001001394.3 NM_019075.2 NM_019093.2 NM_007120.2 NM_019078.1 NM_001072.3 NM_205862.1 NM_019077.2 NM_019076.4 NM_021027.2

Gene Symbol
UGT1A7
Entrez Gene ID
54577
Gene Name
UDP glucuronosyltransferase family 1 member A7
Gene Aliases
UDPGT, UDPGT 1-7, UGT-1G, UGT1-07, UGT1.7, UGT1G
UniGene
Hs.554822
Gene Chromosome Location
Chr.2: 233681938-233773299 on Build GRCh38
Transcripts
NM_001001394.3 NM_019075.2 NM_019093.2 NM_007120.2 NM_019078.1 NM_001072.3 NM_205862.1 NM_019077.2 NM_019076.4 NM_021027.2

Gene Symbol
UGT1A8
Entrez Gene ID
54576
Gene Name
UDP glucuronosyltransferase family 1 member A8
Gene Aliases
UDPGT, UDPGT 1-8, UGT-1H, UGT1-08, UGT1.8, UGT1A8S, UGT1H
UniGene
Hs.554822
Gene Chromosome Location
Chr.2: 233617645-233773299 on Build GRCh38
Transcripts
NM_001001394.3 NM_019075.2 NM_019093.2 NM_007120.2 NM_019078.1 NM_001072.3 NM_205862.1 NM_019077.2 NM_019076.4 NM_021027.2

Gene Symbol
UGT1A9
Entrez Gene ID
54600
Gene Name
UDP glucuronosyltransferase family 1 member A9
Gene Aliases
HLUGP4, LUGP4, UDPGT, UDPGT 1-9, UGT-1I, UGT1-09, UGT1-9, UGT1.9, UGT1A9S, UGT1AI, UGT1I
UniGene
Hs.554822
Gene Chromosome Location
Chr.2: 233671898-233773305 on Build GRCh38
Transcripts
NM_001001394.3 NM_019075.2 NM_019093.2 NM_007120.2 NM_019078.1 NM_001072.3 NM_205862.1 NM_019077.2 NM_019076.4 NM_021027.2

SNP Detail

SNP ID Chromosome Location
rs774791073 Chr.2: 233742807-233742807 on Build GRCh38
rs955838724 Chr.2: 233743056-233743056 on Build GRCh38
rs28900374 Chr.2: 233742896-233742896 on Build GRCh38
rs978880235 Chr.2: 233742761-233742761 on Build GRCh38
rs926867030 Chr.2: 233743086-233743086 on Build GRCh38
rs17863795 Chr.2: 233742723-233742723 on Build GRCh38
rs969729560 Chr.2: 233743066-233743066 on Build GRCh38
rs78644424 Chr.2: 233742936-233742936 on Build GRCh38
rs186747767 Chr.2: 233742893-233742893 on Build GRCh38
rs77486749 Chr.2: 233743142-233743142 on Build GRCh38
rs79897461 Chr.2: 233743058-233743058 on Build GRCh38
rs1030653819 Chr.2: 233742681-233742681 on Build GRCh38
rs3796088 Chr.2: 233743154-233743154 on Build GRCh38
rs574856700 Chr.2: 233743023-233743023 on Build GRCh38
rs942176977 Chr.2: 233742911-233742911 on Build GRCh38
rs546559267 Chr.2: 233742846-233742846 on Build GRCh38
rs1054800 Chr.2: 233743076-233743076 on Build GRCh38
rs890471778 Chr.2: 233742980-233742980 on Build GRCh38
rs1017364884 Chr.2: 233742736-233742736 on Build GRCh38
rs924392805 Chr.2: 233742769-233742769 on Build GRCh38
rs992545321 Chr.2: 233742812-233742812 on Build GRCh38
rs546122146 Chr.2: 233742713-233742713 on Build GRCh38
rs993518638 Chr.2: 233742674-233742674 on Build GRCh38
rs550270409 Chr.2: 233742781-233742781 on Build GRCh38
rs916889730 Chr.2: 233743065-233743065 on Build GRCh38
rs558840772 Chr.2: 233742915-233742915 on Build GRCh38
rs577515653 Chr.2: 233743050-233743050 on Build GRCh38
rs980978105 Chr.2: 233743077-233743077 on Build GRCh38
rs1054809 Chr.2: 233743013-233743013 on Build GRCh38
rs955891060 Chr.2: 233742787-233742787 on Build GRCh38
rs555126231 Chr.2: 233742982-233742982 on Build GRCh38
rs397839835 Chr.2: 233743068-233743068 on Build GRCh38
rs968540830 Chr.2: 233742757-233742757 on Build GRCh38
rs1054803 Chr.2: 233743055-233743055 on Build GRCh38
rs1007409604 Chr.2: 233743005-233743005 on Build GRCh38
rs554129345 Chr.2: 233743042-233743042 on Build GRCh38
rs397706649 Chr.2: 233743045-233743045 on Build GRCh38
rs183327032 Chr.2: 233743035-233743035 on Build GRCh38
rs760786392 Chr.2: 233743032-233743032 on Build GRCh38
rs1038154831 Chr.2: 233742922-233742922 on Build GRCh38
rs78331994 Chr.2: 233743143-233743143 on Build GRCh38
rs949643400 Chr.2: 233742883-233742883 on Build GRCh38
rs76952857 Chr.2: 233742782-233742782 on Build GRCh38
rs187768174 Chr.2: 233743103-233743103 on Build GRCh38
rs191993803 Chr.2: 233742959-233742959 on Build GRCh38
rs968490640 Chr.2: 233743038-233743038 on Build GRCh38
rs919324462 Chr.2: 233743135-233743135 on Build GRCh38
rs764495397 Chr.2: 233742967-233742967 on Build GRCh38
rs564880841 Chr.2: 233742813-233742813 on Build GRCh38
rs942184123 Chr.2: 233743088-233743088 on Build GRCh38
rs1054805 Chr.2: 233743017-233743017 on Build GRCh38
rs879195953 Chr.2: 233743067-233743067 on Build GRCh38
rs181572285 Chr.2: 233742744-233742744 on Build GRCh38
rs750908913 Chr.2: 233742939-233742939 on Build GRCh38
rs1031836290 Chr.2: 233743054-233743054 on Build GRCh38

View Full Product Details