Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 959444-959944 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RSPO4
Entrez Gene ID
343637
Gene Name
R-spondin 4
Gene Aliases
C20orf182, CRISTIN4
UniGene
Hs.444980
Gene Chromosome Location
Chr.20: 958452-1002264 on Build GRCh38
Transcripts
NM_001029871.3 NM_001040007.2

SNP Detail

SNP ID Chromosome Location
rs561775766 Chr.20: 959600-959600 on Build GRCh38
rs534495204 Chr.20: 959821-959821 on Build GRCh38
rs138721103 Chr.20: 959534-959534 on Build GRCh38
rs549960964 Chr.20: 959740-959740 on Build GRCh38
rs923025461 Chr.20: 959593-959593 on Build GRCh38
rs890022462 Chr.20: 959473-959473 on Build GRCh38
rs519478 Chr.20: 959871-959871 on Build GRCh38
rs1013650369 Chr.20: 959781-959781 on Build GRCh38
rs182220957 Chr.20: 959475-959475 on Build GRCh38
rs534453835 Chr.20: 959914-959914 on Build GRCh38
rs1038583988 Chr.20: 959739-959739 on Build GRCh38
rs569735482 Chr.20: 959757-959757 on Build GRCh38
rs942145395 Chr.20: 959792-959792 on Build GRCh38
rs1002770344 Chr.20: 959840-959840 on Build GRCh38
rs1041652722 Chr.20: 959514-959514 on Build GRCh38
rs554325646 Chr.20: 959539-959539 on Build GRCh38
rs547417155 Chr.20: 959714-959714 on Build GRCh38
rs1025720603 Chr.20: 959536-959536 on Build GRCh38
rs147214985 Chr.20: 959773-959773 on Build GRCh38
rs554214124 Chr.20: 959861-959861 on Build GRCh38
rs13037150 Chr.20: 959569-959569 on Build GRCh38
rs140578124 Chr.20: 959864-959864 on Build GRCh38
rs925197355 Chr.20: 959574-959574 on Build GRCh38
rs991976414 Chr.20: 959545-959545 on Build GRCh38
rs935002635 Chr.20: 959583-959583 on Build GRCh38
rs369505776 Chr.20: 959720-959720 on Build GRCh38
rs745363996 Chr.20: 959817-959817 on Build GRCh38
rs1050145532 Chr.20: 959627-959627 on Build GRCh38
rs914216257 Chr.20: 959682-959682 on Build GRCh38
rs149543960 Chr.20: 959594-959594 on Build GRCh38
rs778168615 Chr.20: 959793-959793 on Build GRCh38
rs1041988690 Chr.20: 959730-959730 on Build GRCh38
rs62187523 Chr.20: 959468-959468 on Build GRCh38
rs516714 Chr.20: 959563-959563 on Build GRCh38
rs901643308 Chr.20: 959517-959517 on Build GRCh38
rs372460184 Chr.20: 959672-959672 on Build GRCh38
rs923213581 Chr.20: 959733-959733 on Build GRCh38
rs989795788 Chr.20: 959681-959681 on Build GRCh38
rs994179360 Chr.20: 959756-959756 on Build GRCh38
rs1021390148 Chr.20: 959898-959898 on Build GRCh38
rs543936481 Chr.20: 959795-959795 on Build GRCh38
rs776663727 Chr.20: 959860-959860 on Build GRCh38
rs989869147 Chr.20: 959879-959879 on Build GRCh38
rs754044098 Chr.20: 959529-959529 on Build GRCh38
rs562793850 Chr.20: 959572-959572 on Build GRCh38
rs962703842 Chr.20: 959502-959502 on Build GRCh38
rs987810699 Chr.20: 959905-959905 on Build GRCh38
rs1017159200 Chr.20: 959487-959487 on Build GRCh38
rs956858396 Chr.20: 959880-959880 on Build GRCh38
rs375151240 Chr.20: 959719-959719 on Build GRCh38
rs993521856 Chr.20: 959809-959809 on Build GRCh38
rs755386404 Chr.20: 959552-959552 on Build GRCh38
rs889187719 Chr.20: 959774-959774 on Build GRCh38
rs755620946 Chr.20: 959853-959853 on Build GRCh38
rs539605581 Chr.20: 959870-959870 on Build GRCh38
rs565712072 Chr.20: 959797-959797 on Build GRCh38
rs575155829 Chr.20: 959515-959515 on Build GRCh38
rs997365331 Chr.20: 959518-959518 on Build GRCh38
rs187647199 Chr.20: 959589-959589 on Build GRCh38
rs896560458 Chr.20: 959777-959777 on Build GRCh38
rs897802876 Chr.20: 959798-959798 on Build GRCh38
rs906824172 Chr.20: 959828-959828 on Build GRCh38
rs527566729 Chr.20: 959669-959669 on Build GRCh38
rs1023746551 Chr.20: 959837-959837 on Build GRCh38
rs927027881 Chr.20: 959649-959649 on Build GRCh38

View Full Product Details