Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 1229721-1230229 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAD21L1
Entrez Gene ID
642636
Gene Name
RAD21 cohesin complex component like 1
Gene Aliases
RAD21L, dJ545L17.2
UniGene
Hs.713451
Gene Chromosome Location
Chr.20: 1226043-1255876 on Build GRCh38
Transcripts
NM_001136566.2 XM_006723602.2 XM_006723603.2 XM_006723604.2 XM_006723605.3 XM_011529317.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2702904 c.186A>G p.G62G Chr.20: 1229921-1229921 on Build GRCh38
1024528 c.154G>A p.A52T Chr.20: 1229889-1229889 on Build GRCh38
6558497 c.161G>T p.R54L Chr.20: 1229896-1229896 on Build GRCh38
1024531 c.206G>T p.R69M Chr.20: 1229941-1229941 on Build GRCh38
3840412 c.268T>C p.C90R Chr.20: 1230003-1230003 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs570284468 Chr.20: 1229791-1229791 on Build GRCh38
rs892210511 Chr.20: 1229895-1229895 on Build GRCh38
rs749387399 Chr.20: 1230036-1230038 on Build GRCh38
rs191488531 Chr.20: 1230147-1230147 on Build GRCh38
rs114826915 Chr.20: 1229793-1229793 on Build GRCh38
rs949379103 Chr.20: 1230162-1230162 on Build GRCh38
rs891303418 Chr.20: 1229812-1229812 on Build GRCh38
rs960583928 Chr.20: 1230102-1230102 on Build GRCh38
rs775869641 Chr.20: 1230046-1230046 on Build GRCh38
rs572609202 Chr.20: 1229832-1229832 on Build GRCh38
rs949265673 Chr.20: 1229786-1229786 on Build GRCh38
rs1023743126 Chr.20: 1230146-1230146 on Build GRCh38
rs1018954332 Chr.20: 1229943-1229943 on Build GRCh38
rs563437729 Chr.20: 1230004-1230004 on Build GRCh38
rs148549307 Chr.20: 1229811-1229811 on Build GRCh38
rs759565758 Chr.20: 1229873-1229873 on Build GRCh38
rs930035981 Chr.20: 1230047-1230047 on Build GRCh38
rs779108203 Chr.20: 1230055-1230055 on Build GRCh38
rs746790695 Chr.20: 1229804-1229804 on Build GRCh38
rs988425520 Chr.20: 1230198-1230198 on Build GRCh38
rs956672591 Chr.20: 1230188-1230188 on Build GRCh38
rs1004804502 Chr.20: 1230057-1230057 on Build GRCh38
rs1015220989 Chr.20: 1230062-1230062 on Build GRCh38
rs775484064 Chr.20: 1229990-1229992 on Build GRCh38
rs1029937871 Chr.20: 1229899-1229899 on Build GRCh38
rs925573080 Chr.20: 1229779-1229780 on Build GRCh38
rs982888804 Chr.20: 1230070-1230070 on Build GRCh38
rs979442009 Chr.20: 1230170-1230170 on Build GRCh38
rs995869145 Chr.20: 1229991-1229991 on Build GRCh38
rs964210174 Chr.20: 1229997-1229997 on Build GRCh38
rs929953647 Chr.20: 1229764-1229764 on Build GRCh38
rs896517812 Chr.20: 1229778-1229778 on Build GRCh38
rs1040145226 Chr.20: 1229874-1229874 on Build GRCh38
rs879656830 Chr.20: 1229775-1229775 on Build GRCh38
rs377478024 Chr.20: 1229925-1229925 on Build GRCh38
rs1009308410 Chr.20: 1229896-1229896 on Build GRCh38
rs1053078726 Chr.20: 1229796-1229796 on Build GRCh38
rs450739 Chr.20: 1230003-1230003 on Build GRCh38
rs756440893 Chr.20: 1229929-1229929 on Build GRCh38
rs770821885 Chr.20: 1229843-1229843 on Build GRCh38
rs558252175 Chr.20: 1229946-1229946 on Build GRCh38
rs1013813924 Chr.20: 1230141-1230141 on Build GRCh38
rs552212230 Chr.20: 1230103-1230103 on Build GRCh38
rs79088681 Chr.20: 1230183-1230183 on Build GRCh38
rs187526000 Chr.20: 1229856-1229856 on Build GRCh38
rs866810401 Chr.20: 1229942-1229942 on Build GRCh38
rs776598376 Chr.20: 1229859-1229859 on Build GRCh38
rs141892713 Chr.20: 1229978-1229978 on Build GRCh38

View Full Product Details