Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 6031174-6031698 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRLS1
Entrez Gene ID
54675
Gene Name
cardiolipin synthase 1
Gene Aliases
C20orf155, CLS, CLS1, GCD10, dJ967N21.6
UniGene
Hs.743956
Gene Chromosome Location
Chr.20: 6005938-6040053 on Build GRCh38
Transcripts
NM_001127458.1 NM_001323561.1 NM_001323562.1 NM_001323563.1 NM_001323564.1 NM_019095.5 NR_136617.1 XM_005260738.4 XM_011529263.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6635609 c.326C>T p.A109V Chr.20: 6031333-6031333 on Build GRCh38
4674741 c.623C>T p.A208V Chr.20: 6031333-6031333 on Build GRCh38
6635613 c.303G>A p.S101S Chr.20: 6031310-6031310 on Build GRCh38
5529222 c.649C>T p.L217F Chr.20: 6031359-6031359 on Build GRCh38
444145 c.600G>A p.S200S Chr.20: 6031310-6031310 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs186055501 Chr.20: 6031608-6031608 on Build GRCh38
rs771091839 Chr.20: 6031332-6031332 on Build GRCh38
rs527640184 Chr.20: 6031530-6031530 on Build GRCh38
rs1047256338 Chr.20: 6031207-6031207 on Build GRCh38
rs745695219 Chr.20: 6031294-6031294 on Build GRCh38
rs980451405 Chr.20: 6031668-6031668 on Build GRCh38
rs962800758 Chr.20: 6031610-6031610 on Build GRCh38
rs755027777 Chr.20: 6031363-6031363 on Build GRCh38
rs551426286 Chr.20: 6031462-6031462 on Build GRCh38
rs1039467463 Chr.20: 6031489-6031489 on Build GRCh38
rs761353080 Chr.20: 6031379-6031379 on Build GRCh38
rs954591952 Chr.20: 6031492-6031492 on Build GRCh38
rs763517171 Chr.20: 6031244-6031244 on Build GRCh38
rs757968385 Chr.20: 6031242-6031242 on Build GRCh38
rs568211761 Chr.20: 6031510-6031510 on Build GRCh38
rs749237104 Chr.20: 6031299-6031299 on Build GRCh38
rs750090684 Chr.20: 6031401-6031401 on Build GRCh38
rs535985619 Chr.20: 6031319-6031319 on Build GRCh38
rs560123158 Chr.20: 6031471-6031471 on Build GRCh38
rs41282950 Chr.20: 6031288-6031288 on Build GRCh38
rs756040375 Chr.20: 6031330-6031331 on Build GRCh38
rs771270190 Chr.20: 6031309-6031309 on Build GRCh38
rs550373255 Chr.20: 6031645-6031645 on Build GRCh38
rs374161976 Chr.20: 6031655-6031655 on Build GRCh38
rs938867357 Chr.20: 6031638-6031638 on Build GRCh38
rs773179333 Chr.20: 6031310-6031310 on Build GRCh38
rs750418178 Chr.20: 6031289-6031292 on Build GRCh38
rs202046329 Chr.20: 6031336-6031336 on Build GRCh38
rs756042928 Chr.20: 6031404-6031404 on Build GRCh38
rs527298007 Chr.20: 6031217-6031217 on Build GRCh38
rs781275570 Chr.20: 6031318-6031318 on Build GRCh38
rs759950534 Chr.20: 6031343-6031343 on Build GRCh38
rs145778654 Chr.20: 6031353-6031353 on Build GRCh38
rs751023267 Chr.20: 6031276-6031276 on Build GRCh38
rs767332546 Chr.20: 6031269-6031270 on Build GRCh38
rs908695238 Chr.20: 6031617-6031617 on Build GRCh38
rs148108006 Chr.20: 6031358-6031358 on Build GRCh38
rs549256531 Chr.20: 6031636-6031636 on Build GRCh38
rs987361877 Chr.20: 6031499-6031499 on Build GRCh38
rs929089050 Chr.20: 6031586-6031586 on Build GRCh38
rs899735404 Chr.20: 6031491-6031491 on Build GRCh38
rs775765795 Chr.20: 6031351-6031351 on Build GRCh38
rs780852715 Chr.20: 6031292-6031292 on Build GRCh38
rs767121816 Chr.20: 6031380-6031380 on Build GRCh38
rs973517488 Chr.20: 6031568-6031568 on Build GRCh38
rs192068687 Chr.20: 6031640-6031640 on Build GRCh38
rs756878474 Chr.20: 6031278-6031278 on Build GRCh38
rs778702816 Chr.20: 6031354-6031354 on Build GRCh38
rs764608497 Chr.20: 6031359-6031359 on Build GRCh38
rs998136635 Chr.20: 6031511-6031511 on Build GRCh38
rs549697327 Chr.20: 6031584-6031584 on Build GRCh38
rs549940488 Chr.20: 6031480-6031480 on Build GRCh38
rs77279792 Chr.20: 6031461-6031461 on Build GRCh38
rs770137065 Chr.20: 6031350-6031350 on Build GRCh38
rs536668220 Chr.20: 6031553-6031553 on Build GRCh38
rs58014205 Chr.20: 6031297-6031297 on Build GRCh38
rs1003936434 Chr.20: 6031583-6031583 on Build GRCh38
rs2423132 Chr.20: 6031236-6031236 on Build GRCh38
rs753187646 Chr.20: 6031237-6031237 on Build GRCh38
rs951102647 Chr.20: 6031225-6031225 on Build GRCh38
rs755954900 Chr.20: 6031296-6031296 on Build GRCh38
rs1047200972 Chr.20: 6031599-6031599 on Build GRCh38

View Full Product Details