Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 7906017-7906534 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAO1
Entrez Gene ID
54363
Gene Name
hydroxyacid oxidase 1
Gene Aliases
GOX, GOX1, HAOX1
UniGene
Hs.193640
Gene Chromosome Location
Chr.20: 7882984-7940446 on Build GRCh38
Transcripts
NM_017545.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
724690 c.619G>A p.A207T Chr.20: 7906256-7906256 on Build GRCh38
3423837 c.604G>A p.D202N Chr.20: 7906271-7906271 on Build GRCh38
724691 c.631A>G p.K211E Chr.20: 7906244-7906244 on Build GRCh38
724692 c.686C>A p.T229K Chr.20: 7906189-7906189 on Build GRCh38
332851 c.604G>C p.D202H Chr.20: 7906271-7906271 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751903842 Chr.20: 7906370-7906370 on Build GRCh38
rs369366303 Chr.20: 7906139-7906139 on Build GRCh38
rs529336138 Chr.20: 7906247-7906247 on Build GRCh38
rs201390104 Chr.20: 7906108-7906108 on Build GRCh38
rs1056569070 Chr.20: 7906178-7906178 on Build GRCh38
rs564045779 Chr.20: 7906098-7906098 on Build GRCh38
rs969126020 Chr.20: 7906224-7906224 on Build GRCh38
rs552570900 Chr.20: 7906099-7906099 on Build GRCh38
rs968474389 Chr.20: 7906061-7906061 on Build GRCh38
rs987959356 Chr.20: 7906323-7906323 on Build GRCh38
rs764417339 Chr.20: 7906263-7906263 on Build GRCh38
rs930339242 Chr.20: 7906146-7906146 on Build GRCh38
rs749136809 Chr.20: 7906479-7906479 on Build GRCh38
rs939590810 Chr.20: 7906410-7906410 on Build GRCh38
rs889124125 Chr.20: 7906403-7906403 on Build GRCh38
rs939240254 Chr.20: 7906069-7906069 on Build GRCh38
rs983777066 Chr.20: 7906217-7906217 on Build GRCh38
rs769727018 Chr.20: 7906177-7906177 on Build GRCh38
rs768752030 Chr.20: 7906309-7906309 on Build GRCh38
rs978695894 Chr.20: 7906070-7906070 on Build GRCh38
rs775389428 Chr.20: 7906103-7906103 on Build GRCh38
rs993704181 Chr.20: 7906189-7906189 on Build GRCh38
rs866463468 Chr.20: 7906286-7906286 on Build GRCh38
rs750679582 Chr.20: 7906126-7906126 on Build GRCh38
rs775581760 Chr.20: 7906184-7906184 on Build GRCh38
rs764617706 Chr.20: 7906362-7906362 on Build GRCh38
rs541506883 Chr.20: 7906112-7906112 on Build GRCh38
rs115375518 Chr.20: 7906136-7906136 on Build GRCh38
rs775594099 Chr.20: 7906360-7906360 on Build GRCh38
rs770540248 Chr.20: 7906154-7906154 on Build GRCh38
rs760918760 Chr.20: 7906198-7906198 on Build GRCh38
rs765284804 Chr.20: 7906329-7906329 on Build GRCh38
rs139605533 Chr.20: 7906197-7906197 on Build GRCh38
rs758141038 Chr.20: 7906436-7906436 on Build GRCh38
rs149114100 Chr.20: 7906320-7906320 on Build GRCh38
rs888012900 Chr.20: 7906477-7906477 on Build GRCh38
rs996861023 Chr.20: 7906223-7906223 on Build GRCh38
rs1040478773 Chr.20: 7906425-7906425 on Build GRCh38
rs142998832 Chr.20: 7906222-7906222 on Build GRCh38
rs6055387 Chr.20: 7906378-7906378 on Build GRCh38
rs927806284 Chr.20: 7906041-7906041 on Build GRCh38
rs943384982 Chr.20: 7906404-7906404 on Build GRCh38
rs780725721 Chr.20: 7906165-7906165 on Build GRCh38
rs777628464 Chr.20: 7906450-7906450 on Build GRCh38
rs993653356 Chr.20: 7906437-7906437 on Build GRCh38
rs760000168 Chr.20: 7906299-7906300 on Build GRCh38
rs951706238 Chr.20: 7906357-7906357 on Build GRCh38
rs146825169 Chr.20: 7906187-7906187 on Build GRCh38
rs945567951 Chr.20: 7906440-7906440 on Build GRCh38
rs753903746 Chr.20: 7906138-7906138 on Build GRCh38
rs776852123 Chr.20: 7906287-7906288 on Build GRCh38
rs375983921 Chr.20: 7906264-7906264 on Build GRCh38
rs547717632 Chr.20: 7906427-7906427 on Build GRCh38
rs772743898 Chr.20: 7906117-7906117 on Build GRCh38
rs926848299 Chr.20: 7906386-7906386 on Build GRCh38
rs747917035 Chr.20: 7906311-7906311 on Build GRCh38
rs1043786683 Chr.20: 7906442-7906442 on Build GRCh38
rs1006887026 Chr.20: 7906480-7906480 on Build GRCh38
rs200824892 Chr.20: 7906153-7906153 on Build GRCh38
rs1022280403 Chr.20: 7906058-7906058 on Build GRCh38
rs569371584 Chr.20: 7906300-7906300 on Build GRCh38
rs766540254 Chr.20: 7906205-7906205 on Build GRCh38
rs892418834 Chr.20: 7906125-7906125 on Build GRCh38
rs61751020 Chr.20: 7906272-7906272 on Build GRCh38
rs143718983 Chr.20: 7906174-7906174 on Build GRCh38
rs115813887 Chr.20: 7906140-7906140 on Build GRCh38
rs566128799 Chr.20: 7906493-7906493 on Build GRCh38
rs760199443 Chr.20: 7906142-7906142 on Build GRCh38
rs780887877 Chr.20: 7906090-7906090 on Build GRCh38
rs920346357 Chr.20: 7906326-7906326 on Build GRCh38
rs564179630 Chr.20: 7906149-7906149 on Build GRCh38
rs756897739 Chr.20: 7906268-7906268 on Build GRCh38
rs1028877111 Chr.20: 7906243-7906243 on Build GRCh38
rs186542696 Chr.20: 7906377-7906377 on Build GRCh38
rs778174293 Chr.20: 7906150-7906150 on Build GRCh38
rs866212649 Chr.20: 7906286-7906287 on Build GRCh38
rs913789299 Chr.20: 7906371-7906371 on Build GRCh38
rs759547563 Chr.20: 7906232-7906232 on Build GRCh38
rs780735238 Chr.20: 7906271-7906271 on Build GRCh38
rs1010892616 Chr.20: 7906131-7906131 on Build GRCh38
rs1018778353 Chr.20: 7906270-7906270 on Build GRCh38
rs181543806 Chr.20: 7906303-7906303 on Build GRCh38
rs770632659 Chr.20: 7906262-7906268 on Build GRCh38
rs113948510 Chr.20: 7906274-7906274 on Build GRCh38
rs758959165 Chr.20: 7906253-7906253 on Build GRCh38
rs768568249 Chr.20: 7906196-7906196 on Build GRCh38
rs199630444 Chr.20: 7906302-7906302 on Build GRCh38
rs759865361 Chr.20: 7906328-7906328 on Build GRCh38
rs911622835 Chr.20: 7906128-7906128 on Build GRCh38
rs371141199 Chr.20: 7906322-7906322 on Build GRCh38
rs1044236719 Chr.20: 7906203-7906203 on Build GRCh38

View Full Product Details