Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 18305577-18306074 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF133
Entrez Gene ID
7692
Gene Name
zinc finger protein 133
Gene Aliases
ZNF150, pHZ-13, pHZ-66
UniGene
Hs.472221
Gene Chromosome Location
Chr.20: 18288228-18316996 on Build GRCh38
Transcripts
NM_001083330.2 NM_001282995.1 NM_001282996.1 NM_001282997.1 NM_001282998.1 NM_001282999.1 NM_001283000.1 NM_001283001.1 NM_001283002.1 NM_001283003.1 NM_001283004.1 NM_001283005.1 NM_001283006.1 NM_001283007.1 NM_001283008.1 NM_003434.5 XM_005260819.1 XM_005260820.4 XM_011529336.1 XM_011529337.1 XM_011529338.2 XM_011529339.1 XM_011529340.1 XM_017028041.1 XM_017028042.1 XM_017028043.1 XM_017028044.1 XM_017028045.1 XM_017028046.1 XM_017028047.1 XM_017028048.1 XM_017028049.1 XM_017028050.1 XM_017028051.1 XM_017028052.1 XM_017028053.1 XM_017028054.1 XM_017028055.1 XM_017028056.1 XM_017028057.1 XM_017028058.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4096868 c.4G>A p.A2T Chr.20: 18305690-18305690 on Build GRCh38
5006656 c.71C>T p.T24I Chr.20: 18305757-18305757 on Build GRCh38
1713259 c.59C>T p.P20L Chr.20: 18305745-18305745 on Build GRCh38
4389778 c.117A>G p.S39S Chr.20: 18305803-18305803 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs77421043 Chr.20: 18305712-18305712 on Build GRCh38
rs886123437 Chr.20: 18305880-18305880 on Build GRCh38
rs1005860720 Chr.20: 18305937-18305937 on Build GRCh38
rs892083044 Chr.20: 18305940-18305940 on Build GRCh38
rs1043138132 Chr.20: 18305652-18305652 on Build GRCh38
rs755345515 Chr.20: 18305791-18305791 on Build GRCh38
rs1016028533 Chr.20: 18305957-18305957 on Build GRCh38
rs770767083 Chr.20: 18305674-18305674 on Build GRCh38
rs78728539 Chr.20: 18305709-18305709 on Build GRCh38
rs553527770 Chr.20: 18306005-18306005 on Build GRCh38
rs766926977 Chr.20: 18305767-18305767 on Build GRCh38
rs200354196 Chr.20: 18305805-18305805 on Build GRCh38
rs772574579 Chr.20: 18305658-18305658 on Build GRCh38
rs1000175399 Chr.20: 18305917-18305917 on Build GRCh38
rs947369035 Chr.20: 18305620-18305620 on Build GRCh38
rs570361628 Chr.20: 18305718-18305718 on Build GRCh38
rs769622084 Chr.20: 18305832-18305832 on Build GRCh38
rs367680939 Chr.20: 18305661-18305661 on Build GRCh38
rs937850710 Chr.20: 18305754-18305754 on Build GRCh38
rs199774426 Chr.20: 18305818-18305818 on Build GRCh38
rs559259677 Chr.20: 18305632-18305632 on Build GRCh38
rs202173060 Chr.20: 18305765-18305765 on Build GRCh38
rs150451403 Chr.20: 18305794-18305794 on Build GRCh38
rs974485041 Chr.20: 18305975-18305975 on Build GRCh38
rs1026632281 Chr.20: 18305866-18305866 on Build GRCh38
rs745689658 Chr.20: 18305828-18305828 on Build GRCh38
rs373952348 Chr.20: 18305817-18305817 on Build GRCh38
rs574462572 Chr.20: 18305932-18305932 on Build GRCh38
rs889843204 Chr.20: 18305910-18305910 on Build GRCh38
rs541653884 Chr.20: 18305949-18305949 on Build GRCh38
rs1024523705 Chr.20: 18306033-18306034 on Build GRCh38
rs201136643 Chr.20: 18305770-18305770 on Build GRCh38
rs767687724 Chr.20: 18305725-18305725 on Build GRCh38
rs756713311 Chr.20: 18305761-18305761 on Build GRCh38
rs749667691 Chr.20: 18305635-18305635 on Build GRCh38
rs964418396 Chr.20: 18305962-18305962 on Build GRCh38
rs778651947 Chr.20: 18305641-18305641 on Build GRCh38
rs750459589 Chr.20: 18305746-18305746 on Build GRCh38
rs1049811090 Chr.20: 18305882-18305882 on Build GRCh38
rs754684019 Chr.20: 18305636-18305636 on Build GRCh38
rs778666417 Chr.20: 18305820-18305820 on Build GRCh38
rs752445972 Chr.20: 18305813-18305813 on Build GRCh38
rs1010903777 Chr.20: 18306021-18306021 on Build GRCh38
rs759265829 Chr.20: 18305706-18305706 on Build GRCh38
rs373623911 Chr.20: 18305643-18305643 on Build GRCh38
rs200815501 Chr.20: 18305842-18305842 on Build GRCh38
rs141542221 Chr.20: 18305701-18305701 on Build GRCh38
rs771652439 Chr.20: 18305650-18305650 on Build GRCh38

View Full Product Details