Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 25215598-25216102 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENTPD6
Entrez Gene ID
955
Gene Name
ectonucleoside triphosphate diphosphohydrolase 6 (putative)
Gene Aliases
CD39L2, IL-6SAG, IL6ST2, NTPDase-6, dJ738P15.3
UniGene
Hs.500375
Gene Chromosome Location
Chr.20: 25195670-25228075 on Build GRCh38
Transcripts
NM_001114089.3 NM_001247.4 NM_001317941.2 NM_001322378.1 NM_001322379.1 NM_001322380.1 NM_001322381.1 NM_001322382.1 NM_001322383.1 NM_001322384.1 NM_001322385.1 NM_001322386.1 NM_001322387.1 NM_001322388.1 NM_001322389.1 NM_001322390.1 NM_001322391.1 NM_001322392.1 NM_001322393.1 NM_001322394.1 NM_001322395.1 NM_001322396.1 NM_001322397.1 NM_001322398.1 XM_006723665.2 XM_011529396.1 XM_011529397.1 XM_017028129.1 XM_017028130.1 XM_017028131.1 XM_017028132.1 XM_017028133.1 XM_017028134.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5833938 c.680C>T p.S227L Chr.20: 25215682-25215682 on Build GRCh38
1025321 c.680C>T p.S227L Chr.20: 25215682-25215682 on Build GRCh38
443566 c.684G>A p.A228A Chr.20: 25215686-25215686 on Build GRCh38
5207051 c.684G>A p.A228A Chr.20: 25215686-25215686 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs953188249 Chr.20: 25215994-25215994 on Build GRCh38
rs576682810 Chr.20: 25216003-25216003 on Build GRCh38
rs1054664218 Chr.20: 25215894-25215894 on Build GRCh38
rs760819219 Chr.20: 25215727-25215727 on Build GRCh38
rs143784918 Chr.20: 25215743-25215743 on Build GRCh38
rs1049986688 Chr.20: 25216067-25216067 on Build GRCh38
rs1016037359 Chr.20: 25215738-25215738 on Build GRCh38
rs777289378 Chr.20: 25215682-25215682 on Build GRCh38
rs906008271 Chr.20: 25215874-25215874 on Build GRCh38
rs562705414 Chr.20: 25215942-25215942 on Build GRCh38
rs765182533 Chr.20: 25215669-25215669 on Build GRCh38
rs772949313 Chr.20: 25215678-25215678 on Build GRCh38
rs901460834 Chr.20: 25215668-25215668 on Build GRCh38
rs41308643 Chr.20: 25215780-25215780 on Build GRCh38
rs577335022 Chr.20: 25215887-25215887 on Build GRCh38
rs376583699 Chr.20: 25215712-25215712 on Build GRCh38
rs756988234 Chr.20: 25215686-25215686 on Build GRCh38
rs185710764 Chr.20: 25216035-25216035 on Build GRCh38
rs918566734 Chr.20: 25216036-25216036 on Build GRCh38
rs369102059 Chr.20: 25215718-25215718 on Build GRCh38
rs950113169 Chr.20: 25216042-25216042 on Build GRCh38
rs765773920 Chr.20: 25215665-25215666 on Build GRCh38
rs772249464 Chr.20: 25215721-25215721 on Build GRCh38
rs1010493831 Chr.20: 25215910-25215910 on Build GRCh38
rs539027811 Chr.20: 25215677-25215677 on Build GRCh38
rs927362174 Chr.20: 25216066-25216066 on Build GRCh38
rs776362126 Chr.20: 25215729-25215729 on Build GRCh38
rs181563953 Chr.20: 25215742-25215742 on Build GRCh38
rs371005572 Chr.20: 25215988-25215988 on Build GRCh38
rs752749642 Chr.20: 25215741-25215741 on Build GRCh38
rs984582767 Chr.20: 25215998-25215998 on Build GRCh38
rs997249627 Chr.20: 25215965-25215965 on Build GRCh38
rs1046247270 Chr.20: 25215812-25215812 on Build GRCh38
rs901517263 Chr.20: 25215948-25215948 on Build GRCh38
rs769261062 Chr.20: 25215710-25215710 on Build GRCh38
rs376346393 Chr.20: 25215759-25215759 on Build GRCh38
rs751039294 Chr.20: 25215754-25215754 on Build GRCh38
rs527864164 Chr.20: 25216044-25216044 on Build GRCh38
rs981494424 Chr.20: 25216051-25216051 on Build GRCh38
rs765129569 Chr.20: 25215737-25215737 on Build GRCh38
rs534328527 Chr.20: 25215781-25215781 on Build GRCh38
rs373739975 Chr.20: 25215758-25215758 on Build GRCh38
rs766105410 Chr.20: 25215638-25215638 on Build GRCh38
rs200386205 Chr.20: 25215909-25215909 on Build GRCh38
rs11087505 Chr.20: 25215884-25215884 on Build GRCh38
rs1036992243 Chr.20: 25215768-25215768 on Build GRCh38
rs765613149 Chr.20: 25216018-25216018 on Build GRCh38
rs979151935 Chr.20: 25215903-25215903 on Build GRCh38
rs753634160 Chr.20: 25215646-25215646 on Build GRCh38
rs1009898245 Chr.20: 25215639-25215639 on Build GRCh38
rs915000206 Chr.20: 25215930-25215930 on Build GRCh38
rs991389735 Chr.20: 25215850-25215850 on Build GRCh38
rs999048763 Chr.20: 25215657-25215657 on Build GRCh38
rs537039194 Chr.20: 25215763-25215777 on Build GRCh38
rs1019927231 Chr.20: 25215928-25215928 on Build GRCh38
rs6050447 Chr.20: 25216019-25216019 on Build GRCh38
rs780503838 Chr.20: 25215695-25215695 on Build GRCh38
rs148782311 Chr.20: 25215709-25215709 on Build GRCh38
rs199912392 Chr.20: 25215654-25215654 on Build GRCh38
rs755457841 Chr.20: 25215760-25215760 on Build GRCh38
rs773177477 Chr.20: 25215722-25215722 on Build GRCh38
rs1029143755 Chr.20: 25215977-25215977 on Build GRCh38
rs759378063 Chr.20: 25215731-25215731 on Build GRCh38
rs114620027 Chr.20: 25215664-25215664 on Build GRCh38
rs1016092567 Chr.20: 25216016-25216016 on Build GRCh38
rs746736920 Chr.20: 25215683-25215683 on Build GRCh38

View Full Product Details