Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 32073609-32074101 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HCK
Entrez Gene ID
3055
Gene Name
HCK proto-oncogene, Src family tyrosine kinase
Gene Aliases
JTK9, p59Hck, p61Hck
UniGene
Hs.655210
Gene Chromosome Location
Chr.20: 32052188-32101856 on Build GRCh38
Transcripts
NM_001172129.1 NM_001172130.1 NM_001172131.1 NM_001172132.1 NM_001172133.1 NM_002110.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6191045 c.208G>A p.E70K Chr.20: 32073757-32073757 on Build GRCh38
5596828 c.265G>A p.E89K Chr.20: 32073817-32073817 on Build GRCh38
5219997 c.219C>T p.H73H Chr.20: 32073771-32073771 on Build GRCh38
4976907 c.268G>A p.E90K Chr.20: 32073757-32073757 on Build GRCh38
6282110 c.227-4C>T p.? Chr.20: 32073712-32073712 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs922220222 Chr.20: 32073709-32073709 on Build GRCh38
rs996261383 Chr.20: 32073914-32073914 on Build GRCh38
rs867427724 Chr.20: 32073759-32073759 on Build GRCh38
rs781674698 Chr.20: 32073996-32073996 on Build GRCh38
rs765581370 Chr.20: 32073668-32073668 on Build GRCh38
rs749967035 Chr.20: 32073738-32073738 on Build GRCh38
rs988884449 Chr.20: 32073629-32073629 on Build GRCh38
rs771481116 Chr.20: 32073912-32073912 on Build GRCh38
rs1015147252 Chr.20: 32073701-32073701 on Build GRCh38
rs146350710 Chr.20: 32073735-32073735 on Build GRCh38
rs1005315409 Chr.20: 32074029-32074029 on Build GRCh38
rs952186396 Chr.20: 32073999-32073999 on Build GRCh38
rs556221818 Chr.20: 32073646-32073646 on Build GRCh38
rs60000242 Chr.20: 32074044-32074044 on Build GRCh38
rs528874049 Chr.20: 32073911-32073911 on Build GRCh38
rs952190952 Chr.20: 32073671-32073671 on Build GRCh38
rs370119506 Chr.20: 32073850-32073850 on Build GRCh38
rs373209595 Chr.20: 32073763-32073763 on Build GRCh38
rs55722810 Chr.20: 32073802-32073802 on Build GRCh38
rs765796296 Chr.20: 32073842-32073842 on Build GRCh38
rs143242352 Chr.20: 32074019-32074019 on Build GRCh38
rs6089166 Chr.20: 32073782-32073782 on Build GRCh38
rs763221373 Chr.20: 32073852-32073852 on Build GRCh38
rs908088371 Chr.20: 32074059-32074059 on Build GRCh38
rs757231454 Chr.20: 32073726-32073726 on Build GRCh38
rs375847090 Chr.20: 32073786-32073786 on Build GRCh38
rs975278085 Chr.20: 32073674-32073674 on Build GRCh38
rs532840131 Chr.20: 32074077-32074077 on Build GRCh38
rs866696007 Chr.20: 32073722-32073722 on Build GRCh38
rs900721488 Chr.20: 32073874-32073874 on Build GRCh38
rs1019046512 Chr.20: 32073883-32073883 on Build GRCh38
rs774199270 Chr.20: 32073864-32073864 on Build GRCh38
rs767291366 Chr.20: 32073736-32073736 on Build GRCh38
rs1049501447 Chr.20: 32073921-32073921 on Build GRCh38
rs932258170 Chr.20: 32073902-32073902 on Build GRCh38
rs1019417941 Chr.20: 32073645-32073645 on Build GRCh38
rs545049614 Chr.20: 32073899-32073899 on Build GRCh38
rs745558831 Chr.20: 32073757-32073757 on Build GRCh38
rs369481790 Chr.20: 32073683-32073683 on Build GRCh38
rs113353931 Chr.20: 32074074-32074074 on Build GRCh38
rs751505417 Chr.20: 32073713-32073713 on Build GRCh38
rs887801312 Chr.20: 32073952-32073952 on Build GRCh38
rs2227967 Chr.20: 32073771-32073771 on Build GRCh38
rs139451043 Chr.20: 32073841-32073841 on Build GRCh38
rs866179316 Chr.20: 32073715-32073715 on Build GRCh38
rs763940941 Chr.20: 32073702-32073702 on Build GRCh38
rs758750083 Chr.20: 32073769-32073769 on Build GRCh38
rs775829045 Chr.20: 32073682-32073682 on Build GRCh38
rs560436282 Chr.20: 32073756-32073756 on Build GRCh38
rs777232752 Chr.20: 32073793-32073793 on Build GRCh38
rs1054293490 Chr.20: 32073810-32073810 on Build GRCh38
rs775916901 Chr.20: 32073818-32073818 on Build GRCh38
rs944865299 Chr.20: 32073855-32073855 on Build GRCh38
rs961052001 Chr.20: 32074061-32074061 on Build GRCh38
rs372463603 Chr.20: 32073863-32073863 on Build GRCh38
rs141150727 Chr.20: 32073750-32073750 on Build GRCh38
rs958650426 Chr.20: 32073760-32073760 on Build GRCh38
rs909585818 Chr.20: 32073732-32073732 on Build GRCh38
rs771503154 Chr.20: 32073772-32073772 on Build GRCh38

View Full Product Details