Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33748807-33749293 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF341
Entrez Gene ID
84905
Gene Name
zinc finger protein 341
Gene Aliases
-
UniGene
Hs.516989
Gene Chromosome Location
Chr.20: 33731760-33792269 on Build GRCh38
Transcripts
NM_001282933.1 NM_001282935.1 NM_032819.4 NR_104259.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7132560 c.474delC p.P160fs*7 Chr.20: 33749057-33749057 on Build GRCh38
4001881 c.366G>A p.P122P Chr.20: 33748949-33748949 on Build GRCh38
2757841 c.474delC p.P160fs*7 Chr.20: 33749057-33749057 on Build GRCh38
4185721 c.405G>T p.L135F Chr.20: 33748988-33748988 on Build GRCh38
1025982 c.441C>A p.A147A Chr.20: 33749024-33749024 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs897007641 Chr.20: 33749163-33749163 on Build GRCh38
rs754115354 Chr.20: 33749020-33749020 on Build GRCh38
rs79173873 Chr.20: 33749088-33749088 on Build GRCh38
rs759808358 Chr.20: 33749104-33749104 on Build GRCh38
rs756500418 Chr.20: 33748911-33748911 on Build GRCh38
rs775264826 Chr.20: 33749064-33749064 on Build GRCh38
rs13040424 Chr.20: 33749027-33749027 on Build GRCh38
rs749585835 Chr.20: 33749178-33749178 on Build GRCh38
rs775319895 Chr.20: 33748994-33748994 on Build GRCh38
rs755166219 Chr.20: 33749001-33749002 on Build GRCh38
rs755111972 Chr.20: 33748924-33748924 on Build GRCh38
rs768179431 Chr.20: 33749210-33749210 on Build GRCh38
rs764948194 Chr.20: 33748890-33748890 on Build GRCh38
rs146485548 Chr.20: 33749003-33749003 on Build GRCh38
rs561564478 Chr.20: 33748830-33748830 on Build GRCh38
rs749011648 Chr.20: 33749067-33749067 on Build GRCh38
rs757578061 Chr.20: 33749056-33749056 on Build GRCh38
rs371747028 Chr.20: 33749050-33749050 on Build GRCh38
rs766716178 Chr.20: 33748915-33748915 on Build GRCh38
rs1024202680 Chr.20: 33748905-33748905 on Build GRCh38
rs938244075 Chr.20: 33749113-33749113 on Build GRCh38
rs768423474 Chr.20: 33749081-33749081 on Build GRCh38
rs373675665 Chr.20: 33749120-33749120 on Build GRCh38
rs752976706 Chr.20: 33749042-33749042 on Build GRCh38
rs1026936406 Chr.20: 33749215-33749215 on Build GRCh38
rs753064876 Chr.20: 33749114-33749114 on Build GRCh38
rs369641585 Chr.20: 33749106-33749106 on Build GRCh38
rs141017307 Chr.20: 33749062-33749062 on Build GRCh38
rs765425752 Chr.20: 33749031-33749031 on Build GRCh38
rs552057610 Chr.20: 33749239-33749239 on Build GRCh38
rs756224546 Chr.20: 33749142-33749142 on Build GRCh38
rs1033809826 Chr.20: 33748933-33748933 on Build GRCh38
rs774211258 Chr.20: 33749264-33749264 on Build GRCh38
rs1021071606 Chr.20: 33748876-33748877 on Build GRCh38
rs879939044 Chr.20: 33749243-33749243 on Build GRCh38
rs771314525 Chr.20: 33749093-33749093 on Build GRCh38
rs540687189 Chr.20: 33749007-33749007 on Build GRCh38
rs751552227 Chr.20: 33749124-33749124 on Build GRCh38
rs761033100 Chr.20: 33749013-33749013 on Build GRCh38
rs370690096 Chr.20: 33748891-33748891 on Build GRCh38
rs543565243 Chr.20: 33749212-33749212 on Build GRCh38
rs779995721 Chr.20: 33749171-33749171 on Build GRCh38
rs952743747 Chr.20: 33749223-33749223 on Build GRCh38
rs941092759 Chr.20: 33749134-33749134 on Build GRCh38
rs770796776 Chr.20: 33749256-33749256 on Build GRCh38
rs562569897 Chr.20: 33749258-33749258 on Build GRCh38
rs190646749 Chr.20: 33749179-33749179 on Build GRCh38
rs754231245 Chr.20: 33748917-33748917 on Build GRCh38
rs185795862 Chr.20: 33748970-33748970 on Build GRCh38
rs763103124 Chr.20: 33749117-33749117 on Build GRCh38
rs757092986 Chr.20: 33749057-33749057 on Build GRCh38
rs143444006 Chr.20: 33749051-33749051 on Build GRCh38
rs367894759 Chr.20: 33749015-33749015 on Build GRCh38
rs907114825 Chr.20: 33749207-33749207 on Build GRCh38
rs375824635 Chr.20: 33748907-33748907 on Build GRCh38
rs1055524359 Chr.20: 33749224-33749224 on Build GRCh38
rs766631136 Chr.20: 33748877-33748878 on Build GRCh38
rs368388744 Chr.20: 33748895-33748895 on Build GRCh38
rs978326782 Chr.20: 33749055-33749055 on Build GRCh38
rs374699114 Chr.20: 33748909-33748909 on Build GRCh38
rs770914623 Chr.20: 33748975-33748975 on Build GRCh38
rs745696771 Chr.20: 33748979-33748979 on Build GRCh38
rs759054039 Chr.20: 33748883-33748883 on Build GRCh38
rs746828395 Chr.20: 33749242-33749242 on Build GRCh38
rs142373708 Chr.20: 33748903-33748903 on Build GRCh38
rs777755812 Chr.20: 33749196-33749196 on Build GRCh38
rs75946243 Chr.20: 33748949-33748949 on Build GRCh38
rs1027046592 Chr.20: 33749000-33749000 on Build GRCh38
rs941242070 Chr.20: 33748862-33748862 on Build GRCh38
rs758469627 Chr.20: 33749049-33749049 on Build GRCh38
rs753864900 Chr.20: 33748891-33748892 on Build GRCh38
rs777682620 Chr.20: 33748967-33748967 on Build GRCh38
rs758512398 Chr.20: 33748965-33748965 on Build GRCh38
rs777247152 Chr.20: 33749099-33749099 on Build GRCh38
rs929764190 Chr.20: 33749167-33749167 on Build GRCh38
rs889493762 Chr.20: 33748856-33748856 on Build GRCh38
rs895256811 Chr.20: 33749235-33749235 on Build GRCh38
rs182640233 Chr.20: 33748855-33748855 on Build GRCh38
rs368873311 Chr.20: 33748930-33748930 on Build GRCh38
rs745754781 Chr.20: 33748887-33748887 on Build GRCh38
rs773954021 Chr.20: 33749085-33749085 on Build GRCh38
rs376356735 Chr.20: 33748875-33748875 on Build GRCh38
rs1022402557 Chr.20: 33749022-33749022 on Build GRCh38
rs776223258 Chr.20: 33748978-33748978 on Build GRCh38
rs769289143 Chr.20: 33748992-33748992 on Build GRCh38
rs755413338 Chr.20: 33749029-33749029 on Build GRCh38

View Full Product Details