Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 43594509-43594927 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFT52
Entrez Gene ID
51098
Gene Name
intraflagellar transport 52
Gene Aliases
C20orf9, CGI-53, NGD2, NGD5
UniGene
Hs.444332
Gene Chromosome Location
Chr.20: 43590613-43647296 on Build GRCh38
Transcripts
NM_001303458.2 NM_001303459.2 NM_001323578.1 NM_001323579.1 NM_001323580.1 NM_001323581.1 NM_016004.4 XM_017027863.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1662238 c.38C>A p.A13D Chr.20: 43594736-43594736 on Build GRCh38
1026986 c.28C>A p.L10I Chr.20: 43594726-43594726 on Build GRCh38
4820135 c.117G>C p.Q39H Chr.20: 43594815-43594815 on Build GRCh38
1026987 c.41A>G p.Y14C Chr.20: 43594739-43594739 on Build GRCh38
1026988 c.71G>A p.G24D Chr.20: 43594769-43594769 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1008437187 Chr.20: 43594792-43594792 on Build GRCh38
rs761611965 Chr.20: 43594777-43594777 on Build GRCh38
rs761525993 Chr.20: 43594859-43594859 on Build GRCh38
rs774155176 Chr.20: 43594819-43594820 on Build GRCh38
rs758333521 Chr.20: 43594788-43594788 on Build GRCh38
rs777811646 Chr.20: 43594826-43594826 on Build GRCh38
rs779866421 Chr.20: 43594669-43594669 on Build GRCh38
rs923565458 Chr.20: 43594587-43594587 on Build GRCh38
rs749992688 Chr.20: 43594782-43594782 on Build GRCh38
rs563044236 Chr.20: 43594637-43594637 on Build GRCh38
rs77556214 Chr.20: 43594846-43594846 on Build GRCh38
rs531451513 Chr.20: 43594639-43594639 on Build GRCh38
rs750368811 Chr.20: 43594655-43594655 on Build GRCh38
rs771394377 Chr.20: 43594723-43594723 on Build GRCh38
rs560615744 Chr.20: 43594577-43594577 on Build GRCh38
rs145902295 Chr.20: 43594588-43594588 on Build GRCh38
rs749320242 Chr.20: 43594828-43594828 on Build GRCh38
rs377307012 Chr.20: 43594763-43594763 on Build GRCh38
rs924235751 Chr.20: 43594547-43594547 on Build GRCh38
rs771076234 Chr.20: 43594829-43594829 on Build GRCh38
rs978131938 Chr.20: 43594870-43594870 on Build GRCh38
rs750646902 Chr.20: 43594601-43594601 on Build GRCh38
rs138787768 Chr.20: 43594710-43594710 on Build GRCh38
rs769985111 Chr.20: 43594712-43594712 on Build GRCh38
rs369942659 Chr.20: 43594780-43594780 on Build GRCh38
rs6030976 Chr.20: 43594635-43594635 on Build GRCh38
rs547710015 Chr.20: 43594795-43594795 on Build GRCh38
rs746827895 Chr.20: 43594674-43594674 on Build GRCh38
rs1038152199 Chr.20: 43594643-43594643 on Build GRCh38
rs997843416 Chr.20: 43594625-43594651 on Build GRCh38
rs184213301 Chr.20: 43594822-43594822 on Build GRCh38
rs370368058 Chr.20: 43594677-43594677 on Build GRCh38
rs1056199036 Chr.20: 43594600-43594600 on Build GRCh38
rs776361903 Chr.20: 43594773-43594773 on Build GRCh38
rs202221752 Chr.20: 43594815-43594815 on Build GRCh38
rs373213594 Chr.20: 43594682-43594682 on Build GRCh38
rs569475323 Chr.20: 43594863-43594863 on Build GRCh38
rs1047392040 Chr.20: 43594739-43594739 on Build GRCh38
rs749767548 Chr.20: 43594719-43594719 on Build GRCh38
rs751521218 Chr.20: 43594791-43594791 on Build GRCh38
rs567232435 Chr.20: 43594802-43594802 on Build GRCh38
rs751810076 Chr.20: 43594899-43594899 on Build GRCh38
rs772307855 Chr.20: 43594854-43594854 on Build GRCh38
rs747004115 Chr.20: 43594777-43594778 on Build GRCh38
rs1056645390 Chr.20: 43594569-43594569 on Build GRCh38
rs148888998 Chr.20: 43594714-43594714 on Build GRCh38
rs746395983 Chr.20: 43594832-43594832 on Build GRCh38
rs181146490 Chr.20: 43594558-43594558 on Build GRCh38
rs776059878 Chr.20: 43594857-43594857 on Build GRCh38
rs6065631 Chr.20: 43594737-43594737 on Build GRCh38
rs779218037 Chr.20: 43594830-43594830 on Build GRCh38
rs758476333 Chr.20: 43594659-43594659 on Build GRCh38
rs748384867 Chr.20: 43594697-43594697 on Build GRCh38
rs773019549 Chr.20: 43594867-43594867 on Build GRCh38
rs888027620 Chr.20: 43594684-43594684 on Build GRCh38
rs766834545 Chr.20: 43594741-43594741 on Build GRCh38
rs527285373 Chr.20: 43594790-43594790 on Build GRCh38
rs915162091 Chr.20: 43594540-43594540 on Build GRCh38

View Full Product Details