Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 45221621-45222120 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEMG2
Entrez Gene ID
6407
Gene Name
semenogelin II
Gene Aliases
SGII
UniGene
Hs.537218
Gene Chromosome Location
Chr.20: 45221369-45224458 on Build GRCh38
Transcripts
NM_003008.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3546941 c.80G>A p.G27E Chr.20: 45221712-45221712 on Build GRCh38
6525854 c.267C>A p.A89A Chr.20: 45221899-45221899 on Build GRCh38
5563297 c.436C>G p.Q146E Chr.20: 45222068-45222068 on Build GRCh38
3841105 c.399C>T p.G133G Chr.20: 45222031-45222031 on Build GRCh38
3546945 c.268C>T p.L90L Chr.20: 45221900-45221900 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2233898 Chr.20: 45221884-45221884 on Build GRCh38
rs147702753 Chr.20: 45221893-45221893 on Build GRCh38
rs747684917 Chr.20: 45221721-45221721 on Build GRCh38
rs745679734 Chr.20: 45221898-45221898 on Build GRCh38
rs767767129 Chr.20: 45221755-45221755 on Build GRCh38
rs769207275 Chr.20: 45222070-45222070 on Build GRCh38
rs770704818 Chr.20: 45221912-45221919 on Build GRCh38
rs765107670 Chr.20: 45221999-45221999 on Build GRCh38
rs61729999 Chr.20: 45221841-45221841 on Build GRCh38
rs374565015 Chr.20: 45221905-45221905 on Build GRCh38
rs756635939 Chr.20: 45221676-45221676 on Build GRCh38
rs1057443707 Chr.20: 45222075-45222075 on Build GRCh38
rs374141494 Chr.20: 45221685-45221685 on Build GRCh38
rs1041604900 Chr.20: 45221813-45221813 on Build GRCh38
rs781418524 Chr.20: 45222091-45222091 on Build GRCh38
rs183488917 Chr.20: 45221804-45221804 on Build GRCh38
rs557082400 Chr.20: 45221866-45221866 on Build GRCh38
rs779638348 Chr.20: 45221911-45221911 on Build GRCh38
rs111412692 Chr.20: 45221734-45221734 on Build GRCh38
rs771324877 Chr.20: 45222031-45222031 on Build GRCh38
rs770292316 Chr.20: 45221943-45221943 on Build GRCh38
rs760458546 Chr.20: 45221989-45221989 on Build GRCh38
rs369538412 Chr.20: 45221904-45221904 on Build GRCh38
rs780637566 Chr.20: 45221720-45221720 on Build GRCh38
rs368705905 Chr.20: 45221698-45221698 on Build GRCh38
rs775445924 Chr.20: 45221758-45221758 on Build GRCh38
rs753381619 Chr.20: 45221887-45221887 on Build GRCh38
rs995239738 Chr.20: 45221902-45221902 on Build GRCh38
rs751365055 Chr.20: 45222083-45222083 on Build GRCh38
rs765630776 Chr.20: 45221781-45221781 on Build GRCh38
rs371375093 Chr.20: 45221853-45221853 on Build GRCh38
rs543229204 Chr.20: 45221831-45221831 on Build GRCh38
rs779077576 Chr.20: 45221835-45221835 on Build GRCh38
rs199893935 Chr.20: 45221827-45221827 on Build GRCh38
rs759443747 Chr.20: 45222050-45222051 on Build GRCh38
rs763343653 Chr.20: 45221968-45221968 on Build GRCh38
rs200326613 Chr.20: 45221735-45221735 on Build GRCh38
rs774788806 Chr.20: 45222038-45222038 on Build GRCh38
rs776660955 Chr.20: 45221857-45221857 on Build GRCh38
rs1040086710 Chr.20: 45222074-45222074 on Build GRCh38
rs188329542 Chr.20: 45221863-45221863 on Build GRCh38
rs200846745 Chr.20: 45221823-45221823 on Build GRCh38
rs781328840 Chr.20: 45222018-45222018 on Build GRCh38
rs913966150 Chr.20: 45221710-45221710 on Build GRCh38
rs202124576 Chr.20: 45221927-45221927 on Build GRCh38
rs991161841 Chr.20: 45221942-45221942 on Build GRCh38
rs758239329 Chr.20: 45222003-45222003 on Build GRCh38
rs773732268 Chr.20: 45221950-45221950 on Build GRCh38
rs552609548 Chr.20: 45221688-45221688 on Build GRCh38
rs140134573 Chr.20: 45221810-45221810 on Build GRCh38
rs745427048 Chr.20: 45221908-45221908 on Build GRCh38
rs754730269 Chr.20: 45222084-45222084 on Build GRCh38
rs766804555 Chr.20: 45221975-45221975 on Build GRCh38
rs780842367 Chr.20: 45221799-45221799 on Build GRCh38
rs149777388 Chr.20: 45221910-45221910 on Build GRCh38
rs563034271 Chr.20: 45221871-45221871 on Build GRCh38
rs769819800 Chr.20: 45222028-45222028 on Build GRCh38
rs867576271 Chr.20: 45221870-45221870 on Build GRCh38
rs755599676 Chr.20: 45221802-45221802 on Build GRCh38
rs112798702 Chr.20: 45221939-45221939 on Build GRCh38
rs369152303 Chr.20: 45221882-45221882 on Build GRCh38
rs374211653 Chr.20: 45221680-45221680 on Build GRCh38
rs372121807 Chr.20: 45221716-45221716 on Build GRCh38
rs779966456 Chr.20: 45222015-45222015 on Build GRCh38
rs770984415 Chr.20: 45221814-45221814 on Build GRCh38
rs989568692 Chr.20: 45221657-45221657 on Build GRCh38
rs766173788 Chr.20: 45222078-45222078 on Build GRCh38
rs773219709 Chr.20: 45221730-45221730 on Build GRCh38
rs773132069 Chr.20: 45222029-45222029 on Build GRCh38
rs958045951 Chr.20: 45221856-45221856 on Build GRCh38
rs755688462 Chr.20: 45221883-45221883 on Build GRCh38
rs759912660 Chr.20: 45222042-45222042 on Build GRCh38
rs2233896 Chr.20: 45221759-45221759 on Build GRCh38
rs986705429 Chr.20: 45221677-45221677 on Build GRCh38
rs750424738 Chr.20: 45222002-45222002 on Build GRCh38
rs778926230 Chr.20: 45221897-45221897 on Build GRCh38
rs192628433 Chr.20: 45222016-45222016 on Build GRCh38
rs963871529 Chr.20: 45221746-45221746 on Build GRCh38
rs148802229 Chr.20: 45221750-45221750 on Build GRCh38
rs759739206 Chr.20: 45221754-45221754 on Build GRCh38
rs145599636 Chr.20: 45222062-45222062 on Build GRCh38
rs776372041 Chr.20: 45222047-45222047 on Build GRCh38
rs760693537 Chr.20: 45221847-45221847 on Build GRCh38
rs746714934 Chr.20: 45221712-45221712 on Build GRCh38
rs1019490388 Chr.20: 45221949-45221949 on Build GRCh38
rs764615350 Chr.20: 45221764-45221764 on Build GRCh38
rs1010884801 Chr.20: 45221878-45221878 on Build GRCh38
rs374489769 Chr.20: 45221778-45221778 on Build GRCh38
rs761534127 Chr.20: 45222050-45222050 on Build GRCh38
rs776587049 Chr.20: 45221964-45221966 on Build GRCh38
rs758419957 Chr.20: 45222072-45222072 on Build GRCh38
rs777437827 Chr.20: 45221790-45221790 on Build GRCh38
rs2233897 Chr.20: 45221801-45221801 on Build GRCh38
rs762375167 Chr.20: 45221861-45221861 on Build GRCh38
rs377178712 Chr.20: 45222095-45222095 on Build GRCh38
rs140923429 Chr.20: 45221909-45221909 on Build GRCh38
rs750786246 Chr.20: 45221783-45221783 on Build GRCh38
rs776753000 Chr.20: 45221920-45221920 on Build GRCh38
rs768885979 Chr.20: 45221708-45221708 on Build GRCh38
rs149252982 Chr.20: 45221725-45221725 on Build GRCh38
rs763870015 Chr.20: 45222045-45222045 on Build GRCh38
rs775739835 Chr.20: 45221842-45221842 on Build GRCh38
rs879008073 Chr.20: 45221935-45221935 on Build GRCh38
rs753403910 Chr.20: 45221998-45221998 on Build GRCh38
rs553011094 Chr.20: 45221828-45221828 on Build GRCh38
rs746752132 Chr.20: 45221799-45221800 on Build GRCh38
rs754648794 Chr.20: 45221795-45221795 on Build GRCh38
rs997518242 Chr.20: 45221844-45221844 on Build GRCh38
rs774570826 Chr.20: 45221976-45221976 on Build GRCh38
rs367855275 Chr.20: 45221797-45221797 on Build GRCh38
rs754296353 Chr.20: 45221765-45221765 on Build GRCh38
rs749870633 Chr.20: 45222030-45222030 on Build GRCh38
rs763781475 Chr.20: 45221997-45221997 on Build GRCh38

View Full Product Details