Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 45899644-45900080 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLTP
Entrez Gene ID
5360
Gene Name
phospholipid transfer protein
Gene Aliases
BPIFE, HDLCQ9
UniGene
Hs.439312
Gene Chromosome Location
Chr.20: 45898620-45912364 on Build GRCh38
Transcripts
NM_001242920.1 NM_001242921.1 NM_006227.3 NM_182676.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6868431 c.1286C>A p.P429Q Chr.20: 45899678-45899678 on Build GRCh38
3405155 c.1304A>G p.K435R Chr.20: 45899660-45899660 on Build GRCh38
6030070 c.1241G>A p.R414Q Chr.20: 45899873-45899873 on Build GRCh38
6033310 c.1176-4G>A p.? Chr.20: 45899882-45899882 on Build GRCh38
6033311 c.1236-4G>A p.? Chr.20: 45899882-45899882 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760749486 Chr.20: 45899735-45899735 on Build GRCh38
rs56190989 Chr.20: 45899740-45899740 on Build GRCh38
rs893698911 Chr.20: 45900030-45900030 on Build GRCh38
rs747650570 Chr.20: 45899711-45899711 on Build GRCh38
rs554922207 Chr.20: 45899812-45899813 on Build GRCh38
rs201623225 Chr.20: 45899713-45899713 on Build GRCh38
rs754126168 Chr.20: 45899694-45899694 on Build GRCh38
rs541697748 Chr.20: 45899773-45899773 on Build GRCh38
rs1009547179 Chr.20: 45899743-45899743 on Build GRCh38
rs911441944 Chr.20: 45899830-45899830 on Build GRCh38
rs777343906 Chr.20: 45899714-45899714 on Build GRCh38
rs538015515 Chr.20: 45899747-45899747 on Build GRCh38
rs751335056 Chr.20: 45899815-45899815 on Build GRCh38
rs377024064 Chr.20: 45899697-45899697 on Build GRCh38
rs765603558 Chr.20: 45899812-45899812 on Build GRCh38
rs935222584 Chr.20: 45899968-45899968 on Build GRCh38
rs190186546 Chr.20: 45899703-45899703 on Build GRCh38
rs553793980 Chr.20: 45899774-45899774 on Build GRCh38
rs779676867 Chr.20: 45899788-45899795 on Build GRCh38
rs899661613 Chr.20: 45899690-45899690 on Build GRCh38
rs375620833 Chr.20: 45899907-45899907 on Build GRCh38
rs774758139 Chr.20: 45899719-45899719 on Build GRCh38
rs544066707 Chr.20: 45899920-45899920 on Build GRCh38
rs35033188 Chr.20: 45899963-45899963 on Build GRCh38
rs778463054 Chr.20: 45899882-45899882 on Build GRCh38
rs756114945 Chr.20: 45899825-45899825 on Build GRCh38
rs755942015 Chr.20: 45899691-45899691 on Build GRCh38
rs773233220 Chr.20: 45899676-45899676 on Build GRCh38
rs776915914 Chr.20: 45899800-45899800 on Build GRCh38
rs776827399 Chr.20: 45899926-45899926 on Build GRCh38
rs772336918 Chr.20: 45899906-45899906 on Build GRCh38
rs373792156 Chr.20: 45899730-45899730 on Build GRCh38
rs767436184 Chr.20: 45899820-45899820 on Build GRCh38
rs374467508 Chr.20: 45899716-45899716 on Build GRCh38
rs1056571703 Chr.20: 45900023-45900023 on Build GRCh38
rs572392262 Chr.20: 45899776-45899776 on Build GRCh38
rs923862113 Chr.20: 45899961-45899961 on Build GRCh38
rs932482107 Chr.20: 45899975-45899975 on Build GRCh38
rs532692364 Chr.20: 45900052-45900052 on Build GRCh38
rs545986794 Chr.20: 45899883-45899883 on Build GRCh38
rs754757208 Chr.20: 45899816-45899816 on Build GRCh38
rs917572807 Chr.20: 45900033-45900033 on Build GRCh38
rs756874726 Chr.20: 45899874-45899874 on Build GRCh38
rs1053721062 Chr.20: 45900022-45900022 on Build GRCh38
rs982376518 Chr.20: 45899843-45899843 on Build GRCh38
rs766214064 Chr.20: 45899850-45899850 on Build GRCh38
rs769342957 Chr.20: 45899921-45899921 on Build GRCh38
rs368167633 Chr.20: 45899799-45899799 on Build GRCh38
rs921785898 Chr.20: 45899904-45899904 on Build GRCh38
rs1019611548 Chr.20: 45899787-45899787 on Build GRCh38
rs202134183 Chr.20: 45899687-45899687 on Build GRCh38
rs758199674 Chr.20: 45899696-45899696 on Build GRCh38
rs752585502 Chr.20: 45899824-45899824 on Build GRCh38
rs754919038 Chr.20: 45899702-45899702 on Build GRCh38
rs201369456 Chr.20: 45899729-45899729 on Build GRCh38
rs73306280 Chr.20: 45899772-45899772 on Build GRCh38
rs764362720 Chr.20: 45899689-45899689 on Build GRCh38
rs762112381 Chr.20: 45899811-45899811 on Build GRCh38
rs982306021 Chr.20: 45899752-45899752 on Build GRCh38
rs371226806 Chr.20: 45899842-45899842 on Build GRCh38
rs113023458 Chr.20: 45899835-45899835 on Build GRCh38
rs1040789436 Chr.20: 45899915-45899915 on Build GRCh38
rs866711127 Chr.20: 45899753-45899753 on Build GRCh38
rs56235473 Chr.20: 45899749-45899749 on Build GRCh38
rs777255847 Chr.20: 45899834-45899834 on Build GRCh38
rs868811892 Chr.20: 45899810-45899810 on Build GRCh38

View Full Product Details