Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 45949231-45949733 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF335
Entrez Gene ID
63925
Gene Name
zinc finger protein 335
Gene Aliases
MCPH10, NIF-1, NIF1, NIF2
UniGene
Hs.174193
Gene Chromosome Location
Chr.20: 45948653-45972910 on Build GRCh38
Transcripts
NM_022095.3 XM_005260504.4 XM_017028012.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5543645 c.3800C>T p.P1267L Chr.20: 45949352-45949352 on Build GRCh38
7179504 c.3736G>A p.E1246K Chr.20: 45949502-45949502 on Build GRCh38
4491375 c.3799C>T p.P1267S Chr.20: 45949353-45949353 on Build GRCh38
7261285 c.3670-9_3670-8insC p.? Chr.20: 45949576-45949577 on Build GRCh38
5880907 c.3796G>T p.A1266S Chr.20: 45949356-45949356 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778980849 Chr.20: 45949327-45949327 on Build GRCh38
rs774730215 Chr.20: 45949285-45949286 on Build GRCh38
rs780397810 Chr.20: 45949341-45949341 on Build GRCh38
rs569368236 Chr.20: 45949516-45949516 on Build GRCh38
rs1039607140 Chr.20: 45949445-45949445 on Build GRCh38
rs866581432 Chr.20: 45949430-45949430 on Build GRCh38
rs200177323 Chr.20: 45949425-45949425 on Build GRCh38
rs369305017 Chr.20: 45949602-45949602 on Build GRCh38
rs376567868 Chr.20: 45949586-45949586 on Build GRCh38
rs928025646 Chr.20: 45949265-45949265 on Build GRCh38
rs764608484 Chr.20: 45949453-45949454 on Build GRCh38
rs771181192 Chr.20: 45949577-45949577 on Build GRCh38
rs902538647 Chr.20: 45949563-45949563 on Build GRCh38
rs778695885 Chr.20: 45949466-45949466 on Build GRCh38
rs775007394 Chr.20: 45949487-45949487 on Build GRCh38
rs754403551 Chr.20: 45949609-45949609 on Build GRCh38
rs746986496 Chr.20: 45949606-45949606 on Build GRCh38
rs763575505 Chr.20: 45949518-45949518 on Build GRCh38
rs777558948 Chr.20: 45949456-45949456 on Build GRCh38
rs775095229 Chr.20: 45949390-45949390 on Build GRCh38
rs745927171 Chr.20: 45949328-45949328 on Build GRCh38
rs754608119 Chr.20: 45949258-45949258 on Build GRCh38
rs779476380 Chr.20: 45949577-45949577 on Build GRCh38
rs747180003 Chr.20: 45949434-45949434 on Build GRCh38
rs759668816 Chr.20: 45949589-45949589 on Build GRCh38
rs773066150 Chr.20: 45949619-45949619 on Build GRCh38
rs773162696 Chr.20: 45949355-45949355 on Build GRCh38
rs768201972 Chr.20: 45949578-45949578 on Build GRCh38
rs528955337 Chr.20: 45949694-45949694 on Build GRCh38
rs375133385 Chr.20: 45949469-45949469 on Build GRCh38
rs552673947 Chr.20: 45949298-45949298 on Build GRCh38
rs41280270 Chr.20: 45949411-45949411 on Build GRCh38
rs762059104 Chr.20: 45949514-45949514 on Build GRCh38
rs769759244 Chr.20: 45949289-45949289 on Build GRCh38
rs750227118 Chr.20: 45949597-45949597 on Build GRCh38
rs1044568923 Chr.20: 45949435-45949435 on Build GRCh38
rs775916857 Chr.20: 45949286-45949286 on Build GRCh38
rs917223432 Chr.20: 45949676-45949676 on Build GRCh38
rs367545283 Chr.20: 45949317-45949317 on Build GRCh38
rs748617576 Chr.20: 45949317-45949317 on Build GRCh38
rs372007244 Chr.20: 45949436-45949436 on Build GRCh38
rs1017435940 Chr.20: 45949284-45949284 on Build GRCh38
rs765863504 Chr.20: 45949294-45949294 on Build GRCh38
rs773225126 Chr.20: 45949431-45949432 on Build GRCh38
rs767677365 Chr.20: 45949591-45949591 on Build GRCh38
rs966644687 Chr.20: 45949259-45949259 on Build GRCh38
rs144047985 Chr.20: 45949679-45949679 on Build GRCh38
rs770002166 Chr.20: 45949582-45949582 on Build GRCh38
rs1037588501 Chr.20: 45949366-45949366 on Build GRCh38
rs766012842 Chr.20: 45949370-45949370 on Build GRCh38
rs778894170 Chr.20: 45949279-45949279 on Build GRCh38
rs778802942 Chr.20: 45949555-45949555 on Build GRCh38
rs988849625 Chr.20: 45949657-45949657 on Build GRCh38
rs529342729 Chr.20: 45949305-45949305 on Build GRCh38
rs920482240 Chr.20: 45949656-45949656 on Build GRCh38
rs113958814 Chr.20: 45949352-45949352 on Build GRCh38
rs758561068 Chr.20: 45949329-45949329 on Build GRCh38
rs762829539 Chr.20: 45949292-45949292 on Build GRCh38
rs576029166 Chr.20: 45949616-45949616 on Build GRCh38
rs890193215 Chr.20: 45949427-45949427 on Build GRCh38
rs927476321 Chr.20: 45949706-45949706 on Build GRCh38
rs993156025 Chr.20: 45949566-45949566 on Build GRCh38
rs772185509 Chr.20: 45949282-45949282 on Build GRCh38
rs200761168 Chr.20: 45949618-45949618 on Build GRCh38
rs758519489 Chr.20: 45949429-45949429 on Build GRCh38
rs541241560 Chr.20: 45949701-45949701 on Build GRCh38
rs1038556619 Chr.20: 45949476-45949476 on Build GRCh38
rs754335454 Chr.20: 45949322-45949322 on Build GRCh38
rs764674487 Chr.20: 45949260-45949260 on Build GRCh38
rs762561338 Chr.20: 45949309-45949309 on Build GRCh38
rs928931258 Chr.20: 45949406-45949406 on Build GRCh38
rs1035092542 Chr.20: 45949584-45949584 on Build GRCh38
rs191147826 Chr.20: 45949262-45949262 on Build GRCh38
rs756972556 Chr.20: 45949543-45949543 on Build GRCh38
rs201440041 Chr.20: 45949576-45949577 on Build GRCh38
rs760719803 Chr.20: 45949573-45949575 on Build GRCh38
rs768290060 Chr.20: 45949508-45949508 on Build GRCh38
rs747696164 Chr.20: 45949617-45949617 on Build GRCh38
rs750984765 Chr.20: 45949424-45949424 on Build GRCh38
rs993052154 Chr.20: 45949296-45949296 on Build GRCh38
rs959893772 Chr.20: 45949268-45949268 on Build GRCh38
rs765646672 Chr.20: 45949420-45949420 on Build GRCh38
rs973746718 Chr.20: 45949645-45949645 on Build GRCh38
rs1047831462 Chr.20: 45949568-45949568 on Build GRCh38
rs980119533 Chr.20: 45949688-45949688 on Build GRCh38
rs746041587 Chr.20: 45949281-45949281 on Build GRCh38
rs752824823 Chr.20: 45949592-45949592 on Build GRCh38
rs752406575 Chr.20: 45949316-45949316 on Build GRCh38
rs760267226 Chr.20: 45949503-45949503 on Build GRCh38
rs745862092 Chr.20: 45949468-45949468 on Build GRCh38
rs745759118 Chr.20: 45949572-45949572 on Build GRCh38
rs764886503 Chr.20: 45949595-45949595 on Build GRCh38
rs749847885 Chr.20: 45949276-45949276 on Build GRCh38
rs747273985 Chr.20: 45949351-45949351 on Build GRCh38
rs765670908 Chr.20: 45949515-45949515 on Build GRCh38
rs771016780 Chr.20: 45949277-45949277 on Build GRCh38
rs772015167 Chr.20: 45949576-45949576 on Build GRCh38
rs767155037 Chr.20: 45949315-45949315 on Build GRCh38
rs747940466 Chr.20: 45949287-45949287 on Build GRCh38
rs780309952 Chr.20: 45949431-45949431 on Build GRCh38
rs749033379 Chr.20: 45949459-45949459 on Build GRCh38
rs753015159 Chr.20: 45949542-45949542 on Build GRCh38
rs753066883 Chr.20: 45949263-45949263 on Build GRCh38
rs373445422 Chr.20: 45949583-45949583 on Build GRCh38
rs757703982 Chr.20: 45949414-45949414 on Build GRCh38
rs751694123 Chr.20: 45949523-45949523 on Build GRCh38
rs770820310 Chr.20: 45949461-45949461 on Build GRCh38
rs770779104 Chr.20: 45949377-45949377 on Build GRCh38
rs202015831 Chr.20: 45949255-45949255 on Build GRCh38
rs772168361 Chr.20: 45949429-45949444 on Build GRCh38
rs150750750 Chr.20: 45949257-45949258 on Build GRCh38
rs767957907 Chr.20: 45949438-45949438 on Build GRCh38
rs983095119 Chr.20: 45949674-45949674 on Build GRCh38
rs754170727 Chr.20: 45949413-45949413 on Build GRCh38
rs755263750 Chr.20: 45949453-45949453 on Build GRCh38
rs376616764 Chr.20: 45949579-45949579 on Build GRCh38
rs772754282 Chr.20: 45949301-45949301 on Build GRCh38
rs778241579 Chr.20: 45949274-45949274 on Build GRCh38
rs928981956 Chr.20: 45949668-45949668 on Build GRCh38
rs755176005 Chr.20: 45949524-45949524 on Build GRCh38
rs373911621 Chr.20: 45949387-45949387 on Build GRCh38
rs766508807 Chr.20: 45949521-45949521 on Build GRCh38
rs781257414 Chr.20: 45949532-45949532 on Build GRCh38
rs773252152 Chr.20: 45949290-45949290 on Build GRCh38
rs941505330 Chr.20: 45949708-45949708 on Build GRCh38
rs776399068 Chr.20: 45949512-45949512 on Build GRCh38
rs752170069 Chr.20: 45949587-45949587 on Build GRCh38
rs760353441 Chr.20: 45949395-45949395 on Build GRCh38
rs574409465 Chr.20: 45949324-45949324 on Build GRCh38
rs756538206 Chr.20: 45949269-45949269 on Build GRCh38
rs749190523 Chr.20: 45949365-45949365 on Build GRCh38

View Full Product Details