Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 50574425-50574928 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPN1
Entrez Gene ID
5770
Gene Name
protein tyrosine phosphatase, non-receptor type 1
Gene Aliases
PTP1B
UniGene
Hs.417549
Gene Chromosome Location
Chr.20: 50510321-50584762 on Build GRCh38
Transcripts
NM_001278618.1 NM_002827.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1242728 c.466C>T p.R156* Chr.20: 50574628-50574628 on Build GRCh38
7327212 c.429A>G p.T143T Chr.20: 50574591-50574591 on Build GRCh38
3423721 c.467G>A p.R156Q Chr.20: 50574629-50574629 on Build GRCh38
5921747 c.355-4C>T p.? Chr.20: 50574513-50574513 on Build GRCh38
7101816 c.366A>G p.A122A Chr.20: 50574528-50574528 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371360165 Chr.20: 50574525-50574525 on Build GRCh38
rs1007089931 Chr.20: 50574449-50574449 on Build GRCh38
rs979228261 Chr.20: 50574457-50574457 on Build GRCh38
rs146302578 Chr.20: 50574549-50574549 on Build GRCh38
rs751834185 Chr.20: 50574661-50574661 on Build GRCh38
rs745360487 Chr.20: 50574504-50574504 on Build GRCh38
rs753862735 Chr.20: 50574472-50574472 on Build GRCh38
rs201436540 Chr.20: 50574470-50574470 on Build GRCh38
rs1037171425 Chr.20: 50574839-50574839 on Build GRCh38
rs1023603673 Chr.20: 50574855-50574855 on Build GRCh38
rs6020609 Chr.20: 50574695-50574695 on Build GRCh38
rs142739795 Chr.20: 50574472-50574473 on Build GRCh38
rs117274412 Chr.20: 50574878-50574878 on Build GRCh38
rs758003648 Chr.20: 50574704-50574704 on Build GRCh38
rs139368059 Chr.20: 50574564-50574564 on Build GRCh38
rs776030790 Chr.20: 50574644-50574644 on Build GRCh38
rs562088133 Chr.20: 50574881-50574881 on Build GRCh38
rs541938865 Chr.20: 50574766-50574766 on Build GRCh38
rs759051460 Chr.20: 50574669-50574676 on Build GRCh38
rs753025772 Chr.20: 50574841-50574841 on Build GRCh38
rs200476108 Chr.20: 50574871-50574871 on Build GRCh38
rs928696534 Chr.20: 50574812-50574812 on Build GRCh38
rs753571453 Chr.20: 50574686-50574686 on Build GRCh38
rs763726072 Chr.20: 50574890-50574890 on Build GRCh38
rs567267722 Chr.20: 50574481-50574481 on Build GRCh38
rs778633975 Chr.20: 50574488-50574488 on Build GRCh38
rs144145190 Chr.20: 50574576-50574576 on Build GRCh38
rs773648972 Chr.20: 50574621-50574621 on Build GRCh38
rs1885177 Chr.20: 50574691-50574691 on Build GRCh38
rs573995433 Chr.20: 50574902-50574902 on Build GRCh38
rs35414863 Chr.20: 50574654-50574654 on Build GRCh38
rs1054611126 Chr.20: 50574798-50574798 on Build GRCh38
rs1042115709 Chr.20: 50574756-50574756 on Build GRCh38
rs1013097538 Chr.20: 50574827-50574827 on Build GRCh38
rs779882276 Chr.20: 50574526-50574526 on Build GRCh38
rs759655653 Chr.20: 50574670-50574670 on Build GRCh38
rs950841020 Chr.20: 50574664-50574664 on Build GRCh38
rs746592969 Chr.20: 50574534-50574534 on Build GRCh38
rs970077999 Chr.20: 50574748-50574748 on Build GRCh38
rs1023234289 Chr.20: 50574508-50574508 on Build GRCh38
rs893269621 Chr.20: 50574814-50574814 on Build GRCh38
rs757230978 Chr.20: 50574479-50574479 on Build GRCh38
rs759985095 Chr.20: 50574744-50574744 on Build GRCh38
rs192756235 Chr.20: 50574510-50574510 on Build GRCh38
rs763028609 Chr.20: 50574640-50574640 on Build GRCh38
rs936435174 Chr.20: 50574777-50574777 on Build GRCh38
rs753477127 Chr.20: 50574904-50574904 on Build GRCh38
rs923223369 Chr.20: 50574736-50574736 on Build GRCh38
rs59132941 Chr.20: 50574687-50574687 on Build GRCh38
rs763990171 Chr.20: 50574680-50574680 on Build GRCh38

View Full Product Details