Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 57173078-57173570 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMP7
Entrez Gene ID
655
Gene Name
bone morphogenetic protein 7
Gene Aliases
OP-1
UniGene
Hs.473163
Gene Chromosome Location
Chr.20: 57168753-57266651 on Build GRCh38
Transcripts
NM_001719.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
373064 c.1065C>A p.A355A Chr.20: 57173281-57173281 on Build GRCh38
1028333 c.1062C>T p.Y354Y Chr.20: 57173284-57173284 on Build GRCh38
1412657 c.1050G>A p.A350A Chr.20: 57173296-57173296 on Build GRCh38
7357595 c.1125C>A p.N375K Chr.20: 57173221-57173221 on Build GRCh38
6611974 c.1129G>A p.A377T Chr.20: 57173217-57173217 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764706768 Chr.20: 57173218-57173218 on Build GRCh38
rs369263421 Chr.20: 57173345-57173345 on Build GRCh38
rs759493546 Chr.20: 57173188-57173188 on Build GRCh38
rs780453766 Chr.20: 57173147-57173147 on Build GRCh38
rs773408240 Chr.20: 57173266-57173266 on Build GRCh38
rs775245192 Chr.20: 57173296-57173296 on Build GRCh38
rs1040879793 Chr.20: 57173434-57173434 on Build GRCh38
rs151141435 Chr.20: 57173242-57173242 on Build GRCh38
rs761442203 Chr.20: 57173316-57173316 on Build GRCh38
rs909415889 Chr.20: 57173244-57173244 on Build GRCh38
rs371556453 Chr.20: 57173347-57173347 on Build GRCh38
rs978293630 Chr.20: 57173505-57173507 on Build GRCh38
rs769953359 Chr.20: 57173183-57173183 on Build GRCh38
rs142139866 Chr.20: 57173230-57173230 on Build GRCh38
rs547509078 Chr.20: 57173476-57173476 on Build GRCh38
rs770187349 Chr.20: 57173263-57173263 on Build GRCh38
rs139465481 Chr.20: 57173302-57173302 on Build GRCh38
rs572210562 Chr.20: 57173135-57173135 on Build GRCh38
rs752479332 Chr.20: 57173118-57173118 on Build GRCh38
rs750097211 Chr.20: 57173328-57173328 on Build GRCh38
rs149804552 Chr.20: 57173500-57173500 on Build GRCh38
rs2148328 Chr.20: 57173150-57173150 on Build GRCh38
rs535134055 Chr.20: 57173114-57173114 on Build GRCh38
rs200894126 Chr.20: 57173149-57173149 on Build GRCh38
rs749995679 Chr.20: 57173222-57173222 on Build GRCh38
rs748402363 Chr.20: 57173173-57173173 on Build GRCh38
rs746344981 Chr.20: 57173141-57173141 on Build GRCh38
rs1011512030 Chr.20: 57173130-57173130 on Build GRCh38
rs996537016 Chr.20: 57173177-57173177 on Build GRCh38
rs372973923 Chr.20: 57173297-57173297 on Build GRCh38
rs888121219 Chr.20: 57173489-57173489 on Build GRCh38
rs375831613 Chr.20: 57173325-57173325 on Build GRCh38
rs541901559 Chr.20: 57173351-57173351 on Build GRCh38
rs778428047 Chr.20: 57173138-57173138 on Build GRCh38
rs773463308 Chr.20: 57173185-57173185 on Build GRCh38
rs755547528 Chr.20: 57173349-57173349 on Build GRCh38
rs900981560 Chr.20: 57173442-57173442 on Build GRCh38
rs756397722 Chr.20: 57173360-57173360 on Build GRCh38
rs779114830 Chr.20: 57173145-57173145 on Build GRCh38
rs780359443 Chr.20: 57173229-57173229 on Build GRCh38
rs1028455237 Chr.20: 57173479-57173479 on Build GRCh38
rs762310439 Chr.20: 57173333-57173333 on Build GRCh38
rs527636941 Chr.20: 57173399-57173399 on Build GRCh38
rs751491608 Chr.20: 57173109-57173109 on Build GRCh38
rs748347077 Chr.20: 57173256-57173256 on Build GRCh38
rs561757689 Chr.20: 57173395-57173395 on Build GRCh38
rs755248955 Chr.20: 57173234-57173234 on Build GRCh38
rs775381264 Chr.20: 57173341-57173342 on Build GRCh38
rs769728766 Chr.20: 57173313-57173315 on Build GRCh38
rs775347487 Chr.20: 57173193-57173193 on Build GRCh38
rs962256697 Chr.20: 57173533-57173533 on Build GRCh38
rs369718771 Chr.20: 57173430-57173430 on Build GRCh38
rs763590054 Chr.20: 57173125-57173125 on Build GRCh38
rs555338424 Chr.20: 57173120-57173120 on Build GRCh38
rs563071072 Chr.20: 57173299-57173299 on Build GRCh38
rs35493026 Chr.20: 57173439-57173440 on Build GRCh38
rs779657440 Chr.20: 57173506-57173506 on Build GRCh38
rs934507720 Chr.20: 57173425-57173425 on Build GRCh38
rs924177455 Chr.20: 57173515-57173515 on Build GRCh38
rs749392235 Chr.20: 57173280-57173280 on Build GRCh38
rs533716905 Chr.20: 57173370-57173370 on Build GRCh38
rs377176431 Chr.20: 57173161-57173161 on Build GRCh38
rs189742184 Chr.20: 57173212-57173212 on Build GRCh38
rs745784460 Chr.20: 57173396-57173396 on Build GRCh38
rs1042627315 Chr.20: 57173160-57173160 on Build GRCh38
rs1002945655 Chr.20: 57173406-57173406 on Build GRCh38
rs753506760 Chr.20: 57173128-57173128 on Build GRCh38
rs758939521 Chr.20: 57173224-57173224 on Build GRCh38
rs962224086 Chr.20: 57173283-57173283 on Build GRCh38
rs771059650 Chr.20: 57173284-57173284 on Build GRCh38
rs753451605 Chr.20: 57173203-57173203 on Build GRCh38
rs763975815 Chr.20: 57173202-57173202 on Build GRCh38
rs368543219 Chr.20: 57173189-57173189 on Build GRCh38
rs753165642 Chr.20: 57173353-57173353 on Build GRCh38
rs767309601 Chr.20: 57173116-57173116 on Build GRCh38
rs375516908 Chr.20: 57173352-57173352 on Build GRCh38
rs965109180 Chr.20: 57173152-57173152 on Build GRCh38

View Full Product Details