Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 58989569-58990102 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NELFCD
Entrez Gene ID
51497
Gene Name
negative elongation factor complex member C/D
Gene Aliases
HSPC130, NELF-C, NELF-D, TH1, TH1L
UniGene
Hs.517148
Gene Chromosome Location
Chr.20: 58981208-58995133 on Build GRCh38
Transcripts
NM_198976.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1028522 c.742C>T p.L248L Chr.20: 58989915-58989915 on Build GRCh38
1028523 c.783C>T p.I261I Chr.20: 58989956-58989956 on Build GRCh38
2764960 c.662delA p.N223fs*22 Chr.20: 58989618-58989618 on Build GRCh38
2764963 c.768C>T p.S256S Chr.20: 58989941-58989941 on Build GRCh38
4557866 c.717G>C p.L239L Chr.20: 58989890-58989890 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754277765 Chr.20: 58989759-58989759 on Build GRCh38
rs766148754 Chr.20: 58989623-58989623 on Build GRCh38
rs749104695 Chr.20: 58989720-58989720 on Build GRCh38
rs773231887 Chr.20: 58989718-58989718 on Build GRCh38
rs774890170 Chr.20: 58990022-58990022 on Build GRCh38
rs778671462 Chr.20: 58989911-58989911 on Build GRCh38
rs1031131294 Chr.20: 58990055-58990055 on Build GRCh38
rs775770584 Chr.20: 58989592-58989592 on Build GRCh38
rs531228684 Chr.20: 58989781-58989781 on Build GRCh38
rs750829960 Chr.20: 58989836-58989836 on Build GRCh38
rs758970560 Chr.20: 58989638-58989638 on Build GRCh38
rs199788954 Chr.20: 58989849-58989849 on Build GRCh38
rs750658740 Chr.20: 58989767-58989767 on Build GRCh38
rs920632659 Chr.20: 58989932-58989932 on Build GRCh38
rs757559768 Chr.20: 58989666-58989666 on Build GRCh38
rs17851253 Chr.20: 58989640-58989640 on Build GRCh38
rs563674135 Chr.20: 58989771-58989771 on Build GRCh38
rs751885972 Chr.20: 58989961-58989961 on Build GRCh38
rs746702794 Chr.20: 58989992-58989992 on Build GRCh38
rs773533500 Chr.20: 58989953-58989953 on Build GRCh38
rs1037706224 Chr.20: 58989706-58989706 on Build GRCh38
rs764275132 Chr.20: 58989711-58989711 on Build GRCh38
rs539943354 Chr.20: 58989941-58989941 on Build GRCh38
rs770904830 Chr.20: 58989780-58989780 on Build GRCh38
rs368991410 Chr.20: 58989876-58989876 on Build GRCh38
rs758272595 Chr.20: 58989986-58989986 on Build GRCh38
rs559769118 Chr.20: 58989722-58989722 on Build GRCh38
rs541152557 Chr.20: 58989714-58989714 on Build GRCh38
rs147835273 Chr.20: 58989973-58989973 on Build GRCh38
rs769115215 Chr.20: 58990013-58990013 on Build GRCh38
rs751147774 Chr.20: 58989670-58989672 on Build GRCh38
rs766299985 Chr.20: 58989860-58989860 on Build GRCh38
rs763377658 Chr.20: 58989956-58989956 on Build GRCh38
rs199733975 Chr.20: 58990012-58990012 on Build GRCh38
rs938649411 Chr.20: 58989904-58989904 on Build GRCh38
rs370017836 Chr.20: 58990015-58990015 on Build GRCh38
rs765558453 Chr.20: 58989833-58989833 on Build GRCh38
rs754224780 Chr.20: 58989662-58989662 on Build GRCh38
rs758613009 Chr.20: 58989768-58989768 on Build GRCh38
rs200401626 Chr.20: 58989807-58989807 on Build GRCh38
rs938490726 Chr.20: 58989951-58989951 on Build GRCh38
rs753196531 Chr.20: 58989641-58989641 on Build GRCh38
rs773832437 Chr.20: 58990014-58990014 on Build GRCh38
rs762794992 Chr.20: 58989607-58989607 on Build GRCh38
rs748076762 Chr.20: 58989709-58989709 on Build GRCh38
rs757688195 Chr.20: 58989762-58989762 on Build GRCh38
rs778915200 Chr.20: 58989977-58989977 on Build GRCh38
rs577010469 Chr.20: 58989708-58989708 on Build GRCh38
rs756094033 Chr.20: 58989974-58989974 on Build GRCh38
rs374402949 Chr.20: 58990019-58990019 on Build GRCh38
rs1012551040 Chr.20: 58989775-58989775 on Build GRCh38
rs749591281 Chr.20: 58989615-58989615 on Build GRCh38
rs776086900 Chr.20: 58989945-58989945 on Build GRCh38
rs776438916 Chr.20: 58989912-58989912 on Build GRCh38
rs376148366 Chr.20: 58989828-58989828 on Build GRCh38
rs766633113 Chr.20: 58989844-58989844 on Build GRCh38
rs775101513 Chr.20: 58989756-58989756 on Build GRCh38
rs999088939 Chr.20: 58989799-58989799 on Build GRCh38
rs770356436 Chr.20: 58989949-58989949 on Build GRCh38
rs749230875 Chr.20: 58989778-58989778 on Build GRCh38
rs753842441 Chr.20: 58989798-58989798 on Build GRCh38
rs751195063 Chr.20: 58989624-58989624 on Build GRCh38
rs766644663 Chr.20: 58989957-58989957 on Build GRCh38
rs746100841 Chr.20: 58989684-58989684 on Build GRCh38
rs201430803 Chr.20: 58989820-58989820 on Build GRCh38
rs552944245 Chr.20: 58989803-58989803 on Build GRCh38
rs373372450 Chr.20: 58989937-58989937 on Build GRCh38
rs147676359 Chr.20: 58990056-58990056 on Build GRCh38
rs897630095 Chr.20: 58989884-58989884 on Build GRCh38
rs896513202 Chr.20: 58989922-58989922 on Build GRCh38
rs770780332 Chr.20: 58989723-58989723 on Build GRCh38
rs752904067 Chr.20: 58990036-58990036 on Build GRCh38
rs758464494 Chr.20: 58989701-58989701 on Build GRCh38
rs780773406 Chr.20: 58990008-58990008 on Build GRCh38
rs760875955 Chr.20: 58990040-58990040 on Build GRCh38
rs908473790 Chr.20: 58989942-58989942 on Build GRCh38
rs774626381 Chr.20: 58989774-58989774 on Build GRCh38
rs554301270 Chr.20: 58989845-58989845 on Build GRCh38
rs534146653 Chr.20: 58989806-58989806 on Build GRCh38
rs765573828 Chr.20: 58989765-58989765 on Build GRCh38
rs79806098 Chr.20: 58989769-58989769 on Build GRCh38
rs759807033 Chr.20: 58989972-58989972 on Build GRCh38
rs759933370 Chr.20: 58990026-58990026 on Build GRCh38
rs755047050 Chr.20: 58989773-58989773 on Build GRCh38
rs750324749 Chr.20: 58989983-58989983 on Build GRCh38
rs778217155 Chr.20: 58989656-58989656 on Build GRCh38
rs768327000 Chr.20: 58989999-58989999 on Build GRCh38
rs757733941 Chr.20: 58989917-58989917 on Build GRCh38
rs1023567196 Chr.20: 58989955-58989955 on Build GRCh38
rs766978761 Chr.20: 58989749-58989749 on Build GRCh38
rs758639976 Chr.20: 58989839-58989839 on Build GRCh38
rs763591163 Chr.20: 58989815-58989815 on Build GRCh38
rs764599165 Chr.20: 58989757-58989757 on Build GRCh38
rs781708442 Chr.20: 58989999-58990000 on Build GRCh38
rs762285085 Chr.20: 58989832-58989832 on Build GRCh38
rs771783056 Chr.20: 58989785-58989785 on Build GRCh38
rs779114487 Chr.20: 58989676-58989676 on Build GRCh38
rs760183408 Chr.20: 58989808-58989808 on Build GRCh38
rs530150217 Chr.20: 58989724-58989724 on Build GRCh38
rs976396063 Chr.20: 58990081-58990081 on Build GRCh38
rs761271004 Chr.20: 58989948-58989948 on Build GRCh38
rs941739579 Chr.20: 58989811-58989811 on Build GRCh38
rs571293549 Chr.20: 58989742-58989742 on Build GRCh38
rs181957233 Chr.20: 58990029-58990029 on Build GRCh38
rs756616986 Chr.20: 58989643-58989643 on Build GRCh38

View Full Product Details