Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 62201715-62202207 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTG2
Entrez Gene ID
26164
Gene Name
mitochondrial ribosome associated GTPase 2
Gene Aliases
GTPBP5, ObgH1, dJ1005F21.2
UniGene
Hs.340636
Gene Chromosome Location
Chr.20: 62183025-62203568 on Build GRCh38
Transcripts
NM_015666.3 XM_005260393.1 XM_006723774.3 XM_011528776.1 XM_017027805.1 XM_017027806.1 XM_017027807.1 XM_017027808.1

SNP Detail

SNP ID Chromosome Location
rs923822900 Chr.20: 62202126-62202126 on Build GRCh38
rs1033207191 Chr.20: 62202005-62202005 on Build GRCh38
rs59232077 Chr.20: 62201859-62201859 on Build GRCh38
rs1042362768 Chr.20: 62201742-62201742 on Build GRCh38
rs145296448 Chr.20: 62201962-62201962 on Build GRCh38
rs185041954 Chr.20: 62202015-62202015 on Build GRCh38
rs1051977440 Chr.20: 62202029-62202029 on Build GRCh38
rs1025503492 Chr.20: 62202109-62202109 on Build GRCh38
rs936346449 Chr.20: 62201762-62201762 on Build GRCh38
rs189841174 Chr.20: 62202090-62202090 on Build GRCh38
rs533923398 Chr.20: 62202104-62202104 on Build GRCh38
rs528336532 Chr.20: 62202071-62202071 on Build GRCh38
rs1055859436 Chr.20: 62201933-62201933 on Build GRCh38
rs530145686 Chr.20: 62202113-62202113 on Build GRCh38
rs2184156 Chr.20: 62202115-62202115 on Build GRCh38
rs977984670 Chr.20: 62201971-62201971 on Build GRCh38
rs897420327 Chr.20: 62202055-62202055 on Build GRCh38
rs925715271 Chr.20: 62201807-62201807 on Build GRCh38
rs950211140 Chr.20: 62202116-62202116 on Build GRCh38
rs917751522 Chr.20: 62201959-62201959 on Build GRCh38
rs138494643 Chr.20: 62201781-62201781 on Build GRCh38
rs2184157 Chr.20: 62202099-62202099 on Build GRCh38
rs977935261 Chr.20: 62202118-62202118 on Build GRCh38
rs569995867 Chr.20: 62201888-62201888 on Build GRCh38
rs535213130 Chr.20: 62201941-62201941 on Build GRCh38
rs775219836 Chr.20: 62201780-62201780 on Build GRCh38
rs552405986 Chr.20: 62201744-62201744 on Build GRCh38
rs1008419513 Chr.20: 62201869-62201869 on Build GRCh38
rs2144279 Chr.20: 62202014-62202014 on Build GRCh38
rs564165403 Chr.20: 62201753-62201753 on Build GRCh38
rs762039699 Chr.20: 62201813-62201813 on Build GRCh38
rs1012321516 Chr.20: 62202101-62202101 on Build GRCh38
rs888521771 Chr.20: 62201912-62201912 on Build GRCh38
rs902167818 Chr.20: 62201751-62201751 on Build GRCh38
rs80174504 Chr.20: 62201832-62201832 on Build GRCh38
rs750572239 Chr.20: 62201833-62201833 on Build GRCh38
rs75901783 Chr.20: 62202165-62202165 on Build GRCh38
rs140224364 Chr.20: 62201860-62201860 on Build GRCh38
rs559201291 Chr.20: 62201915-62201915 on Build GRCh38
rs922561805 Chr.20: 62201877-62201877 on Build GRCh38
rs773403715 Chr.20: 62202169-62202169 on Build GRCh38
rs949281534 Chr.20: 62201739-62201739 on Build GRCh38
rs900265763 Chr.20: 62201796-62201796 on Build GRCh38
rs192275558 Chr.20: 62201882-62201882 on Build GRCh38
rs910487124 Chr.20: 62201858-62201858 on Build GRCh38
rs181835462 Chr.20: 62202043-62202043 on Build GRCh38
rs1041070337 Chr.20: 62201871-62201871 on Build GRCh38
rs137978533 Chr.20: 62202030-62202030 on Build GRCh38
rs1760050 Chr.20: 62201903-62201903 on Build GRCh38
rs73137297 Chr.20: 62202056-62202056 on Build GRCh38
rs754326030 Chr.20: 62201883-62201883 on Build GRCh38
rs189385121 Chr.20: 62202016-62202016 on Build GRCh38
rs12625898 Chr.20: 62201940-62201940 on Build GRCh38
rs752254296 Chr.20: 62201989-62201989 on Build GRCh38
rs758181666 Chr.20: 62201975-62201975 on Build GRCh38
rs745820910 Chr.20: 62202032-62202032 on Build GRCh38
rs6061457 Chr.20: 62202176-62202176 on Build GRCh38

View Full Product Details