Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 31672217-31672760 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCAF4
Entrez Gene ID
57466
Gene Name
SR-related CTD associated factor 4
Gene Aliases
SFRS15, SRA4
UniGene
Hs.17255
Gene Chromosome Location
Chr.21: 31671000-31732118 on Build GRCh38
Transcripts
NM_001145444.1 NM_001145445.1 NM_020706.2 XM_005261017.2 XM_006724035.2 XM_006724036.2 XM_017028415.1 XM_017028416.1 XM_017028417.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1030080 c.2609T>C p.L870S Chr.21: 31672234-31672234 on Build GRCh38
3405354 c.2554G>A p.A852T Chr.21: 31672289-31672289 on Build GRCh38
6772355 c.2600delC p.P867fs*9 Chr.21: 31672243-31672243 on Build GRCh38
6896690 c.2559G>A p.R853R Chr.21: 31672284-31672284 on Build GRCh38
6401450 c.2527C>A p.Q843K Chr.21: 31672316-31672316 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147745077 Chr.21: 31672734-31672734 on Build GRCh38
rs748574302 Chr.21: 31672497-31672497 on Build GRCh38
rs184675235 Chr.21: 31672407-31672407 on Build GRCh38
rs747940266 Chr.21: 31672347-31672347 on Build GRCh38
rs773056583 Chr.21: 31672363-31672363 on Build GRCh38
rs184384572 Chr.21: 31672635-31672635 on Build GRCh38
rs967803400 Chr.21: 31672426-31672426 on Build GRCh38
rs752345878 Chr.21: 31672386-31672386 on Build GRCh38
rs757591341 Chr.21: 31672389-31672389 on Build GRCh38
rs774971958 Chr.21: 31672357-31672357 on Build GRCh38
rs371447774 Chr.21: 31672346-31672346 on Build GRCh38
rs940410640 Chr.21: 31672286-31672286 on Build GRCh38
rs1028527276 Chr.21: 31672723-31672723 on Build GRCh38
rs754624681 Chr.21: 31672244-31672244 on Build GRCh38
rs756601813 Chr.21: 31672375-31672376 on Build GRCh38
rs149687210 Chr.21: 31672281-31672281 on Build GRCh38
rs753676664 Chr.21: 31672302-31672302 on Build GRCh38
rs759115948 Chr.21: 31672378-31672378 on Build GRCh38
rs116261565 Chr.21: 31672560-31672560 on Build GRCh38
rs529390940 Chr.21: 31672715-31672715 on Build GRCh38
rs766841131 Chr.21: 31672407-31672407 on Build GRCh38
rs774067797 Chr.21: 31672376-31672376 on Build GRCh38
rs199989307 Chr.21: 31672290-31672290 on Build GRCh38
rs886614262 Chr.21: 31672679-31672679 on Build GRCh38
rs368053787 Chr.21: 31672339-31672339 on Build GRCh38
rs747076602 Chr.21: 31672337-31672337 on Build GRCh38
rs200966344 Chr.21: 31672285-31672285 on Build GRCh38
rs529034982 Chr.21: 31672245-31672245 on Build GRCh38
rs201723551 Chr.21: 31672250-31672250 on Build GRCh38
rs1005022317 Chr.21: 31672713-31672713 on Build GRCh38
rs181436038 Chr.21: 31672614-31672614 on Build GRCh38
rs560258689 Chr.21: 31672663-31672663 on Build GRCh38
rs747149760 Chr.21: 31672541-31672541 on Build GRCh38
rs766489760 Chr.21: 31672736-31672736 on Build GRCh38
rs371653392 Chr.21: 31672322-31672322 on Build GRCh38
rs986681944 Chr.21: 31672540-31672540 on Build GRCh38
rs566795257 Chr.21: 31672529-31672529 on Build GRCh38
rs568997478 Chr.21: 31672566-31672566 on Build GRCh38
rs997407153 Chr.21: 31672710-31672710 on Build GRCh38
rs188643974 Chr.21: 31672548-31672548 on Build GRCh38
rs1036527053 Chr.21: 31672287-31672287 on Build GRCh38
rs759000823 Chr.21: 31672396-31672396 on Build GRCh38
rs750903957 Chr.21: 31672394-31672394 on Build GRCh38
rs746342814 Chr.21: 31672297-31672297 on Build GRCh38
rs367892452 Chr.21: 31672704-31672704 on Build GRCh38
rs2833475 Chr.21: 31672507-31672507 on Build GRCh38
rs765820592 Chr.21: 31672365-31672365 on Build GRCh38
rs12152067 Chr.21: 31672306-31672306 on Build GRCh38
rs930738751 Chr.21: 31672664-31672664 on Build GRCh38
rs551748841 Chr.21: 31672335-31672335 on Build GRCh38
rs374428803 Chr.21: 31672579-31672579 on Build GRCh38
rs1049735220 Chr.21: 31672673-31672673 on Build GRCh38
rs955582038 Chr.21: 31672736-31672736 on Build GRCh38
rs762882113 Chr.21: 31672364-31672364 on Build GRCh38
rs765567804 Chr.21: 31672390-31672390 on Build GRCh38
rs766220154 Chr.21: 31672295-31672295 on Build GRCh38
rs548979165 Chr.21: 31672459-31672459 on Build GRCh38
rs763138985 Chr.21: 31672414-31672415 on Build GRCh38
rs1033504230 Chr.21: 31672669-31672669 on Build GRCh38
rs767321404 Chr.21: 31672381-31672381 on Build GRCh38
rs949983256 Chr.21: 31672427-31672427 on Build GRCh38
rs766335442 Chr.21: 31672419-31672420 on Build GRCh38
rs920391064 Chr.21: 31672289-31672289 on Build GRCh38
rs374591358 Chr.21: 31672324-31672324 on Build GRCh38

View Full Product Details