Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 31685468-31685923 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCAF4
Entrez Gene ID
57466
Gene Name
SR-related CTD associated factor 4
Gene Aliases
SFRS15, SRA4
UniGene
Hs.17255
Gene Chromosome Location
Chr.21: 31671000-31732118 on Build GRCh38
Transcripts
NM_001145444.1 NM_001145445.1 NM_020706.2 XM_005261017.2 XM_006724035.2 XM_006724036.2 XM_017028415.1 XM_017028416.1 XM_017028417.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5953646 c.2158C>T p.P720S Chr.21: 31685619-31685619 on Build GRCh38
1615986 c.2090C>T p.P697L Chr.21: 31685687-31685687 on Build GRCh38
5937881 c.2149C>T p.P717S Chr.21: 31685628-31685628 on Build GRCh38
4752378 c.2218C>T p.P740S Chr.21: 31685476-31685476 on Build GRCh38
6772348 c.2213T>G p.F738C Chr.21: 31685481-31685481 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758181527 Chr.21: 31685781-31685781 on Build GRCh38
rs778314236 Chr.21: 31685715-31685715 on Build GRCh38
rs757565205 Chr.21: 31685856-31685856 on Build GRCh38
rs181372986 Chr.21: 31685727-31685727 on Build GRCh38
rs369638100 Chr.21: 31685699-31685699 on Build GRCh38
rs752639758 Chr.21: 31685656-31685656 on Build GRCh38
rs201206769 Chr.21: 31685619-31685619 on Build GRCh38
rs535651303 Chr.21: 31685503-31685503 on Build GRCh38
rs984003493 Chr.21: 31685617-31685617 on Build GRCh38
rs1028117707 Chr.21: 31685594-31685594 on Build GRCh38
rs745997090 Chr.21: 31685885-31685885 on Build GRCh38
rs115568998 Chr.21: 31685863-31685863 on Build GRCh38
rs750890297 Chr.21: 31685621-31685621 on Build GRCh38
rs191306091 Chr.21: 31685534-31685534 on Build GRCh38
rs78281614 Chr.21: 31685762-31685762 on Build GRCh38
rs771870355 Chr.21: 31685741-31685741 on Build GRCh38
rs900841372 Chr.21: 31685873-31685873 on Build GRCh38
rs1028649796 Chr.21: 31685884-31685884 on Build GRCh38
rs570022991 Chr.21: 31685875-31685875 on Build GRCh38
rs759275436 Chr.21: 31685630-31685630 on Build GRCh38
rs759346430 Chr.21: 31685530-31685530 on Build GRCh38
rs774611355 Chr.21: 31685626-31685626 on Build GRCh38
rs866741788 Chr.21: 31685613-31685613 on Build GRCh38
rs184524498 Chr.21: 31685563-31685563 on Build GRCh38
rs765858143 Chr.21: 31685513-31685513 on Build GRCh38
rs3841743 Chr.21: 31685622-31685623 on Build GRCh38
rs369802404 Chr.21: 31685676-31685676 on Build GRCh38
rs370308214 Chr.21: 31685749-31685749 on Build GRCh38
rs550829570 Chr.21: 31685789-31685789 on Build GRCh38
rs184958033 Chr.21: 31685774-31685774 on Build GRCh38
rs374793994 Chr.21: 31685765-31685765 on Build GRCh38
rs375999273 Chr.21: 31685536-31685536 on Build GRCh38
rs938553168 Chr.21: 31685702-31685702 on Build GRCh38
rs994780298 Chr.21: 31685843-31685843 on Build GRCh38
rs764010062 Chr.21: 31685661-31685661 on Build GRCh38
rs199940701 Chr.21: 31685686-31685686 on Build GRCh38
rs756923189 Chr.21: 31685553-31685553 on Build GRCh38
rs574147232 Chr.21: 31685611-31685611 on Build GRCh38
rs753325665 Chr.21: 31685670-31685670 on Build GRCh38
rs919270078 Chr.21: 31685675-31685675 on Build GRCh38
rs1002417628 Chr.21: 31685522-31685522 on Build GRCh38
rs557832665 Chr.21: 31685545-31685545 on Build GRCh38
rs568209777 Chr.21: 31685495-31685495 on Build GRCh38
rs759740671 Chr.21: 31685737-31685737 on Build GRCh38
rs748510825 Chr.21: 31685697-31685697 on Build GRCh38
rs769091419 Chr.21: 31685572-31685572 on Build GRCh38
rs13052367 Chr.21: 31685758-31685758 on Build GRCh38
rs781771912 Chr.21: 31685579-31685579 on Build GRCh38
rs142985392 Chr.21: 31685539-31685539 on Build GRCh38
rs569151965 Chr.21: 31685825-31685825 on Build GRCh38
rs753133252 Chr.21: 31685736-31685745 on Build GRCh38
rs561248924 Chr.21: 31685763-31685763 on Build GRCh38
rs750057406 Chr.21: 31685777-31685777 on Build GRCh38
rs758541096 Chr.21: 31685743-31685747 on Build GRCh38
rs146113352 Chr.21: 31685716-31685716 on Build GRCh38
rs778594323 Chr.21: 31685557-31685557 on Build GRCh38
rs893537517 Chr.21: 31685767-31685767 on Build GRCh38
rs140696970 Chr.21: 31685657-31685657 on Build GRCh38
rs758691118 Chr.21: 31685650-31685650 on Build GRCh38
rs201536213 Chr.21: 31685547-31685547 on Build GRCh38
rs767263244 Chr.21: 31685651-31685651 on Build GRCh38
rs770996481 Chr.21: 31685717-31685717 on Build GRCh38
rs771100211 Chr.21: 31685622-31685622 on Build GRCh38
rs984341273 Chr.21: 31685899-31685899 on Build GRCh38
rs766610602 Chr.21: 31685516-31685516 on Build GRCh38
rs201740202 Chr.21: 31685596-31685596 on Build GRCh38
rs368708758 Chr.21: 31685764-31685764 on Build GRCh38
rs199587645 Chr.21: 31685529-31685529 on Build GRCh38
rs533305503 Chr.21: 31685828-31685828 on Build GRCh38
rs545000525 Chr.21: 31685687-31685687 on Build GRCh38
rs988736695 Chr.21: 31685704-31685704 on Build GRCh38
rs560614560 Chr.21: 31685611-31685612 on Build GRCh38
rs763028443 Chr.21: 31685508-31685508 on Build GRCh38
rs1025862984 Chr.21: 31685850-31685850 on Build GRCh38
rs145915853 Chr.21: 31685671-31685671 on Build GRCh38

View Full Product Details