Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 32392678-32393182 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
URB1
Entrez Gene ID
9875
Gene Name
URB1 ribosome biogenesis 1 homolog (S. cerevisiae)
Gene Aliases
C21orf108, NPA1
UniGene
Hs.473611
Gene Chromosome Location
Chr.21: 32311019-32393003 on Build GRCh38
Transcripts
NM_014825.2 NR_026845.1

Gene Symbol
URB1-AS1
Entrez Gene ID
84996
Gene Name
URB1 antisense RNA 1 (head to head)
Gene Aliases
C21orf119, PRED84
UniGene
Hs.58149
Gene Chromosome Location
Chr.21: 32393133-32393960 on Build GRCh38
Transcripts
NM_014825.2 NR_026845.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6290389 c.133C>T p.P45S Chr.21: 32392778-32392778 on Build GRCh38
6336215 c.105C>A p.F35L Chr.21: 32392806-32392806 on Build GRCh38
6539545 c.101G>C p.R34P Chr.21: 32392810-32392810 on Build GRCh38
4770563 c.24delC p.S9fs*31 Chr.21: 32392887-32392887 on Build GRCh38
1616002 c.127C>A p.Q43K Chr.21: 32392784-32392784 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772974314 Chr.21: 32392809-32392811 on Build GRCh38
rs1051667468 Chr.21: 32393072-32393072 on Build GRCh38
rs529094250 Chr.21: 32392969-32392969 on Build GRCh38
rs963069777 Chr.21: 32393004-32393004 on Build GRCh38
rs923194948 Chr.21: 32392999-32392999 on Build GRCh38
rs1046187353 Chr.21: 32392897-32392897 on Build GRCh38
rs1037881557 Chr.21: 32393052-32393052 on Build GRCh38
rs150903996 Chr.21: 32393148-32393163 on Build GRCh38
rs368817965 Chr.21: 32392998-32392998 on Build GRCh38
rs889007570 Chr.21: 32393082-32393082 on Build GRCh38
rs918752802 Chr.21: 32393009-32393009 on Build GRCh38
rs1029957565 Chr.21: 32392717-32392717 on Build GRCh38
rs985890328 Chr.21: 32392730-32392730 on Build GRCh38
rs1010484411 Chr.21: 32393103-32393103 on Build GRCh38
rs893189287 Chr.21: 32392943-32392943 on Build GRCh38
rs968770287 Chr.21: 32392979-32392979 on Build GRCh38
rs886142024 Chr.21: 32393039-32393039 on Build GRCh38
rs903107044 Chr.21: 32392972-32392972 on Build GRCh38
rs979515693 Chr.21: 32393032-32393032 on Build GRCh38
rs34323951 Chr.21: 32392775-32392776 on Build GRCh38
rs997869760 Chr.21: 32393064-32393064 on Build GRCh38
rs923142405 Chr.21: 32393146-32393146 on Build GRCh38
rs928651952 Chr.21: 32393030-32393030 on Build GRCh38
rs551051101 Chr.21: 32393130-32393130 on Build GRCh38
rs60255358 Chr.21: 32393045-32393062 on Build GRCh38
rs993200560 Chr.21: 32393105-32393105 on Build GRCh38
rs1055866236 Chr.21: 32392922-32392922 on Build GRCh38
rs771887868 Chr.21: 32392773-32392773 on Build GRCh38
rs1056172987 Chr.21: 32393037-32393037 on Build GRCh38
rs533063964 Chr.21: 32393115-32393115 on Build GRCh38
rs747673851 Chr.21: 32393084-32393084 on Build GRCh38
rs986306400 Chr.21: 32392993-32392993 on Build GRCh38
rs779327927 Chr.21: 32392764-32392764 on Build GRCh38
rs766646669 Chr.21: 32392932-32392962 on Build GRCh38
rs761156930 Chr.21: 32392814-32392815 on Build GRCh38
rs953060266 Chr.21: 32392992-32392992 on Build GRCh38
rs969151283 Chr.21: 32393108-32393108 on Build GRCh38
rs775507271 Chr.21: 32392762-32392762 on Build GRCh38
rs963100865 Chr.21: 32393100-32393100 on Build GRCh38
rs772404700 Chr.21: 32392760-32392760 on Build GRCh38
rs918852750 Chr.21: 32392838-32392838 on Build GRCh38
rs745921215 Chr.21: 32392741-32392741 on Build GRCh38
rs937378615 Chr.21: 32393036-32393036 on Build GRCh38
rs569410475 Chr.21: 32393154-32393154 on Build GRCh38
rs1030442809 Chr.21: 32392988-32392988 on Build GRCh38
rs779558462 Chr.21: 32392940-32392940 on Build GRCh38
rs550239668 Chr.21: 32393008-32393008 on Build GRCh38
rs951532625 Chr.21: 32393127-32393127 on Build GRCh38
rs1016309515 Chr.21: 32393094-32393094 on Build GRCh38
rs907521508 Chr.21: 32393153-32393153 on Build GRCh38
rs1047708310 Chr.21: 32393147-32393147 on Build GRCh38
rs960003214 Chr.21: 32392924-32392924 on Build GRCh38
rs893235637 Chr.21: 32393043-32393043 on Build GRCh38
rs974187005 Chr.21: 32392837-32392837 on Build GRCh38
rs930262051 Chr.21: 32393010-32393010 on Build GRCh38
rs1026038566 Chr.21: 32392811-32392811 on Build GRCh38
rs953286999 Chr.21: 32392725-32392725 on Build GRCh38
rs374019516 Chr.21: 32393012-32393029 on Build GRCh38
rs947415541 Chr.21: 32393044-32393044 on Build GRCh38
rs11702289 Chr.21: 32392997-32392997 on Build GRCh38
rs113297222 Chr.21: 32392708-32392708 on Build GRCh38
rs974219617 Chr.21: 32393007-32393007 on Build GRCh38
rs1016032701 Chr.21: 32393002-32393002 on Build GRCh38
rs776730007 Chr.21: 32392872-32392872 on Build GRCh38
rs571286620 Chr.21: 32392975-32392975 on Build GRCh38
rs906009883 Chr.21: 32393070-32393070 on Build GRCh38
rs181609239 Chr.21: 32393140-32393140 on Build GRCh38
rs772052390 Chr.21: 32393122-32393123 on Build GRCh38
rs960282742 Chr.21: 32393097-32393097 on Build GRCh38
rs901488835 Chr.21: 32392769-32392769 on Build GRCh38
rs932936316 Chr.21: 32393034-32393034 on Build GRCh38
rs978849592 Chr.21: 32393111-32393111 on Build GRCh38
rs750011419 Chr.21: 32392860-32392860 on Build GRCh38
rs1035870975 Chr.21: 32393088-32393088 on Build GRCh38
rs776578939 Chr.21: 32392813-32392813 on Build GRCh38
rs768388177 Chr.21: 32392810-32392810 on Build GRCh38
rs546492368 Chr.21: 32392908-32392908 on Build GRCh38
rs760531451 Chr.21: 32392791-32392791 on Build GRCh38
rs920801313 Chr.21: 32393112-32393113 on Build GRCh38
rs937452567 Chr.21: 32392921-32392921 on Build GRCh38
rs941513447 Chr.21: 32392820-32392820 on Build GRCh38
rs757889156 Chr.21: 32392935-32392935 on Build GRCh38
rs981705567 Chr.21: 32393016-32393016 on Build GRCh38
rs761774049 Chr.21: 32392853-32392853 on Build GRCh38
rs143045889 Chr.21: 32393043-32393044 on Build GRCh38
rs754224339 Chr.21: 32392931-32392931 on Build GRCh38
rs1025798027 Chr.21: 32393121-32393121 on Build GRCh38
rs1016427663 Chr.21: 32392870-32392870 on Build GRCh38
rs373827834 Chr.21: 32392918-32392918 on Build GRCh38
rs568832690 Chr.21: 32393027-32393027 on Build GRCh38
rs1041609512 Chr.21: 32393049-32393049 on Build GRCh38
rs903008578 Chr.21: 32393060-32393060 on Build GRCh38
rs979289151 Chr.21: 32392991-32392991 on Build GRCh38
rs1020097694 Chr.21: 32392971-32392971 on Build GRCh38
rs561542255 Chr.21: 32392954-32392954 on Build GRCh38
rs1039390949 Chr.21: 32392729-32392729 on Build GRCh38
rs765133597 Chr.21: 32392855-32392855 on Build GRCh38
rs374065244 Chr.21: 32392902-32392902 on Build GRCh38
rs967261680 Chr.21: 32392707-32392707 on Build GRCh38
rs1020234195 Chr.21: 32393107-32393107 on Build GRCh38
rs539302873 Chr.21: 32393056-32393056 on Build GRCh38

View Full Product Details