Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 32793713-32794216 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C21orf62
Entrez Gene ID
56245
Gene Name
chromosome 21 open reading frame 62
Gene Aliases
B37, C21orf120, PRED81
UniGene
Hs.517235
Gene Chromosome Location
Chr.21: 32790673-32813808 on Build GRCh38
Transcripts
NM_001162495.2 NM_001162496.2 NM_019596.5 NR_024622.1 NR_024623.1

Gene Symbol
C21orf62-AS1
Entrez Gene ID
54067
Gene Name
C21orf62 antisense RNA 1
Gene Aliases
C21orf49
UniGene
Hs.54725
Gene Chromosome Location
Chr.21: 32772100-32797707 on Build GRCh38
Transcripts
NM_001162495.2 NM_001162496.2 NM_019596.5 NR_024622.1 NR_024623.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7256502 c.425T>A p.I142N Chr.21: 32793997-32793997 on Build GRCh38
7300743 c.449C>T p.S150F Chr.21: 32793973-32793973 on Build GRCh38
3550331 c.439G>A p.D147N Chr.21: 32793983-32793983 on Build GRCh38
6467892 c.551A>G p.Y184C Chr.21: 32793871-32793871 on Build GRCh38
3550333 c.301G>A p.D101N Chr.21: 32794121-32794121 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780133299 Chr.21: 32794150-32794150 on Build GRCh38
rs770685535 Chr.21: 32794180-32794180 on Build GRCh38
rs750410793 Chr.21: 32794137-32794137 on Build GRCh38
rs982090978 Chr.21: 32794165-32794165 on Build GRCh38
rs758482724 Chr.21: 32794003-32794003 on Build GRCh38
rs778007251 Chr.21: 32793902-32793902 on Build GRCh38
rs761305523 Chr.21: 32793966-32793966 on Build GRCh38
rs762369991 Chr.21: 32794131-32794131 on Build GRCh38
rs758702783 Chr.21: 32794144-32794144 on Build GRCh38
rs767430057 Chr.21: 32793741-32793741 on Build GRCh38
rs746968756 Chr.21: 32793954-32793954 on Build GRCh38
rs761466482 Chr.21: 32793870-32793870 on Build GRCh38
rs374818772 Chr.21: 32793981-32793981 on Build GRCh38
rs751866590 Chr.21: 32793785-32793785 on Build GRCh38
rs201931377 Chr.21: 32794156-32794156 on Build GRCh38
rs775476230 Chr.21: 32793849-32793849 on Build GRCh38
rs751931600 Chr.21: 32793876-32793876 on Build GRCh38
rs748212020 Chr.21: 32793802-32793802 on Build GRCh38
rs375969368 Chr.21: 32794162-32794162 on Build GRCh38
rs767962508 Chr.21: 32793885-32793885 on Build GRCh38
rs200869965 Chr.21: 32794167-32794167 on Build GRCh38
rs375363659 Chr.21: 32794192-32794192 on Build GRCh38
rs752201249 Chr.21: 32793748-32793749 on Build GRCh38
rs541170407 Chr.21: 32793932-32793932 on Build GRCh38
rs749550140 Chr.21: 32793822-32793822 on Build GRCh38
rs756704238 Chr.21: 32793985-32793985 on Build GRCh38
rs558753345 Chr.21: 32793858-32793858 on Build GRCh38
rs756864702 Chr.21: 32793781-32793781 on Build GRCh38
rs377322005 Chr.21: 32794132-32794132 on Build GRCh38
rs2231390 Chr.21: 32794032-32794032 on Build GRCh38
rs1011809864 Chr.21: 32793744-32793744 on Build GRCh38
rs183684265 Chr.21: 32794168-32794168 on Build GRCh38
rs377526174 Chr.21: 32793993-32793993 on Build GRCh38
rs375164231 Chr.21: 32793753-32793753 on Build GRCh38
rs762674995 Chr.21: 32794062-32794062 on Build GRCh38
rs754375407 Chr.21: 32794128-32794128 on Build GRCh38
rs745661904 Chr.21: 32793835-32793835 on Build GRCh38
rs200790175 Chr.21: 32794084-32794084 on Build GRCh38
rs770435664 Chr.21: 32794081-32794081 on Build GRCh38
rs112582543 Chr.21: 32793986-32793986 on Build GRCh38
rs779273673 Chr.21: 32793921-32793921 on Build GRCh38
rs777358543 Chr.21: 32793824-32793825 on Build GRCh38
rs749324930 Chr.21: 32793907-32793907 on Build GRCh38
rs780403437 Chr.21: 32793783-32793783 on Build GRCh38
rs938486931 Chr.21: 32793871-32793871 on Build GRCh38
rs575904703 Chr.21: 32793790-32793790 on Build GRCh38
rs1029971128 Chr.21: 32793821-32793821 on Build GRCh38
rs753272960 Chr.21: 32793888-32793888 on Build GRCh38
rs771969003 Chr.21: 32793844-32793844 on Build GRCh38
rs745690168 Chr.21: 32793927-32793927 on Build GRCh38
rs375215701 Chr.21: 32793831-32793831 on Build GRCh38
rs373350275 Chr.21: 32793989-32793989 on Build GRCh38
rs751064410 Chr.21: 32793812-32793812 on Build GRCh38
rs368281384 Chr.21: 32793763-32793763 on Build GRCh38
rs772902465 Chr.21: 32793974-32793974 on Build GRCh38
rs867450780 Chr.21: 32794193-32794193 on Build GRCh38
rs760481485 Chr.21: 32793857-32793857 on Build GRCh38
rs369556552 Chr.21: 32793996-32793996 on Build GRCh38
rs372493378 Chr.21: 32793786-32793786 on Build GRCh38
rs913048180 Chr.21: 32794173-32794173 on Build GRCh38
rs764852823 Chr.21: 32793872-32793872 on Build GRCh38
rs969388251 Chr.21: 32793850-32793850 on Build GRCh38
rs779911066 Chr.21: 32794011-32794011 on Build GRCh38
rs773844372 Chr.21: 32794188-32794188 on Build GRCh38
rs771922449 Chr.21: 32793929-32793929 on Build GRCh38
rs79194766 Chr.21: 32793881-32793881 on Build GRCh38
rs757612998 Chr.21: 32793915-32793915 on Build GRCh38
rs371303342 Chr.21: 32793980-32793980 on Build GRCh38
rs988782377 Chr.21: 32794170-32794170 on Build GRCh38
rs751393654 Chr.21: 32794155-32794155 on Build GRCh38
rs762467814 Chr.21: 32793975-32793975 on Build GRCh38
rs764738726 Chr.21: 32794101-32794101 on Build GRCh38
rs761117395 Chr.21: 32794089-32794089 on Build GRCh38
rs879228383 Chr.21: 32793876-32793876 on Build GRCh38
rs148054755 Chr.21: 32794046-32794046 on Build GRCh38
rs901433097 Chr.21: 32794025-32794025 on Build GRCh38
rs79815715 Chr.21: 32793875-32793875 on Build GRCh38
rs369533216 Chr.21: 32793868-32793868 on Build GRCh38
rs377405712 Chr.21: 32794166-32794166 on Build GRCh38
rs201096811 Chr.21: 32794169-32794169 on Build GRCh38
rs141523679 Chr.21: 32793834-32793834 on Build GRCh38
rs757914579 Chr.21: 32793812-32793812 on Build GRCh38
rs535418618 Chr.21: 32793740-32793740 on Build GRCh38
rs373712189 Chr.21: 32794002-32794002 on Build GRCh38
rs74617086 Chr.21: 32793879-32793879 on Build GRCh38
rs768521325 Chr.21: 32793958-32793958 on Build GRCh38
rs768644293 Chr.21: 32794039-32794039 on Build GRCh38
rs777947927 Chr.21: 32793818-32793818 on Build GRCh38
rs752207127 Chr.21: 32793745-32793745 on Build GRCh38
rs11390393 Chr.21: 32793879-32793880 on Build GRCh38
rs757381747 Chr.21: 32793990-32793990 on Build GRCh38
rs764905771 Chr.21: 32793972-32793972 on Build GRCh38
rs746678100 Chr.21: 32794158-32794158 on Build GRCh38
rs947266128 Chr.21: 32794186-32794186 on Build GRCh38
rs756541645 Chr.21: 32793890-32793890 on Build GRCh38
rs1007182761 Chr.21: 32793910-32793910 on Build GRCh38
rs781073580 Chr.21: 32794045-32794045 on Build GRCh38
rs772560563 Chr.21: 32794048-32794048 on Build GRCh38
rs144644858 Chr.21: 32793965-32793965 on Build GRCh38

View Full Product Details