Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 41804453-41804954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRDM15
Entrez Gene ID
63977
Gene Name
PR domain 15
Gene Aliases
C21orf83, PFM15, ZNF298
UniGene
Hs.473893
Gene Chromosome Location
Chr.21: 41798225-41879482 on Build GRCh38
Transcripts
NM_001040424.2 NM_001282934.1 NM_022115.4 NR_104257.1 NR_104258.1 NR_104260.1 NR_135464.1 XM_006724039.2 XM_006724040.2 XM_011529674.1 XM_011529675.1 XM_011529676.2 XM_011529677.2 XM_011529678.2 XM_011529679.2 XM_011529681.2 XM_011529683.1 XM_017028425.1 XM_017028426.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5537557 c.3825C>T p.I1275I Chr.21: 41804540-41804540 on Build GRCh38
5537558 c.2787C>T p.I929I Chr.21: 41804540-41804540 on Build GRCh38
4491291 c.3786C>T p.L1262L Chr.21: 41804579-41804579 on Build GRCh38
6302917 c.3756C>T p.L1252L Chr.21: 41804609-41804609 on Build GRCh38
6302918 c.2718C>T p.L906L Chr.21: 41804609-41804609 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369714611 Chr.21: 41804596-41804596 on Build GRCh38
rs780173634 Chr.21: 41804780-41804780 on Build GRCh38
rs986803300 Chr.21: 41804672-41804672 on Build GRCh38
rs911211794 Chr.21: 41804711-41804711 on Build GRCh38
rs143284535 Chr.21: 41804561-41804561 on Build GRCh38
rs547339791 Chr.21: 41804628-41804628 on Build GRCh38
rs1034522862 Chr.21: 41804868-41804868 on Build GRCh38
rs951328830 Chr.21: 41804652-41804652 on Build GRCh38
rs781218417 Chr.21: 41804903-41804903 on Build GRCh38
rs1005958000 Chr.21: 41804611-41804611 on Build GRCh38
rs147029126 Chr.21: 41804609-41804609 on Build GRCh38
rs564156186 Chr.21: 41804585-41804585 on Build GRCh38
rs746817799 Chr.21: 41804626-41804626 on Build GRCh38
rs778474869 Chr.21: 41804607-41804607 on Build GRCh38
rs149203937 Chr.21: 41804539-41804539 on Build GRCh38
rs575645187 Chr.21: 41804929-41804929 on Build GRCh38
rs907096747 Chr.21: 41804588-41804588 on Build GRCh38
rs544290739 Chr.21: 41804555-41804555 on Build GRCh38
rs990267516 Chr.21: 41804744-41804744 on Build GRCh38
rs763159282 Chr.21: 41804577-41804577 on Build GRCh38
rs73373472 Chr.21: 41804642-41804642 on Build GRCh38
rs73373477 Chr.21: 41804687-41804687 on Build GRCh38
rs928596265 Chr.21: 41804857-41804857 on Build GRCh38
rs771909811 Chr.21: 41804540-41804540 on Build GRCh38
rs753297772 Chr.21: 41804599-41804599 on Build GRCh38
rs373317468 Chr.21: 41804558-41804558 on Build GRCh38
rs34506941 Chr.21: 41804860-41804860 on Build GRCh38
rs569233661 Chr.21: 41804928-41804928 on Build GRCh38
rs79751505 Chr.21: 41804597-41804597 on Build GRCh38
rs778101394 Chr.21: 41804515-41804515 on Build GRCh38
rs1055417187 Chr.21: 41804866-41804866 on Build GRCh38
rs935759465 Chr.21: 41804707-41804707 on Build GRCh38
rs570611160 Chr.21: 41804690-41804690 on Build GRCh38
rs942780720 Chr.21: 41804491-41804491 on Build GRCh38
rs764530806 Chr.21: 41804579-41804579 on Build GRCh38
rs115593311 Chr.21: 41804898-41804898 on Build GRCh38
rs894155615 Chr.21: 41804871-41804871 on Build GRCh38
rs553054364 Chr.21: 41804722-41804722 on Build GRCh38
rs760650245 Chr.21: 41804557-41804557 on Build GRCh38
rs539036361 Chr.21: 41804839-41804839 on Build GRCh38
rs945911507 Chr.21: 41804831-41804831 on Build GRCh38
rs377344671 Chr.21: 41804559-41804559 on Build GRCh38
rs144803804 Chr.21: 41804688-41804688 on Build GRCh38
rs772968682 Chr.21: 41804552-41804552 on Build GRCh38
rs918589660 Chr.21: 41804641-41804641 on Build GRCh38
rs571926246 Chr.21: 41804833-41804833 on Build GRCh38
rs763259278 Chr.21: 41804807-41804807 on Build GRCh38
rs370538051 Chr.21: 41804664-41804664 on Build GRCh38
rs375035916 Chr.21: 41804494-41804494 on Build GRCh38
rs369184784 Chr.21: 41804509-41804509 on Build GRCh38
rs751925272 Chr.21: 41804581-41804581 on Build GRCh38
rs758808892 Chr.21: 41804514-41804514 on Build GRCh38
rs955605970 Chr.21: 41804697-41804697 on Build GRCh38
rs573193326 Chr.21: 41804798-41804798 on Build GRCh38
rs2298687 Chr.21: 41804523-41804523 on Build GRCh38
rs371408887 Chr.21: 41804516-41804516 on Build GRCh38
rs753085770 Chr.21: 41804513-41804513 on Build GRCh38
rs777622520 Chr.21: 41804612-41804612 on Build GRCh38
rs73373482 Chr.21: 41804706-41804706 on Build GRCh38
rs199671559 Chr.21: 41804627-41804627 on Build GRCh38
rs752081023 Chr.21: 41804880-41804881 on Build GRCh38
rs200186659 Chr.21: 41804510-41804510 on Build GRCh38
rs752994704 Chr.21: 41804576-41804576 on Build GRCh38
rs748838695 Chr.21: 41804522-41804522 on Build GRCh38
rs61744413 Chr.21: 41804606-41804606 on Build GRCh38
rs891551993 Chr.21: 41804825-41804825 on Build GRCh38
rs755245021 Chr.21: 41804508-41804508 on Build GRCh38
rs974603290 Chr.21: 41804639-41804639 on Build GRCh38
rs779537573 Chr.21: 41804519-41804519 on Build GRCh38
rs542119415 Chr.21: 41804483-41804483 on Build GRCh38
rs774014532 Chr.21: 41804533-41804533 on Build GRCh38
rs866005588 Chr.21: 41804616-41804616 on Build GRCh38
rs374097080 Chr.21: 41804608-41804608 on Build GRCh38
rs759735465 Chr.21: 41804560-41804560 on Build GRCh38

View Full Product Details