Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 41989086-41989590 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZBTB21
Entrez Gene ID
49854
Gene Name
zinc finger and BTB domain containing 21
Gene Aliases
ZNF295
UniGene
Hs.434947
Gene Chromosome Location
Chr.21: 41986831-42010418 on Build GRCh38
Transcripts
NM_001098402.1 NM_001098403.1 NM_001320729.1 NM_001320731.1 NM_020727.4 XM_005261121.3 XM_011529588.2 XM_011529590.2 XM_017028360.1 XM_017028361.1

SNP Detail

SNP ID Chromosome Location
rs535299696 Chr.21: 41989304-41989304 on Build GRCh38
rs933968234 Chr.21: 41989426-41989426 on Build GRCh38
rs1002475214 Chr.21: 41989524-41989524 on Build GRCh38
rs1054337130 Chr.21: 41989283-41989283 on Build GRCh38
rs552918946 Chr.21: 41989489-41989489 on Build GRCh38
rs959673384 Chr.21: 41989340-41989340 on Build GRCh38
rs539547433 Chr.21: 41989478-41989478 on Build GRCh38
rs972772865 Chr.21: 41989418-41989418 on Build GRCh38
rs906990135 Chr.21: 41989490-41989490 on Build GRCh38
rs889084562 Chr.21: 41989264-41989264 on Build GRCh38
rs1037024096 Chr.21: 41989156-41989156 on Build GRCh38
rs903733718 Chr.21: 41989282-41989283 on Build GRCh38
rs942162406 Chr.21: 41989184-41989185 on Build GRCh38
rs777614936 Chr.21: 41989502-41989502 on Build GRCh38
rs369571175 Chr.21: 41989189-41989190 on Build GRCh38
rs747617664 Chr.21: 41989441-41989441 on Build GRCh38
rs183121328 Chr.21: 41989318-41989318 on Build GRCh38
rs1025131244 Chr.21: 41989542-41989542 on Build GRCh38
rs114605784 Chr.21: 41989326-41989326 on Build GRCh38
rs971015007 Chr.21: 41989327-41989327 on Build GRCh38
rs797017522 Chr.21: 41989481-41989481 on Build GRCh38
rs892271126 Chr.21: 41989493-41989493 on Build GRCh38
rs142361492 Chr.21: 41989432-41989432 on Build GRCh38
rs558232329 Chr.21: 41989181-41989181 on Build GRCh38
rs1000653699 Chr.21: 41989283-41989283 on Build GRCh38
rs368061733 Chr.21: 41989532-41989532 on Build GRCh38
rs780865974 Chr.21: 41989391-41989391 on Build GRCh38
rs917540842 Chr.21: 41989129-41989131 on Build GRCh38
rs919384336 Chr.21: 41989398-41989398 on Build GRCh38
rs911129646 Chr.21: 41989433-41989433 on Build GRCh38
rs1050975803 Chr.21: 41989270-41989270 on Build GRCh38
rs561773525 Chr.21: 41989188-41989189 on Build GRCh38
rs892870725 Chr.21: 41989310-41989310 on Build GRCh38
rs190930340 Chr.21: 41989346-41989346 on Build GRCh38
rs1054789218 Chr.21: 41989452-41989452 on Build GRCh38
rs897123808 Chr.21: 41989163-41989163 on Build GRCh38
rs566349807 Chr.21: 41989240-41989240 on Build GRCh38
rs201647386 Chr.21: 41989367-41989367 on Build GRCh38
rs941493203 Chr.21: 41989155-41989155 on Build GRCh38
rs576487615 Chr.21: 41989244-41989249 on Build GRCh38
rs1007120702 Chr.21: 41989273-41989273 on Build GRCh38
rs918691718 Chr.21: 41989421-41989421 on Build GRCh38
rs889739604 Chr.21: 41989272-41989272 on Build GRCh38
rs992290837 Chr.21: 41989330-41989330 on Build GRCh38
rs972227254 Chr.21: 41989398-41989398 on Build GRCh38
rs1010044079 Chr.21: 41989483-41989483 on Build GRCh38
rs933979944 Chr.21: 41989166-41989166 on Build GRCh38
rs186025464 Chr.21: 41989336-41989336 on Build GRCh38
rs969075691 Chr.21: 41989550-41989550 on Build GRCh38
rs538860517 Chr.21: 41989344-41989344 on Build GRCh38
rs897906706 Chr.21: 41989222-41989222 on Build GRCh38
rs10568487 Chr.21: 41989347-41989348 on Build GRCh38
rs755650117 Chr.21: 41989491-41989491 on Build GRCh38
rs1002506449 Chr.21: 41989329-41989329 on Build GRCh38
rs573170303 Chr.21: 41989527-41989527 on Build GRCh38

View Full Product Details