Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 42376463-42376969 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMPRSS3
Entrez Gene ID
64699
Gene Name
transmembrane protease, serine 3
Gene Aliases
DFNB10, DFNB8, ECHOS1, TADG12
UniGene
Hs.208600
Gene Chromosome Location
Chr.21: 42371887-42396846 on Build GRCh38
Transcripts
NM_001256317.1 NM_024022.2 NM_032404.2 NR_046020.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6316179 c.1301C>A p.A434E Chr.21: 42376686-42376686 on Build GRCh38
76769 c.1160C>A p.A387E Chr.21: 42376575-42376575 on Build GRCh38
6403978 c.1144C>A p.P382T Chr.21: 42376591-42376591 on Build GRCh38
4402651 c.1087C>T p.P363S Chr.21: 42376648-42376648 on Build GRCh38
352990 c.1161G>T p.A387A Chr.21: 42376574-42376574 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs560380002 Chr.21: 42376505-42376505 on Build GRCh38
rs375477507 Chr.21: 42376782-42376782 on Build GRCh38
rs776173937 Chr.21: 42376600-42376600 on Build GRCh38
rs760379793 Chr.21: 42376561-42376561 on Build GRCh38
rs978035535 Chr.21: 42376672-42376672 on Build GRCh38
rs202020952 Chr.21: 42376526-42376526 on Build GRCh38
rs567727376 Chr.21: 42376863-42376863 on Build GRCh38
rs770372956 Chr.21: 42376541-42376541 on Build GRCh38
rs978383411 Chr.21: 42376515-42376515 on Build GRCh38
rs193035630 Chr.21: 42376775-42376775 on Build GRCh38
rs184951299 Chr.21: 42376879-42376879 on Build GRCh38
rs111033537 Chr.21: 42376552-42376552 on Build GRCh38
rs727503492 Chr.21: 42376583-42376583 on Build GRCh38
rs747606494 Chr.21: 42376548-42376548 on Build GRCh38
rs180878237 Chr.21: 42376931-42376931 on Build GRCh38
rs113747896 Chr.21: 42376613-42376613 on Build GRCh38
rs568897245 Chr.21: 42376854-42376854 on Build GRCh38
rs1009019681 Chr.21: 42376586-42376586 on Build GRCh38
rs777243740 Chr.21: 42376661-42376661 on Build GRCh38
rs746293388 Chr.21: 42376577-42376577 on Build GRCh38
rs373365200 Chr.21: 42376903-42376903 on Build GRCh38
rs371920504 Chr.21: 42376919-42376919 on Build GRCh38
rs1044045779 Chr.21: 42376482-42376482 on Build GRCh38
rs527651051 Chr.21: 42376802-42376802 on Build GRCh38
rs891408559 Chr.21: 42376781-42376781 on Build GRCh38
rs371402895 Chr.21: 42376567-42376567 on Build GRCh38
rs145077154 Chr.21: 42376619-42376619 on Build GRCh38
rs769934547 Chr.21: 42376842-42376842 on Build GRCh38
rs756455958 Chr.21: 42376579-42376579 on Build GRCh38
rs1005640798 Chr.21: 42376878-42376878 on Build GRCh38
rs564663766 Chr.21: 42376810-42376810 on Build GRCh38
rs375580956 Chr.21: 42376726-42376726 on Build GRCh38
rs550324586 Chr.21: 42376840-42376840 on Build GRCh38
rs1005963165 Chr.21: 42376682-42376682 on Build GRCh38
rs35051860 Chr.21: 42376913-42376914 on Build GRCh38
rs111817056 Chr.21: 42376512-42376512 on Build GRCh38
rs147140159 Chr.21: 42376814-42376814 on Build GRCh38
rs760671977 Chr.21: 42376936-42376936 on Build GRCh38
rs749798053 Chr.21: 42376584-42376584 on Build GRCh38
rs764262141 Chr.21: 42376614-42376614 on Build GRCh38
rs750642034 Chr.21: 42376652-42376652 on Build GRCh38
rs13048440 Chr.21: 42376913-42376913 on Build GRCh38
rs962652474 Chr.21: 42376858-42376858 on Build GRCh38
rs778832786 Chr.21: 42376525-42376525 on Build GRCh38
rs767202382 Chr.21: 42376509-42376509 on Build GRCh38
rs766177584 Chr.21: 42376518-42376518 on Build GRCh38
rs769615599 Chr.21: 42376695-42376695 on Build GRCh38
rs371448615 Chr.21: 42376606-42376606 on Build GRCh38
rs749334208 Chr.21: 42376910-42376910 on Build GRCh38
rs775489548 Chr.21: 42376699-42376699 on Build GRCh38
rs780568459 Chr.21: 42376580-42376580 on Build GRCh38
rs727504304 Chr.21: 42376543-42376543 on Build GRCh38
rs902256395 Chr.21: 42376806-42376806 on Build GRCh38
rs755519837 Chr.21: 42376660-42376660 on Build GRCh38
rs201451028 Chr.21: 42376531-42376531 on Build GRCh38
rs985620612 Chr.21: 42376740-42376740 on Build GRCh38
rs930097107 Chr.21: 42376765-42376765 on Build GRCh38
rs55652649 Chr.21: 42376745-42376745 on Build GRCh38
rs999797220 Chr.21: 42376814-42376814 on Build GRCh38
rs745522641 Chr.21: 42376681-42376681 on Build GRCh38
rs999765835 Chr.21: 42376498-42376498 on Build GRCh38
rs760420559 Chr.21: 42376513-42376513 on Build GRCh38
rs374082620 Chr.21: 42376565-42376565 on Build GRCh38
rs770666997 Chr.21: 42376599-42376599 on Build GRCh38
rs773977655 Chr.21: 42376496-42376496 on Build GRCh38
rs780708451 Chr.21: 42376759-42376759 on Build GRCh38
rs151315833 Chr.21: 42376555-42376555 on Build GRCh38
rs754341063 Chr.21: 42376658-42376658 on Build GRCh38
rs756432794 Chr.21: 42376655-42376655 on Build GRCh38
rs767728685 Chr.21: 42376651-42376651 on Build GRCh38
rs1039951039 Chr.21: 42376927-42376927 on Build GRCh38
rs111033292 Chr.21: 42376607-42376607 on Build GRCh38
rs918764124 Chr.21: 42376761-42376761 on Build GRCh38
rs560318076 Chr.21: 42376785-42376785 on Build GRCh38
rs397517370 Chr.21: 42376645-42376645 on Build GRCh38
rs145766262 Chr.21: 42376563-42376563 on Build GRCh38
rs1078272 Chr.21: 42376804-42376804 on Build GRCh38
rs915066211 Chr.21: 42376493-42376493 on Build GRCh38
rs201172277 Chr.21: 42376575-42376575 on Build GRCh38
rs1016968797 Chr.21: 42376687-42376687 on Build GRCh38
rs13052961 Chr.21: 42376914-42376914 on Build GRCh38
rs757529665 Chr.21: 42376648-42376648 on Build GRCh38
rs781662983 Chr.21: 42376536-42376536 on Build GRCh38
rs755013014 Chr.21: 42376520-42376520 on Build GRCh38
rs200559617 Chr.21: 42376519-42376519 on Build GRCh38
rs397517369 Chr.21: 42376679-42376679 on Build GRCh38
rs727505305 Chr.21: 42376530-42376530 on Build GRCh38
rs757635909 Chr.21: 42376533-42376533 on Build GRCh38
rs772776809 Chr.21: 42376588-42376588 on Build GRCh38
rs987405584 Chr.21: 42376911-42376911 on Build GRCh38
rs746633574 Chr.21: 42376589-42376589 on Build GRCh38
rs577160419 Chr.21: 42376703-42376703 on Build GRCh38
rs200777266 Chr.21: 42376656-42376656 on Build GRCh38
rs895871845 Chr.21: 42376523-42376523 on Build GRCh38
rs1038745428 Chr.21: 42376784-42376784 on Build GRCh38
rs114156234 Chr.21: 42376727-42376727 on Build GRCh38
rs752332652 Chr.21: 42376680-42376680 on Build GRCh38
rs566668462 Chr.21: 42376894-42376894 on Build GRCh38
rs201425269 Chr.21: 42376537-42376537 on Build GRCh38
rs532114055 Chr.21: 42376833-42376833 on Build GRCh38
rs199751071 Chr.21: 42376673-42376673 on Build GRCh38
rs774452631 Chr.21: 42376721-42376721 on Build GRCh38
rs773711263 Chr.21: 42376545-42376545 on Build GRCh38
rs574546006 Chr.21: 42376941-42376941 on Build GRCh38
rs574609099 Chr.21: 42376763-42376763 on Build GRCh38
rs748363450 Chr.21: 42376834-42376834 on Build GRCh38
rs768759952 Chr.21: 42376708-42376708 on Build GRCh38
rs758488441 Chr.21: 42376568-42376568 on Build GRCh38
rs781376204 Chr.21: 42376576-42376576 on Build GRCh38
rs1022012928 Chr.21: 42376805-42376805 on Build GRCh38
rs140620963 Chr.21: 42376562-42376562 on Build GRCh38
rs188750127 Chr.21: 42376678-42376678 on Build GRCh38
rs568884207 Chr.21: 42376574-42376574 on Build GRCh38
rs768037982 Chr.21: 42376494-42376494 on Build GRCh38
rs941433875 Chr.21: 42376556-42376556 on Build GRCh38
rs775508015 Chr.21: 42376612-42376612 on Build GRCh38
rs771559352 Chr.21: 42376587-42376587 on Build GRCh38

View Full Product Details