Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 89526824-89527354 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC8B
Entrez Gene ID
23507
Gene Name
leucine rich repeat containing 8 family member B
Gene Aliases
TA-LRRP, TALRRP
UniGene
Hs.482017
Gene Chromosome Location
Chr.1: 89522627-89597861 on Build GRCh38
Transcripts
NM_001134476.1 XM_005270701.4 XM_005270702.4 XM_005270703.4 XM_011541143.2 XM_011541144.2 XM_011541145.2 XM_011541146.2 XM_011541148.2 XM_017000885.1 XM_017000886.1

SNP Detail

SNP ID Chromosome Location
rs1039618089 Chr.1: 89527184-89527184 on Build GRCh38
rs1033044173 Chr.1: 89526876-89526876 on Build GRCh38
rs776980747 Chr.1: 89527308-89527308 on Build GRCh38
rs941146550 Chr.1: 89527097-89527097 on Build GRCh38
rs532899715 Chr.1: 89527264-89527264 on Build GRCh38
rs922241895 Chr.1: 89527013-89527013 on Build GRCh38
rs755946773 Chr.1: 89527062-89527062 on Build GRCh38
rs527944595 Chr.1: 89527319-89527319 on Build GRCh38
rs1007724653 Chr.1: 89526941-89526941 on Build GRCh38
rs539598774 Chr.1: 89527307-89527307 on Build GRCh38
rs915710084 Chr.1: 89527024-89527024 on Build GRCh38
rs934313236 Chr.1: 89527072-89527072 on Build GRCh38
rs550800727 Chr.1: 89527165-89527165 on Build GRCh38
rs982472017 Chr.1: 89527032-89527032 on Build GRCh38
rs72957862 Chr.1: 89526972-89526972 on Build GRCh38
rs187330854 Chr.1: 89527112-89527112 on Build GRCh38
rs562434755 Chr.1: 89527195-89527195 on Build GRCh38
rs981358230 Chr.1: 89527039-89527039 on Build GRCh38
rs1016390305 Chr.1: 89527282-89527282 on Build GRCh38
rs1030752169 Chr.1: 89526854-89526854 on Build GRCh38
rs12409358 Chr.1: 89527275-89527275 on Build GRCh38
rs970380104 Chr.1: 89527025-89527025 on Build GRCh38
rs1008066141 Chr.1: 89527260-89527260 on Build GRCh38
rs41313391 Chr.1: 89527168-89527168 on Build GRCh38
rs1004984789 Chr.1: 89526906-89526906 on Build GRCh38
rs997705436 Chr.1: 89527281-89527281 on Build GRCh38
rs566153200 Chr.1: 89527292-89527292 on Build GRCh38
rs780329207 Chr.1: 89527273-89527273 on Build GRCh38
rs976347618 Chr.1: 89527000-89527000 on Build GRCh38
rs1045605750 Chr.1: 89527196-89527196 on Build GRCh38
rs1054426357 Chr.1: 89527233-89527233 on Build GRCh38
rs778611708 Chr.1: 89527112-89527113 on Build GRCh38
rs1045674111 Chr.1: 89526848-89526848 on Build GRCh38
rs111739774 Chr.1: 89527237-89527237 on Build GRCh38
rs957721174 Chr.1: 89526903-89526903 on Build GRCh38
rs749120706 Chr.1: 89527253-89527253 on Build GRCh38
rs951006504 Chr.1: 89527027-89527027 on Build GRCh38
rs934655701 Chr.1: 89527232-89527232 on Build GRCh38
rs915736834 Chr.1: 89527140-89527140 on Build GRCh38
rs1052824250 Chr.1: 89527073-89527073 on Build GRCh38
rs998836879 Chr.1: 89526857-89526857 on Build GRCh38
rs1012290774 Chr.1: 89527302-89527302 on Build GRCh38
rs928302814 Chr.1: 89527048-89527048 on Build GRCh38
rs573150635 Chr.1: 89526889-89526889 on Build GRCh38
rs903137198 Chr.1: 89527228-89527228 on Build GRCh38
rs192731509 Chr.1: 89527139-89527139 on Build GRCh38
rs963462679 Chr.1: 89527284-89527284 on Build GRCh38
rs903205597 Chr.1: 89526850-89526850 on Build GRCh38
rs376161256 Chr.1: 89526948-89526948 on Build GRCh38
rs902029668 Chr.1: 89527280-89527280 on Build GRCh38
rs560503903 Chr.1: 89526846-89526846 on Build GRCh38
rs774013498 Chr.1: 89527274-89527274 on Build GRCh38
rs900630743 Chr.1: 89527224-89527224 on Build GRCh38
rs149986293 Chr.1: 89526946-89526946 on Build GRCh38

View Full Product Details