Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 39434835-39435331 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100506472
Entrez Gene ID
100506472
Gene Name
uncharacterized LOC100506472
Gene Aliases
-
UniGene
Hs.729080
Gene Chromosome Location
Chr.22: 39432160-39437128 on Build GRCh38
Transcripts
NR_040535.1 NM_153497.2

Gene Symbol
TAB1
Entrez Gene ID
10454
Gene Name
TGF-beta activated kinase 1/MAP3K7 binding protein 1
Gene Aliases
3'-Tab1, MAP3K7IP1
UniGene
Hs.507681
Gene Chromosome Location
Chr.22: 39399754-39437127 on Build GRCh38
Transcripts
NR_040535.1 NM_153497.2

SNP Detail

SNP ID Chromosome Location
rs750283649 Chr.22: 39435303-39435303 on Build GRCh38
rs1000657857 Chr.22: 39434881-39434881 on Build GRCh38
rs573047446 Chr.22: 39435016-39435016 on Build GRCh38
rs971704528 Chr.22: 39434920-39434920 on Build GRCh38
rs528207528 Chr.22: 39434947-39434947 on Build GRCh38
rs377337009 Chr.22: 39435087-39435087 on Build GRCh38
rs958748452 Chr.22: 39435120-39435120 on Build GRCh38
rs899498692 Chr.22: 39435038-39435038 on Build GRCh38
rs137870709 Chr.22: 39434861-39434861 on Build GRCh38
rs5750825 Chr.22: 39435273-39435273 on Build GRCh38
rs957532136 Chr.22: 39434888-39434888 on Build GRCh38
rs901626605 Chr.22: 39435134-39435135 on Build GRCh38
rs999075807 Chr.22: 39435189-39435189 on Build GRCh38
rs919907592 Chr.22: 39435128-39435128 on Build GRCh38
rs929828479 Chr.22: 39434972-39434972 on Build GRCh38
rs17001112 Chr.22: 39435225-39435225 on Build GRCh38
rs1034982801 Chr.22: 39435106-39435106 on Build GRCh38
rs971971970 Chr.22: 39435102-39435102 on Build GRCh38
rs982819334 Chr.22: 39435212-39435212 on Build GRCh38
rs866049268 Chr.22: 39435213-39435213 on Build GRCh38
rs988956959 Chr.22: 39434907-39434907 on Build GRCh38
rs1022966431 Chr.22: 39435024-39435024 on Build GRCh38
rs563817478 Chr.22: 39434860-39434860 on Build GRCh38
rs546380193 Chr.22: 39434989-39434989 on Build GRCh38
rs1000497907 Chr.22: 39435061-39435061 on Build GRCh38
rs1038881817 Chr.22: 39435129-39435129 on Build GRCh38
rs568293255 Chr.22: 39435076-39435076 on Build GRCh38
rs1042077646 Chr.22: 39435141-39435141 on Build GRCh38
rs902152401 Chr.22: 39435173-39435173 on Build GRCh38
rs981721987 Chr.22: 39435104-39435104 on Build GRCh38
rs190478541 Chr.22: 39435068-39435068 on Build GRCh38
rs933051659 Chr.22: 39435140-39435140 on Build GRCh38
rs1008179780 Chr.22: 39435000-39435000 on Build GRCh38
rs927356096 Chr.22: 39434937-39434937 on Build GRCh38
rs1021030291 Chr.22: 39434863-39434863 on Build GRCh38
rs34413791 Chr.22: 39435143-39435144 on Build GRCh38
rs149460488 Chr.22: 39434938-39434938 on Build GRCh38
rs1032114620 Chr.22: 39435066-39435066 on Build GRCh38
rs1038766496 Chr.22: 39434854-39434854 on Build GRCh38
rs930088553 Chr.22: 39435182-39435182 on Build GRCh38
rs1053478261 Chr.22: 39435286-39435286 on Build GRCh38
rs929265849 Chr.22: 39435094-39435094 on Build GRCh38
rs1040021996 Chr.22: 39435005-39435005 on Build GRCh38
rs913386300 Chr.22: 39434915-39434915 on Build GRCh38
rs974020335 Chr.22: 39435125-39435125 on Build GRCh38
rs375394511 Chr.22: 39435267-39435267 on Build GRCh38
rs1032054522 Chr.22: 39434884-39434884 on Build GRCh38
rs982095408 Chr.22: 39434926-39434926 on Build GRCh38
rs1046902739 Chr.22: 39434975-39434975 on Build GRCh38
rs981501047 Chr.22: 39435054-39435054 on Build GRCh38
rs577595570 Chr.22: 39435284-39435284 on Build GRCh38
rs971886686 Chr.22: 39435051-39435051 on Build GRCh38
rs903463036 Chr.22: 39434883-39434885 on Build GRCh38
rs761582101 Chr.22: 39435135-39435135 on Build GRCh38
rs117093889 Chr.22: 39435099-39435099 on Build GRCh38
rs919983441 Chr.22: 39434961-39434961 on Build GRCh38
rs763697777 Chr.22: 39435088-39435088 on Build GRCh38

View Full Product Details