Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 41396051-41396540 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TEF
Entrez Gene ID
7008
Gene Name
TEF, PAR bZIP transcription factor
Gene Aliases
-
UniGene
Hs.181159
Gene Chromosome Location
Chr.22: 41367333-41399328 on Build GRCh38
Transcripts
NM_001145398.2 NM_003216.3

SNP Detail

SNP ID Chromosome Location
rs113140298 Chr.22: 41396476-41396476 on Build GRCh38
rs753291357 Chr.22: 41396389-41396389 on Build GRCh38
rs1010647593 Chr.22: 41396388-41396388 on Build GRCh38
rs1028508919 Chr.22: 41396386-41396386 on Build GRCh38
rs529755234 Chr.22: 41396354-41396354 on Build GRCh38
rs928202742 Chr.22: 41396422-41396422 on Build GRCh38
rs916666072 Chr.22: 41396480-41396480 on Build GRCh38
rs540599272 Chr.22: 41396258-41396258 on Build GRCh38
rs1038553165 Chr.22: 41396265-41396265 on Build GRCh38
rs139035623 Chr.22: 41396430-41396433 on Build GRCh38
rs368243386 Chr.22: 41396219-41396219 on Build GRCh38
rs182267390 Chr.22: 41396130-41396130 on Build GRCh38
rs535899304 Chr.22: 41396084-41396084 on Build GRCh38
rs74503034 Chr.22: 41396129-41396129 on Build GRCh38
rs868289230 Chr.22: 41396520-41396520 on Build GRCh38
rs766523599 Chr.22: 41396246-41396246 on Build GRCh38
rs566310711 Chr.22: 41396098-41396098 on Build GRCh38
rs1049023843 Chr.22: 41396284-41396284 on Build GRCh38
rs8136493 Chr.22: 41396278-41396278 on Build GRCh38
rs534116596 Chr.22: 41396126-41396126 on Build GRCh38
rs17002425 Chr.22: 41396203-41396203 on Build GRCh38
rs957164150 Chr.22: 41396364-41396365 on Build GRCh38
rs756541840 Chr.22: 41396415-41396415 on Build GRCh38
rs143478818 Chr.22: 41396229-41396229 on Build GRCh38
rs187570422 Chr.22: 41396134-41396134 on Build GRCh38
rs915828589 Chr.22: 41396376-41396376 on Build GRCh38
rs928092003 Chr.22: 41396390-41396390 on Build GRCh38
rs542712286 Chr.22: 41396199-41396199 on Build GRCh38
rs994812375 Chr.22: 41396076-41396076 on Build GRCh38
rs763176366 Chr.22: 41396238-41396238 on Build GRCh38
rs895194347 Chr.22: 41396242-41396242 on Build GRCh38
rs550808241 Chr.22: 41396320-41396320 on Build GRCh38
rs1049118255 Chr.22: 41396497-41396497 on Build GRCh38
rs990982415 Chr.22: 41396456-41396456 on Build GRCh38
rs17002428 Chr.22: 41396440-41396440 on Build GRCh38
rs998723003 Chr.22: 41396357-41396357 on Build GRCh38
rs1015411806 Chr.22: 41396123-41396123 on Build GRCh38
rs556391726 Chr.22: 41396184-41396184 on Build GRCh38
rs192666274 Chr.22: 41396264-41396264 on Build GRCh38
rs754195460 Chr.22: 41396436-41396436 on Build GRCh38
rs759722224 Chr.22: 41396336-41396336 on Build GRCh38
rs939449978 Chr.22: 41396414-41396414 on Build GRCh38
rs963650721 Chr.22: 41396071-41396071 on Build GRCh38
rs1055211492 Chr.22: 41396072-41396072 on Build GRCh38
rs530369480 Chr.22: 41396365-41396365 on Build GRCh38
rs1055623499 Chr.22: 41396231-41396231 on Build GRCh38
rs112460788 Chr.22: 41396091-41396091 on Build GRCh38
rs764571112 Chr.22: 41396418-41396418 on Build GRCh38
rs768479967 Chr.22: 41396174-41396174 on Build GRCh38
rs562702981 Chr.22: 41396339-41396339 on Build GRCh38
rs9623390 Chr.22: 41396511-41396511 on Build GRCh38
rs947995536 Chr.22: 41396141-41396141 on Build GRCh38
rs1015464065 Chr.22: 41396330-41396330 on Build GRCh38
rs533588750 Chr.22: 41396492-41396492 on Build GRCh38
rs895169061 Chr.22: 41396074-41396074 on Build GRCh38
rs953791026 Chr.22: 41396387-41396387 on Build GRCh38
rs552143589 Chr.22: 41396493-41396493 on Build GRCh38
rs183662622 Chr.22: 41396429-41396429 on Build GRCh38
rs1004110473 Chr.22: 41396322-41396322 on Build GRCh38
rs767522447 Chr.22: 41396486-41396486 on Build GRCh38
rs1031941149 Chr.22: 41396361-41396361 on Build GRCh38
rs575809638 Chr.22: 41396192-41396192 on Build GRCh38
rs568650954 Chr.22: 41396079-41396079 on Build GRCh38
rs915508179 Chr.22: 41396138-41396138 on Build GRCh38
rs965224140 Chr.22: 41396125-41396125 on Build GRCh38

View Full Product Details