Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 42382725-42383219 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NFAM1
Entrez Gene ID
150372
Gene Name
NFAT activating protein with ITAM motif 1
Gene Aliases
CNAIP
UniGene
Hs.436677
Gene Chromosome Location
Chr.22: 42380407-42438155 on Build GRCh38
Transcripts
NM_001318323.1 NM_145912.6 XM_005261374.4

SNP Detail

SNP ID Chromosome Location
rs150754152 Chr.22: 42383021-42383021 on Build GRCh38
rs1010742073 Chr.22: 42383050-42383050 on Build GRCh38
rs41277491 Chr.22: 42382900-42382900 on Build GRCh38
rs763332117 Chr.22: 42382928-42382928 on Build GRCh38
rs992072462 Chr.22: 42383051-42383051 on Build GRCh38
rs968479369 Chr.22: 42382791-42382791 on Build GRCh38
rs529488168 Chr.22: 42382946-42382946 on Build GRCh38
rs868677648 Chr.22: 42382802-42382802 on Build GRCh38
rs995601088 Chr.22: 42382855-42382855 on Build GRCh38
rs866786766 Chr.22: 42382976-42382976 on Build GRCh38
rs966739048 Chr.22: 42382885-42382885 on Build GRCh38
rs567496709 Chr.22: 42382755-42382755 on Build GRCh38
rs542344880 Chr.22: 42382926-42382926 on Build GRCh38
rs112856367 Chr.22: 42383022-42383022 on Build GRCh38
rs1049870695 Chr.22: 42383023-42383023 on Build GRCh38
rs931045902 Chr.22: 42383087-42383087 on Build GRCh38
rs763740341 Chr.22: 42382877-42382877 on Build GRCh38
rs535077385 Chr.22: 42383052-42383052 on Build GRCh38
rs1022219915 Chr.22: 42382843-42382843 on Build GRCh38
rs898232745 Chr.22: 42382854-42382854 on Build GRCh38
rs998255970 Chr.22: 42382803-42382803 on Build GRCh38
rs534808309 Chr.22: 42382778-42382778 on Build GRCh38
rs534147325 Chr.22: 42382796-42382797 on Build GRCh38
rs771609654 Chr.22: 42382779-42382779 on Build GRCh38
rs571390848 Chr.22: 42383148-42383148 on Build GRCh38
rs533479412 Chr.22: 42382975-42382975 on Build GRCh38
rs752834232 Chr.22: 42383165-42383165 on Build GRCh38
rs374319363 Chr.22: 42382797-42382799 on Build GRCh38
rs141138393 Chr.22: 42382958-42382958 on Build GRCh38
rs113414543 Chr.22: 42383075-42383075 on Build GRCh38
rs114472822 Chr.22: 42382833-42382833 on Build GRCh38
rs1034664831 Chr.22: 42383135-42383135 on Build GRCh38
rs1034736839 Chr.22: 42382906-42382906 on Build GRCh38
rs1008154164 Chr.22: 42382750-42382750 on Build GRCh38
rs115035758 Chr.22: 42382973-42382973 on Build GRCh38
rs1056707725 Chr.22: 42382805-42382805 on Build GRCh38
rs889583571 Chr.22: 42383040-42383040 on Build GRCh38
rs541573752 Chr.22: 42383114-42383114 on Build GRCh38
rs1036801334 Chr.22: 42383071-42383071 on Build GRCh38
rs1057157 Chr.22: 42383012-42383012 on Build GRCh38
rs552018366 Chr.22: 42382998-42382998 on Build GRCh38
rs560650529 Chr.22: 42382929-42382929 on Build GRCh38
rs148205809 Chr.22: 42382812-42382812 on Build GRCh38
rs1019090862 Chr.22: 42382754-42382754 on Build GRCh38
rs552894618 Chr.22: 42383088-42383088 on Build GRCh38
rs767622007 Chr.22: 42383136-42383136 on Build GRCh38
rs184300545 Chr.22: 42383007-42383007 on Build GRCh38
rs752073148 Chr.22: 42383026-42383026 on Build GRCh38
rs553271971 Chr.22: 42382788-42382788 on Build GRCh38
rs556702258 Chr.22: 42382891-42382891 on Build GRCh38
rs1036771556 Chr.22: 42382846-42382846 on Build GRCh38
rs916058878 Chr.22: 42382959-42382959 on Build GRCh38
rs1040675972 Chr.22: 42383153-42383153 on Build GRCh38
rs939881213 Chr.22: 42383067-42383067 on Build GRCh38
rs1037054408 Chr.22: 42382792-42382792 on Build GRCh38
rs889664818 Chr.22: 42382816-42382816 on Build GRCh38

View Full Product Details