Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 44724862-44725352 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRR5
Entrez Gene ID
55615
Gene Name
proline rich 5
Gene Aliases
FLJ20185k, PP610, PROTOR-1, PROTOR1
UniGene
Hs.102336
Gene Chromosome Location
Chr.22: 44668547-44737681 on Build GRCh38
Transcripts
NM_001017528.2 NM_001017529.2 NM_001017530.1 NM_001198721.1 NM_015366.3 NM_181333.3 NM_181334.5

Gene Symbol
PRR5-ARHGAP8
Entrez Gene ID
553158
Gene Name
PRR5-ARHGAP8 readthrough
Gene Aliases
-
UniGene
Hs.102336
Gene Chromosome Location
Chr.22: 44702198-44862784 on Build GRCh38
Transcripts
NM_001017528.2 NM_001017529.2 NM_001017530.1 NM_001198721.1 NM_015366.3 NM_181333.3 NM_181334.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7153712 c.205G>C p.E69Q Chr.22: 44725260-44725260 on Build GRCh38
7106109 c.246C>T p.F82F Chr.22: 44725274-44725274 on Build GRCh38
7153711 c.232G>C p.E78Q Chr.22: 44725260-44725260 on Build GRCh38
7106108 c.315C>T p.F105F Chr.22: 44725274-44725274 on Build GRCh38
7153710 c.301G>C p.E101Q Chr.22: 44725260-44725260 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200927358 Chr.22: 44725217-44725217 on Build GRCh38
rs551231284 Chr.22: 44725029-44725029 on Build GRCh38
rs544745890 Chr.22: 44724956-44724956 on Build GRCh38
rs1039477326 Chr.22: 44724915-44724915 on Build GRCh38
rs759513633 Chr.22: 44725229-44725229 on Build GRCh38
rs761514470 Chr.22: 44725268-44725268 on Build GRCh38
rs933118693 Chr.22: 44725089-44725089 on Build GRCh38
rs747132044 Chr.22: 44725209-44725209 on Build GRCh38
rs572601726 Chr.22: 44724950-44724950 on Build GRCh38
rs35530859 Chr.22: 44725056-44725057 on Build GRCh38
rs892477457 Chr.22: 44724900-44724900 on Build GRCh38
rs772951980 Chr.22: 44724988-44724988 on Build GRCh38
rs914447294 Chr.22: 44725093-44725093 on Build GRCh38
rs896660598 Chr.22: 44725180-44725180 on Build GRCh38
rs376694994 Chr.22: 44725317-44725317 on Build GRCh38
rs368159230 Chr.22: 44725230-44725230 on Build GRCh38
rs753524437 Chr.22: 44725266-44725266 on Build GRCh38
rs541315180 Chr.22: 44725287-44725287 on Build GRCh38
rs1009537142 Chr.22: 44724920-44724920 on Build GRCh38
rs989945824 Chr.22: 44725066-44725066 on Build GRCh38
rs557153870 Chr.22: 44725279-44725279 on Build GRCh38
rs770835424 Chr.22: 44725225-44725225 on Build GRCh38
rs907421158 Chr.22: 44725293-44725293 on Build GRCh38
rs899557538 Chr.22: 44724928-44724928 on Build GRCh38
rs145196799 Chr.22: 44725248-44725248 on Build GRCh38
rs41368848 Chr.22: 44725058-44725058 on Build GRCh38
rs1022390765 Chr.22: 44724946-44724946 on Build GRCh38
rs943734500 Chr.22: 44725158-44725158 on Build GRCh38
rs200344209 Chr.22: 44725315-44725315 on Build GRCh38
rs761238842 Chr.22: 44725139-44725139 on Build GRCh38
rs139168828 Chr.22: 44725280-44725280 on Build GRCh38
rs904040196 Chr.22: 44725074-44725074 on Build GRCh38
rs908876282 Chr.22: 44725045-44725045 on Build GRCh38
rs184950947 Chr.22: 44724958-44724958 on Build GRCh38
rs192849195 Chr.22: 44724935-44724935 on Build GRCh38
rs368029285 Chr.22: 44725009-44725009 on Build GRCh38
rs531015049 Chr.22: 44724967-44724967 on Build GRCh38
rs947283476 Chr.22: 44725129-44725129 on Build GRCh38
rs1043782225 Chr.22: 44725146-44725146 on Build GRCh38
rs974443924 Chr.22: 44725072-44725072 on Build GRCh38
rs1026948463 Chr.22: 44725017-44725017 on Build GRCh38
rs951004345 Chr.22: 44725213-44725213 on Build GRCh38
rs763587713 Chr.22: 44725259-44725259 on Build GRCh38
rs1053047975 Chr.22: 44725102-44725102 on Build GRCh38
rs1033876832 Chr.22: 44724957-44724957 on Build GRCh38
rs552705079 Chr.22: 44725202-44725202 on Build GRCh38
rs1026482951 Chr.22: 44725204-44725204 on Build GRCh38
rs546673303 Chr.22: 44725061-44725061 on Build GRCh38
rs748357549 Chr.22: 44725330-44725330 on Build GRCh38
rs774815902 Chr.22: 44725238-44725238 on Build GRCh38
rs370453693 Chr.22: 44725073-44725073 on Build GRCh38
rs371763329 Chr.22: 44725303-44725303 on Build GRCh38
rs536868117 Chr.22: 44725226-44725226 on Build GRCh38
rs1002309238 Chr.22: 44725174-44725174 on Build GRCh38
rs751523837 Chr.22: 44725308-44725308 on Build GRCh38
rs780621389 Chr.22: 44725289-44725289 on Build GRCh38
rs903842054 Chr.22: 44725154-44725154 on Build GRCh38
rs912449031 Chr.22: 44725142-44725142 on Build GRCh38
rs1015908274 Chr.22: 44725241-44725241 on Build GRCh38
rs977397412 Chr.22: 44725124-44725124 on Build GRCh38
rs1005448081 Chr.22: 44725271-44725271 on Build GRCh38
rs891668517 Chr.22: 44725120-44725120 on Build GRCh38
rs758865281 Chr.22: 44725286-44725286 on Build GRCh38
rs888031782 Chr.22: 44724983-44724983 on Build GRCh38
rs1002884500 Chr.22: 44725331-44725331 on Build GRCh38
rs752789216 Chr.22: 44725272-44725272 on Build GRCh38
rs765256930 Chr.22: 44725222-44725223 on Build GRCh38
rs201231073 Chr.22: 44725312-44725312 on Build GRCh38
rs748445191 Chr.22: 44725219-44725219 on Build GRCh38
rs370419846 Chr.22: 44725201-44725201 on Build GRCh38
rs950392888 Chr.22: 44725038-44725038 on Build GRCh38
rs150774541 Chr.22: 44725044-44725044 on Build GRCh38
rs961136935 Chr.22: 44724966-44724966 on Build GRCh38
rs768700455 Chr.22: 44725212-44725212 on Build GRCh38
rs373536864 Chr.22: 44725179-44725179 on Build GRCh38
rs1003262346 Chr.22: 44725057-44725057 on Build GRCh38

View Full Product Details