Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 46366294-46366788 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELSR1
Entrez Gene ID
9620
Gene Name
cadherin EGF LAG seven-pass G-type receptor 1
Gene Aliases
ADGRC1, CDHF9, FMI2, HFMI2, ME2
UniGene
Hs.252387
Gene Chromosome Location
Chr.22: 46360834-46537502 on Build GRCh38
Transcripts
NM_014246.1 XM_006724383.3 XM_011530554.2 XM_011530555.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6316276 c.8238C>A p.D2746E Chr.22: 46366448-46366448 on Build GRCh38
5435544 c.8233G>A p.G2745S Chr.22: 46366453-46366453 on Build GRCh38
6585539 c.8245G>C p.D2749H Chr.22: 46366441-46366441 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs777036938 Chr.22: 46366349-46366349 on Build GRCh38
rs373749197 Chr.22: 46366370-46366370 on Build GRCh38
rs1037835173 Chr.22: 46366441-46366441 on Build GRCh38
rs765095219 Chr.22: 46366525-46366525 on Build GRCh38
rs1001806394 Chr.22: 46366650-46366650 on Build GRCh38
rs4635638 Chr.22: 46366638-46366638 on Build GRCh38
rs553721996 Chr.22: 46366570-46366570 on Build GRCh38
rs562394666 Chr.22: 46366647-46366647 on Build GRCh38
rs576937168 Chr.22: 46366590-46366590 on Build GRCh38
rs771193237 Chr.22: 46366447-46366447 on Build GRCh38
rs553541931 Chr.22: 46366631-46366631 on Build GRCh38
rs538998117 Chr.22: 46366378-46366378 on Build GRCh38
rs770301403 Chr.22: 46366360-46366360 on Build GRCh38
rs201956319 Chr.22: 46366547-46366548 on Build GRCh38
rs934965000 Chr.22: 46366580-46366580 on Build GRCh38
rs375345581 Chr.22: 46366443-46366443 on Build GRCh38
rs762273382 Chr.22: 46366353-46366353 on Build GRCh38
rs34100600 Chr.22: 46366564-46366565 on Build GRCh38
rs112587264 Chr.22: 46366538-46366538 on Build GRCh38
rs73889254 Chr.22: 46366317-46366317 on Build GRCh38
rs778864028 Chr.22: 46366391-46366391 on Build GRCh38
rs992067594 Chr.22: 46366367-46366367 on Build GRCh38
rs1022939784 Chr.22: 46366634-46366634 on Build GRCh38
rs183490268 Chr.22: 46366556-46366556 on Build GRCh38
rs1034675766 Chr.22: 46366683-46366683 on Build GRCh38
rs757174874 Chr.22: 46366390-46366390 on Build GRCh38
rs144039991 Chr.22: 46366404-46366404 on Build GRCh38
rs943541735 Chr.22: 46366438-46366438 on Build GRCh38
rs1025316799 Chr.22: 46366483-46366483 on Build GRCh38
rs902107194 Chr.22: 46366399-46366399 on Build GRCh38
rs111934710 Chr.22: 46366343-46366343 on Build GRCh38
rs532711227 Chr.22: 46366759-46366759 on Build GRCh38
rs943319891 Chr.22: 46366558-46366558 on Build GRCh38
rs781447851 Chr.22: 46366431-46366431 on Build GRCh38
rs1047844011 Chr.22: 46366606-46366606 on Build GRCh38
rs779627756 Chr.22: 46366341-46366341 on Build GRCh38
rs186247668 Chr.22: 46366364-46366364 on Build GRCh38
rs770223437 Chr.22: 46366434-46366434 on Build GRCh38
rs957895419 Chr.22: 46366667-46366667 on Build GRCh38
rs925024450 Chr.22: 46366381-46366381 on Build GRCh38
rs749952660 Chr.22: 46366403-46366403 on Build GRCh38
rs950142105 Chr.22: 46366761-46366761 on Build GRCh38
rs74699704 Chr.22: 46366448-46366448 on Build GRCh38
rs568874367 Chr.22: 46366439-46366439 on Build GRCh38
rs985041208 Chr.22: 46366717-46366717 on Build GRCh38
rs779531463 Chr.22: 46366409-46366409 on Build GRCh38
rs140990388 Chr.22: 46366432-46366432 on Build GRCh38
rs893265652 Chr.22: 46366604-46366604 on Build GRCh38
rs148905592 Chr.22: 46366470-46366470 on Build GRCh38
rs910645927 Chr.22: 46366557-46366557 on Build GRCh38
rs1040413991 Chr.22: 46366559-46366559 on Build GRCh38
rs1046948223 Chr.22: 46366320-46366320 on Build GRCh38
rs934663413 Chr.22: 46366736-46366736 on Build GRCh38
rs927894482 Chr.22: 46366321-46366321 on Build GRCh38
rs919130307 Chr.22: 46366537-46366537 on Build GRCh38
rs190965610 Chr.22: 46366445-46366445 on Build GRCh38
rs773193859 Chr.22: 46366362-46366362 on Build GRCh38
rs56325154 Chr.22: 46366463-46366463 on Build GRCh38
rs987965181 Chr.22: 46366673-46366673 on Build GRCh38
rs750431500 Chr.22: 46366531-46366531 on Build GRCh38
rs547055976 Chr.22: 46366719-46366719 on Build GRCh38
rs114561229 Chr.22: 46366637-46366637 on Build GRCh38
rs976075738 Chr.22: 46366720-46366720 on Build GRCh38
rs116625238 Chr.22: 46366707-46366707 on Build GRCh38
rs141907986 Chr.22: 46366363-46366363 on Build GRCh38
rs1014208832 Chr.22: 46366478-46366478 on Build GRCh38
rs146465806 Chr.22: 46366417-46366417 on Build GRCh38
rs932953352 Chr.22: 46366412-46366412 on Build GRCh38
rs960899589 Chr.22: 46366685-46366685 on Build GRCh38
rs759879880 Chr.22: 46366454-46366454 on Build GRCh38
rs770626227 Chr.22: 46366483-46366486 on Build GRCh38
rs111324092 Chr.22: 46366750-46366750 on Build GRCh38
rs368583642 Chr.22: 46366339-46366339 on Build GRCh38
rs879407649 Chr.22: 46366705-46366705 on Build GRCh38
rs981029533 Chr.22: 46366502-46366502 on Build GRCh38
rs527538316 Chr.22: 46366706-46366706 on Build GRCh38
rs112707306 Chr.22: 46366535-46366535 on Build GRCh38
rs947555906 Chr.22: 46366748-46366748 on Build GRCh38
rs574890397 Chr.22: 46366686-46366686 on Build GRCh38
rs929512810 Chr.22: 46366724-46366724 on Build GRCh38
rs763845724 Chr.22: 46366377-46366377 on Build GRCh38
rs756068197 Chr.22: 46366352-46366352 on Build GRCh38
rs187504981 Chr.22: 46366688-46366688 on Build GRCh38
rs77823653 Chr.22: 46366369-46366369 on Build GRCh38
rs746719085 Chr.22: 46366413-46366413 on Build GRCh38
rs1053340782 Chr.22: 46366741-46366741 on Build GRCh38
rs920835105 Chr.22: 46366721-46366721 on Build GRCh38
rs929460238 Chr.22: 46366600-46366600 on Build GRCh38
rs895899379 Chr.22: 46366608-46366608 on Build GRCh38
rs753600009 Chr.22: 46366464-46366464 on Build GRCh38
rs969291188 Chr.22: 46366491-46366491 on Build GRCh38
rs923307678 Chr.22: 46366731-46366731 on Build GRCh38
rs957843773 Chr.22: 46366532-46366532 on Build GRCh38
rs893392429 Chr.22: 46366744-46366744 on Build GRCh38
rs4541339 Chr.22: 46366407-46366407 on Build GRCh38
rs1032145259 Chr.22: 46366504-46366504 on Build GRCh38
rs910614700 Chr.22: 46366718-46366718 on Build GRCh38
rs964719567 Chr.22: 46366716-46366716 on Build GRCh38

View Full Product Details