Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 10925690-10926194 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC6A11
Entrez Gene ID
6538
Gene Name
solute carrier family 6 member 11
Gene Aliases
GAT-3, GAT3, GAT4
UniGene
Hs.655078
Gene Chromosome Location
Chr.3: 10816200-10940733 on Build GRCh38
Transcripts
NM_014229.2 XM_011534033.2 XM_017007073.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
378760 c.1219G>C p.G407R Chr.3: 10926102-10926102 on Build GRCh38
6095792 c.1137T>C p.F379F Chr.3: 10926020-10926020 on Build GRCh38
4925496 c.1133C>A p.A378D Chr.3: 10926016-10926016 on Build GRCh38
6544242 c.1167G>T p.M389I Chr.3: 10926050-10926050 on Build GRCh38
5033275 c.1125C>T p.P375P Chr.3: 10926008-10926008 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140515744 Chr.3: 10926062-10926062 on Build GRCh38
rs550018389 Chr.3: 10925852-10925852 on Build GRCh38
rs968373786 Chr.3: 10925960-10925960 on Build GRCh38
rs772462891 Chr.3: 10926160-10926160 on Build GRCh38
rs761615989 Chr.3: 10925973-10925973 on Build GRCh38
rs751498777 Chr.3: 10926069-10926069 on Build GRCh38
rs766365191 Chr.3: 10925991-10925991 on Build GRCh38
rs772471540 Chr.3: 10926032-10926032 on Build GRCh38
rs975892271 Chr.3: 10925875-10925875 on Build GRCh38
rs941146210 Chr.3: 10925944-10925944 on Build GRCh38
rs367863096 Chr.3: 10925722-10925723 on Build GRCh38
rs1030149754 Chr.3: 10925880-10925880 on Build GRCh38
rs757356033 Chr.3: 10926092-10926092 on Build GRCh38
rs1010488027 Chr.3: 10926104-10926104 on Build GRCh38
rs959612632 Chr.3: 10926172-10926172 on Build GRCh38
rs958421621 Chr.3: 10925737-10925737 on Build GRCh38
rs545968317 Chr.3: 10925765-10925765 on Build GRCh38
rs761493433 Chr.3: 10926049-10926049 on Build GRCh38
rs918756308 Chr.3: 10926021-10926021 on Build GRCh38
rs749961937 Chr.3: 10925984-10925984 on Build GRCh38
rs901199318 Chr.3: 10926061-10926061 on Build GRCh38
rs147839581 Chr.3: 10926038-10926038 on Build GRCh38
rs1035032946 Chr.3: 10926153-10926153 on Build GRCh38
rs200922963 Chr.3: 10925996-10925996 on Build GRCh38
rs959328459 Chr.3: 10925886-10925886 on Build GRCh38
rs1021022639 Chr.3: 10925787-10925787 on Build GRCh38
rs2272395 Chr.3: 10926053-10926053 on Build GRCh38
rs988568256 Chr.3: 10925925-10925925 on Build GRCh38
rs149901093 Chr.3: 10926008-10926008 on Build GRCh38
rs368046949 Chr.3: 10926033-10926033 on Build GRCh38
rs752519185 Chr.3: 10926077-10926077 on Build GRCh38
rs1057345344 Chr.3: 10926014-10926014 on Build GRCh38
rs968885013 Chr.3: 10926128-10926128 on Build GRCh38
rs1036853911 Chr.3: 10925949-10925949 on Build GRCh38
rs1030285454 Chr.3: 10926073-10926073 on Build GRCh38
rs956094030 Chr.3: 10925921-10925921 on Build GRCh38
rs374204629 Chr.3: 10926070-10926070 on Build GRCh38
rs754098162 Chr.3: 10926089-10926089 on Build GRCh38
rs767812961 Chr.3: 10925965-10925965 on Build GRCh38
rs112500885 Chr.3: 10925806-10925806 on Build GRCh38
rs774891377 Chr.3: 10926025-10926025 on Build GRCh38
rs1017501311 Chr.3: 10925841-10925841 on Build GRCh38
rs955895033 Chr.3: 10926102-10926102 on Build GRCh38
rs770963281 Chr.3: 10926009-10926009 on Build GRCh38
rs933718591 Chr.3: 10925975-10925975 on Build GRCh38
rs756280414 Chr.3: 10926078-10926078 on Build GRCh38
rs764783549 Chr.3: 10925977-10925977 on Build GRCh38
rs764683366 Chr.3: 10925983-10925984 on Build GRCh38
rs564634236 Chr.3: 10925774-10925774 on Build GRCh38
rs778920734 Chr.3: 10926145-10926145 on Build GRCh38
rs765869429 Chr.3: 10926065-10926065 on Build GRCh38
rs572441598 Chr.3: 10925754-10925754 on Build GRCh38
rs769536123 Chr.3: 10925811-10925811 on Build GRCh38
rs531379252 Chr.3: 10925851-10925851 on Build GRCh38
rs775305586 Chr.3: 10925910-10925910 on Build GRCh38
rs897353882 Chr.3: 10925739-10925739 on Build GRCh38
rs1021339841 Chr.3: 10926127-10926127 on Build GRCh38
rs1057241724 Chr.3: 10925719-10925720 on Build GRCh38
rs370801289 Chr.3: 10926007-10926007 on Build GRCh38
rs758373668 Chr.3: 10925986-10925986 on Build GRCh38
rs2272394 Chr.3: 10926026-10926026 on Build GRCh38
rs746328420 Chr.3: 10926149-10926149 on Build GRCh38
rs748307591 Chr.3: 10925997-10925997 on Build GRCh38
rs888451906 Chr.3: 10925807-10925807 on Build GRCh38
rs991105904 Chr.3: 10925908-10925908 on Build GRCh38
rs776421038 Chr.3: 10925968-10925968 on Build GRCh38
rs756333513 Chr.3: 10926000-10926000 on Build GRCh38
rs982118691 Chr.3: 10925826-10925826 on Build GRCh38
rs561924824 Chr.3: 10925909-10925909 on Build GRCh38
rs754795156 Chr.3: 10925994-10925994 on Build GRCh38
rs888538842 Chr.3: 10926037-10926037 on Build GRCh38
rs767600599 Chr.3: 10925992-10925992 on Build GRCh38

View Full Product Details