Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 15073420-15073921 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBSN
Entrez Gene ID
64145
Gene Name
rabenosyn, RAB effector
Gene Aliases
Rabenosyn-5, ZFYVE20
UniGene
Hs.475565
Gene Chromosome Location
Chr.3: 15070069-15099163 on Build GRCh38
Transcripts
NM_001302378.1 NM_022340.3 XM_005265384.4 XM_005265385.4 XM_011534001.2 XM_017007023.1 XM_017007024.1 XM_017007025.1 XM_017007026.1 XM_017007027.1 XM_017007028.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6835949 c.2350G>A p.D784N Chr.3: 15073787-15073787 on Build GRCh38
4912314 c.2301G>T p.E767D Chr.3: 15073836-15073836 on Build GRCh38
5863761 c.2342G>A p.G781E Chr.3: 15073795-15073795 on Build GRCh38
5398405 c.2289G>A p.E763E Chr.3: 15073848-15073848 on Build GRCh38
36593 c.2229C>T p.I743I Chr.3: 15073908-15073908 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756564515 Chr.3: 15073626-15073626 on Build GRCh38
rs1028374591 Chr.3: 15073659-15073659 on Build GRCh38
rs751937571 Chr.3: 15073455-15073455 on Build GRCh38
rs761049645 Chr.3: 15073813-15073813 on Build GRCh38
rs61740931 Chr.3: 15073869-15073869 on Build GRCh38
rs945387901 Chr.3: 15073462-15073462 on Build GRCh38
rs149898306 Chr.3: 15073853-15073853 on Build GRCh38
rs777022072 Chr.3: 15073896-15073896 on Build GRCh38
rs760562801 Chr.3: 15073878-15073878 on Build GRCh38
rs911771832 Chr.3: 15073448-15073448 on Build GRCh38
rs767948182 Chr.3: 15073480-15073480 on Build GRCh38
rs946479658 Chr.3: 15073502-15073502 on Build GRCh38
rs370531432 Chr.3: 15073581-15073581 on Build GRCh38
rs1044794681 Chr.3: 15073510-15073510 on Build GRCh38
rs751932987 Chr.3: 15073787-15073787 on Build GRCh38
rs533990013 Chr.3: 15073829-15073829 on Build GRCh38
rs146245590 Chr.3: 15073653-15073653 on Build GRCh38
rs1004410390 Chr.3: 15073632-15073632 on Build GRCh38
rs775503983 Chr.3: 15073873-15073873 on Build GRCh38
rs3831 Chr.3: 15073763-15073763 on Build GRCh38
rs770428003 Chr.3: 15073770-15073770 on Build GRCh38
rs932733368 Chr.3: 15073513-15073513 on Build GRCh38
rs747366907 Chr.3: 15073842-15073842 on Build GRCh38
rs772046823 Chr.3: 15073872-15073872 on Build GRCh38
rs754689523 Chr.3: 15073655-15073655 on Build GRCh38
rs956636791 Chr.3: 15073737-15073737 on Build GRCh38
rs967708426 Chr.3: 15073777-15073777 on Build GRCh38
rs1056556887 Chr.3: 15073533-15073533 on Build GRCh38
rs772774541 Chr.3: 15073753-15073753 on Build GRCh38
rs922613878 Chr.3: 15073796-15073796 on Build GRCh38
rs558955215 Chr.3: 15073668-15073668 on Build GRCh38
rs747846912 Chr.3: 15073741-15073741 on Build GRCh38
rs757303573 Chr.3: 15073826-15073826 on Build GRCh38
rs1013880872 Chr.3: 15073623-15073623 on Build GRCh38
rs754058789 Chr.3: 15073821-15073821 on Build GRCh38
rs189564761 Chr.3: 15073701-15073701 on Build GRCh38
rs546921280 Chr.3: 15073785-15073785 on Build GRCh38
rs1001257169 Chr.3: 15073656-15073656 on Build GRCh38
rs146141673 Chr.3: 15073789-15073789 on Build GRCh38
rs755266924 Chr.3: 15073846-15073846 on Build GRCh38
rs987839644 Chr.3: 15073466-15073466 on Build GRCh38
rs546607652 Chr.3: 15073523-15073523 on Build GRCh38
rs987340802 Chr.3: 15073670-15073670 on Build GRCh38
rs780454425 Chr.3: 15073840-15073840 on Build GRCh38
rs958216728 Chr.3: 15073667-15073667 on Build GRCh38
rs757628802 Chr.3: 15073474-15073474 on Build GRCh38
rs764417962 Chr.3: 15073818-15073818 on Build GRCh38
rs779552046 Chr.3: 15073638-15073638 on Build GRCh38
rs935231819 Chr.3: 15073461-15073461 on Build GRCh38
rs780431561 Chr.3: 15073733-15073733 on Build GRCh38
rs139267270 Chr.3: 15073868-15073868 on Build GRCh38
rs34911587 Chr.3: 15073481-15073481 on Build GRCh38
rs201461257 Chr.3: 15073522-15073522 on Build GRCh38
rs142390372 Chr.3: 15073797-15073797 on Build GRCh38
rs775415918 Chr.3: 15073811-15073811 on Build GRCh38
rs765318494 Chr.3: 15073828-15073828 on Build GRCh38
rs774189376 Chr.3: 15073772-15073772 on Build GRCh38
rs566302360 Chr.3: 15073590-15073590 on Build GRCh38
rs770388561 Chr.3: 15073865-15073865 on Build GRCh38
rs763929314 Chr.3: 15073884-15073884 on Build GRCh38
rs750968806 Chr.3: 15073579-15073579 on Build GRCh38
rs201944804 Chr.3: 15073742-15073742 on Build GRCh38
rs141948895 Chr.3: 15073864-15073864 on Build GRCh38
rs1015498791 Chr.3: 15073586-15073586 on Build GRCh38

View Full Product Details