Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 95246567-95247090 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RWDD3
Entrez Gene ID
25950
Gene Name
RWD domain containing 3
Gene Aliases
RSUME
UniGene
Hs.483512
Gene Chromosome Location
Chr.1: 95234155-95247225 on Build GRCh38
Transcripts
NM_001128142.1 NM_001199682.1 NM_001278247.1 NM_001278248.1 NM_015485.4 NR_103483.1 NR_103484.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3493601 c.611T>C p.V204A Chr.1: 95246579-95246579 on Build GRCh38
5117281 c.690-10T>C p.? Chr.1: 95246746-95246746 on Build GRCh38
3493602 c.746T>G p.F249C Chr.1: 95246812-95246812 on Build GRCh38
913549 c.609T>C p.D203D Chr.1: 95246577-95246577 on Build GRCh38
913550 c.637G>A p.E213K Chr.1: 95246605-95246605 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768357993 Chr.1: 95246775-95246775 on Build GRCh38
rs576948307 Chr.1: 95246698-95246698 on Build GRCh38
rs767512529 Chr.1: 95246907-95246929 on Build GRCh38
rs751580311 Chr.1: 95246723-95246723 on Build GRCh38
rs781109438 Chr.1: 95246845-95246845 on Build GRCh38
rs559321774 Chr.1: 95247007-95247007 on Build GRCh38
rs753653417 Chr.1: 95246704-95246704 on Build GRCh38
rs1048331615 Chr.1: 95246706-95246706 on Build GRCh38
rs111453823 Chr.1: 95246785-95246785 on Build GRCh38
rs912524146 Chr.1: 95246675-95246675 on Build GRCh38
rs762674258 Chr.1: 95246633-95246633 on Build GRCh38
rs770047964 Chr.1: 95246607-95246607 on Build GRCh38
rs760234998 Chr.1: 95246900-95246900 on Build GRCh38
rs192560785 Chr.1: 95247034-95247034 on Build GRCh38
rs753560482 Chr.1: 95246828-95246828 on Build GRCh38
rs746329139 Chr.1: 95246739-95246739 on Build GRCh38
rs761416077 Chr.1: 95246673-95246673 on Build GRCh38
rs770383198 Chr.1: 95246753-95246753 on Build GRCh38
rs777971687 Chr.1: 95246718-95246718 on Build GRCh38
rs763861733 Chr.1: 95246646-95246646 on Build GRCh38
rs926481400 Chr.1: 95247065-95247065 on Build GRCh38
rs149350415 Chr.1: 95246829-95246831 on Build GRCh38
rs780234612 Chr.1: 95246763-95246763 on Build GRCh38
rs1049105463 Chr.1: 95246797-95246797 on Build GRCh38
rs780712537 Chr.1: 95246856-95246856 on Build GRCh38
rs139153792 Chr.1: 95246906-95246907 on Build GRCh38
rs757270820 Chr.1: 95246729-95246729 on Build GRCh38
rs770713587 Chr.1: 95246901-95246901 on Build GRCh38
rs369502560 Chr.1: 95246882-95246882 on Build GRCh38
rs759337170 Chr.1: 95246840-95246840 on Build GRCh38
rs369779655 Chr.1: 95246796-95246796 on Build GRCh38
rs377309701 Chr.1: 95246879-95246879 on Build GRCh38
rs558278162 Chr.1: 95247030-95247030 on Build GRCh38
rs889953420 Chr.1: 95246899-95246899 on Build GRCh38
rs771052210 Chr.1: 95246774-95246775 on Build GRCh38
rs953653699 Chr.1: 95246622-95246622 on Build GRCh38
rs766367083 Chr.1: 95246814-95246814 on Build GRCh38
rs761893335 Chr.1: 95246911-95246911 on Build GRCh38
rs752537308 Chr.1: 95246909-95246909 on Build GRCh38
rs1028040000 Chr.1: 95247036-95247036 on Build GRCh38
rs200205495 Chr.1: 95246853-95246853 on Build GRCh38
rs773037798 Chr.1: 95247023-95247023 on Build GRCh38
rs750316695 Chr.1: 95246688-95246688 on Build GRCh38
rs758574924 Chr.1: 95246711-95246711 on Build GRCh38
rs751774407 Chr.1: 95246844-95246844 on Build GRCh38
rs770131380 Chr.1: 95246780-95246780 on Build GRCh38
rs762840754 Chr.1: 95246614-95246616 on Build GRCh38
rs749702259 Chr.1: 95246772-95246772 on Build GRCh38
rs141917580 Chr.1: 95247063-95247063 on Build GRCh38
rs771775151 Chr.1: 95246891-95246891 on Build GRCh38
rs750539788 Chr.1: 95246852-95246852 on Build GRCh38
rs956269315 Chr.1: 95246958-95246958 on Build GRCh38
rs749609666 Chr.1: 95246863-95246863 on Build GRCh38
rs592097 Chr.1: 95246902-95246902 on Build GRCh38
rs769132189 Chr.1: 95246875-95246875 on Build GRCh38
rs778597049 Chr.1: 95246834-95246834 on Build GRCh38
rs1021551569 Chr.1: 95247004-95247004 on Build GRCh38
rs760477235 Chr.1: 95246809-95246809 on Build GRCh38
rs1028008799 Chr.1: 95246592-95246592 on Build GRCh38
rs371709891 Chr.1: 95246674-95246674 on Build GRCh38
rs773959999 Chr.1: 95246649-95246649 on Build GRCh38
rs781397779 Chr.1: 95246736-95246736 on Build GRCh38
rs368987417 Chr.1: 95246788-95246788 on Build GRCh38
rs767812256 Chr.1: 95246915-95246915 on Build GRCh38
rs775964778 Chr.1: 95246623-95246623 on Build GRCh38
rs756146593 Chr.1: 95246693-95246693 on Build GRCh38
rs766503471 Chr.1: 95246966-95246966 on Build GRCh38
rs374946719 Chr.1: 95246789-95246789 on Build GRCh38
rs374426010 Chr.1: 95246843-95246843 on Build GRCh38
rs761618068 Chr.1: 95246787-95246787 on Build GRCh38
rs746444889 Chr.1: 95246594-95246594 on Build GRCh38
rs764986286 Chr.1: 95246904-95246904 on Build GRCh38
rs772552523 Chr.1: 95246898-95246898 on Build GRCh38

View Full Product Details