Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 95246968-95247468 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RWDD3
Entrez Gene ID
25950
Gene Name
RWD domain containing 3
Gene Aliases
RSUME
UniGene
Hs.483512
Gene Chromosome Location
Chr.1: 95234155-95247225 on Build GRCh38
Transcripts
NM_001128142.1 NM_001199682.1 NM_001278247.1 NM_001278248.1 NM_015485.4 NR_103483.1 NR_103484.1

SNP Detail

SNP ID Chromosome Location
rs1019279674 Chr.1: 95247140-95247140 on Build GRCh38
rs889570189 Chr.1: 95247349-95247349 on Build GRCh38
rs904025753 Chr.1: 95247318-95247318 on Build GRCh38
rs115648243 Chr.1: 95247396-95247396 on Build GRCh38
rs1028040000 Chr.1: 95247036-95247036 on Build GRCh38
rs773037798 Chr.1: 95247023-95247023 on Build GRCh38
rs867995055 Chr.1: 95247081-95247081 on Build GRCh38
rs76786702 Chr.1: 95247199-95247199 on Build GRCh38
rs559321774 Chr.1: 95247007-95247007 on Build GRCh38
rs959357295 Chr.1: 95247428-95247428 on Build GRCh38
rs961606446 Chr.1: 95247157-95247157 on Build GRCh38
rs141917580 Chr.1: 95247063-95247063 on Build GRCh38
rs868357943 Chr.1: 95247354-95247354 on Build GRCh38
rs1010404228 Chr.1: 95247319-95247319 on Build GRCh38
rs1028478505 Chr.1: 95247347-95247347 on Build GRCh38
rs560493628 Chr.1: 95247147-95247147 on Build GRCh38
rs562015803 Chr.1: 95247275-95247275 on Build GRCh38
rs192560785 Chr.1: 95247034-95247034 on Build GRCh38
rs938077994 Chr.1: 95247227-95247227 on Build GRCh38
rs972733761 Chr.1: 95247217-95247217 on Build GRCh38
rs3767312 Chr.1: 95247331-95247331 on Build GRCh38
rs972652849 Chr.1: 95247161-95247161 on Build GRCh38
rs3767313 Chr.1: 95247241-95247241 on Build GRCh38
rs952735528 Chr.1: 95247435-95247435 on Build GRCh38
rs1021551569 Chr.1: 95247004-95247004 on Build GRCh38
rs574358345 Chr.1: 95247191-95247191 on Build GRCh38
rs959220471 Chr.1: 95247116-95247116 on Build GRCh38
rs369495593 Chr.1: 95247300-95247300 on Build GRCh38
rs926481400 Chr.1: 95247065-95247065 on Build GRCh38
rs1007953745 Chr.1: 95247378-95247378 on Build GRCh38
rs945635087 Chr.1: 95247279-95247279 on Build GRCh38
rs1026938288 Chr.1: 95247424-95247424 on Build GRCh38
rs994499934 Chr.1: 95247420-95247420 on Build GRCh38
rs922627668 Chr.1: 95247218-95247218 on Build GRCh38
rs573085524 Chr.1: 95247168-95247168 on Build GRCh38
rs1019310849 Chr.1: 95247387-95247387 on Build GRCh38
rs919756908 Chr.1: 95247164-95247164 on Build GRCh38
rs926747127 Chr.1: 95247224-95247224 on Build GRCh38
rs889975164 Chr.1: 95247274-95247274 on Build GRCh38
rs906191364 Chr.1: 95247366-95247366 on Build GRCh38
rs558278162 Chr.1: 95247030-95247030 on Build GRCh38
rs531974562 Chr.1: 95247384-95247384 on Build GRCh38
rs986489731 Chr.1: 95247082-95247082 on Build GRCh38

View Full Product Details