Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 24118590-24119086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
THRB
Entrez Gene ID
7068
Gene Name
thyroid hormone receptor beta
Gene Aliases
C-ERBA-2, C-ERBA-BETA, ERBA2, GRTH, NR1A2, PRTH, THR1, THRB1, THRB2
UniGene
Hs.187861
Gene Chromosome Location
Chr.3: 24117153-24495281 on Build GRCh38
Transcripts
NM_000461.4 NM_001128176.2 NM_001128177.1 NM_001252634.1 XM_006713317.3

SNP Detail

SNP ID Chromosome Location
rs751368376 Chr.3: 24118903-24118903 on Build GRCh38
rs1020769364 Chr.3: 24118939-24118939 on Build GRCh38
rs886058279 Chr.3: 24119005-24119005 on Build GRCh38
rs538000688 Chr.3: 24118835-24118835 on Build GRCh38
rs765099000 Chr.3: 24118743-24118743 on Build GRCh38
rs954585458 Chr.3: 24118688-24118688 on Build GRCh38
rs1017568894 Chr.3: 24118960-24118960 on Build GRCh38
rs1043029913 Chr.3: 24118925-24118925 on Build GRCh38
rs982786930 Chr.3: 24118970-24118970 on Build GRCh38
rs890298316 Chr.3: 24118948-24118948 on Build GRCh38
rs142926795 Chr.3: 24118755-24118755 on Build GRCh38
rs973171220 Chr.3: 24118730-24118730 on Build GRCh38
rs548921995 Chr.3: 24118847-24118847 on Build GRCh38
rs1008709491 Chr.3: 24118992-24118992 on Build GRCh38
rs1047861923 Chr.3: 24118757-24118757 on Build GRCh38
rs1027342820 Chr.3: 24118973-24118973 on Build GRCh38
rs569455707 Chr.3: 24118860-24118860 on Build GRCh38
rs71622766 Chr.3: 24118961-24118962 on Build GRCh38
rs13320551 Chr.3: 24118938-24118938 on Build GRCh38
rs954811267 Chr.3: 24118645-24118645 on Build GRCh38
rs886058273 Chr.3: 24118969-24118969 on Build GRCh38
rs772201945 Chr.3: 24118633-24118633 on Build GRCh38
rs886058274 Chr.3: 24118973-24118973 on Build GRCh38
rs886058276 Chr.3: 24118974-24118974 on Build GRCh38
rs575539914 Chr.3: 24118829-24118829 on Build GRCh38
rs983172916 Chr.3: 24118981-24118981 on Build GRCh38
rs961920176 Chr.3: 24119018-24119018 on Build GRCh38
rs964397630 Chr.3: 24118961-24118961 on Build GRCh38
rs973502816 Chr.3: 24118968-24118968 on Build GRCh38
rs868646865 Chr.3: 24118787-24118787 on Build GRCh38
rs980314103 Chr.3: 24119055-24119055 on Build GRCh38
rs973023047 Chr.3: 24118711-24118711 on Build GRCh38
rs537922567 Chr.3: 24118859-24118859 on Build GRCh38
rs905859487 Chr.3: 24118828-24118828 on Build GRCh38
rs866468264 Chr.3: 24118788-24118788 on Build GRCh38
rs575525867 Chr.3: 24118695-24118695 on Build GRCh38
rs754256615 Chr.3: 24118974-24118980 on Build GRCh38
rs749860593 Chr.3: 24118973-24118974 on Build GRCh38
rs917428792 Chr.3: 24118774-24118774 on Build GRCh38
rs890306058 Chr.3: 24118966-24118966 on Build GRCh38
rs760745743 Chr.3: 24118710-24118710 on Build GRCh38
rs549973481 Chr.3: 24118955-24118955 on Build GRCh38
rs151088155 Chr.3: 24118851-24118851 on Build GRCh38
rs1020862895 Chr.3: 24118685-24118685 on Build GRCh38
rs980720098 Chr.3: 24118768-24118768 on Build GRCh38
rs376819021 Chr.3: 24118679-24118679 on Build GRCh38
rs1014819036 Chr.3: 24118691-24118691 on Build GRCh38
rs917376444 Chr.3: 24119037-24119037 on Build GRCh38
rs886058275 Chr.3: 24118974-24118978 on Build GRCh38
rs760776123 Chr.3: 24118902-24118902 on Build GRCh38
rs964774423 Chr.3: 24118725-24118725 on Build GRCh38
rs886058278 Chr.3: 24119004-24119004 on Build GRCh38
rs559325556 Chr.3: 24118974-24118979 on Build GRCh38
rs950491355 Chr.3: 24118978-24118978 on Build GRCh38
rs1014850269 Chr.3: 24119004-24119004 on Build GRCh38
rs369341188 Chr.3: 24119054-24119054 on Build GRCh38
rs578023648 Chr.3: 24118897-24118897 on Build GRCh38
rs957887505 Chr.3: 24118629-24118629 on Build GRCh38
rs1010336847 Chr.3: 24118905-24118905 on Build GRCh38
rs886058277 Chr.3: 24118999-24119004 on Build GRCh38
rs938966989 Chr.3: 24118832-24118832 on Build GRCh38
rs774543354 Chr.3: 24118980-24118981 on Build GRCh38
rs1044630857 Chr.3: 24118810-24118810 on Build GRCh38
rs996256712 Chr.3: 24118955-24118956 on Build GRCh38

View Full Product Details