Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 39183133-39183644 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
XIRP1
Entrez Gene ID
165904
Gene Name
xin actin binding repeat containing 1
Gene Aliases
CMYA1, Xin
UniGene
Hs.447868
Gene Chromosome Location
Chr.3: 39183214-39192621 on Build GRCh38
Transcripts
NM_001198621.2 NM_194293.2 XM_005264909.3

SNP Detail

SNP ID Chromosome Location
rs1038302296 Chr.3: 39183576-39183576 on Build GRCh38
rs938820543 Chr.3: 39183477-39183477 on Build GRCh38
rs1003012107 Chr.3: 39183245-39183245 on Build GRCh38
rs965184901 Chr.3: 39183323-39183323 on Build GRCh38
rs375913299 Chr.3: 39183361-39183361 on Build GRCh38
rs527470928 Chr.3: 39183232-39183232 on Build GRCh38
rs966930880 Chr.3: 39183386-39183386 on Build GRCh38
rs533190487 Chr.3: 39183327-39183327 on Build GRCh38
rs776285181 Chr.3: 39183482-39183482 on Build GRCh38
rs1034526957 Chr.3: 39183253-39183253 on Build GRCh38
rs915646375 Chr.3: 39183470-39183470 on Build GRCh38
rs770520735 Chr.3: 39183452-39183452 on Build GRCh38
rs547917730 Chr.3: 39183294-39183294 on Build GRCh38
rs1012267058 Chr.3: 39183580-39183580 on Build GRCh38
rs562654418 Chr.3: 39183170-39183170 on Build GRCh38
rs894730349 Chr.3: 39183260-39183260 on Build GRCh38
rs906292620 Chr.3: 39183451-39183451 on Build GRCh38
rs1008169170 Chr.3: 39183352-39183352 on Build GRCh38
rs190526205 Chr.3: 39183350-39183350 on Build GRCh38
rs181207329 Chr.3: 39183158-39183158 on Build GRCh38
rs932521176 Chr.3: 39183582-39183582 on Build GRCh38
rs112894855 Chr.3: 39183299-39183299 on Build GRCh38
rs553472576 Chr.3: 39183545-39183545 on Build GRCh38
rs997306102 Chr.3: 39183303-39183303 on Build GRCh38
rs891202149 Chr.3: 39183594-39183594 on Build GRCh38
rs1032888492 Chr.3: 39183413-39183413 on Build GRCh38
rs545563374 Chr.3: 39183581-39183581 on Build GRCh38
rs539909254 Chr.3: 39183484-39183484 on Build GRCh38
rs546875926 Chr.3: 39183262-39183262 on Build GRCh38
rs1004168784 Chr.3: 39183266-39183266 on Build GRCh38
rs1028422024 Chr.3: 39183304-39183304 on Build GRCh38
rs908231265 Chr.3: 39183357-39183357 on Build GRCh38
rs887013468 Chr.3: 39183205-39183205 on Build GRCh38
rs754617536 Chr.3: 39183499-39183499 on Build GRCh38
rs908249924 Chr.3: 39183498-39183498 on Build GRCh38
rs138868130 Chr.3: 39183462-39183462 on Build GRCh38
rs535793322 Chr.3: 39183405-39183405 on Build GRCh38
rs114470485 Chr.3: 39183584-39183584 on Build GRCh38
rs1019671159 Chr.3: 39183614-39183614 on Build GRCh38
rs915532186 Chr.3: 39183156-39183156 on Build GRCh38
rs542121726 Chr.3: 39183192-39183192 on Build GRCh38
rs555631375 Chr.3: 39183410-39183410 on Build GRCh38
rs878876522 Chr.3: 39183557-39183557 on Build GRCh38
rs1036474243 Chr.3: 39183155-39183155 on Build GRCh38
rs531664862 Chr.3: 39183191-39183191 on Build GRCh38
rs939880068 Chr.3: 39183530-39183530 on Build GRCh38
rs949869780 Chr.3: 39183480-39183480 on Build GRCh38
rs894726290 Chr.3: 39183521-39183521 on Build GRCh38
rs1022086069 Chr.3: 39183272-39183272 on Build GRCh38
rs1045450498 Chr.3: 39183173-39183173 on Build GRCh38
rs576836412 Chr.3: 39183547-39183547 on Build GRCh38
rs534842303 Chr.3: 39183515-39183515 on Build GRCh38
rs185812431 Chr.3: 39183161-39183161 on Build GRCh38
rs975251181 Chr.3: 39183324-39183324 on Build GRCh38
rs561908323 Chr.3: 39183219-39183219 on Build GRCh38
rs557666371 Chr.3: 39183605-39183605 on Build GRCh38
rs1056403043 Chr.3: 39183503-39183503 on Build GRCh38
rs921184410 Chr.3: 39183334-39183334 on Build GRCh38

View Full Product Details