Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 46577812-46578351 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC2
Entrez Gene ID
79442
Gene Name
leucine rich repeat containing 2
Gene Aliases
-
UniGene
Hs.657345
Gene Chromosome Location
Chr.3: 46515385-46580099 on Build GRCh38
Transcripts
XM_011534110.2 NM_001174136.1 NM_003212.3

Gene Symbol
TDGF1
Entrez Gene ID
6997
Gene Name
teratocarcinoma-derived growth factor 1
Gene Aliases
CR, CRGF, CRIPTO
UniGene
Hs.385870
Gene Chromosome Location
Chr.3: 46574555-46582463 on Build GRCh38
Transcripts
XM_011534110.2 NM_001174136.1 NM_003212.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4975639 c.22C>T p.R8C Chr.3: 46577991-46577991 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs751574884 Chr.3: 46577991-46577991 on Build GRCh38
rs948845771 Chr.3: 46578186-46578186 on Build GRCh38
rs190338101 Chr.3: 46578126-46578126 on Build GRCh38
rs1046639840 Chr.3: 46578258-46578261 on Build GRCh38
rs762558362 Chr.3: 46577979-46577979 on Build GRCh38
rs188178709 Chr.3: 46578165-46578165 on Build GRCh38
rs892768147 Chr.3: 46578299-46578299 on Build GRCh38
rs113542669 Chr.3: 46578042-46578042 on Build GRCh38
rs372738951 Chr.3: 46577953-46577953 on Build GRCh38
rs986632810 Chr.3: 46578117-46578117 on Build GRCh38
rs1009259238 Chr.3: 46577841-46577841 on Build GRCh38
rs1040770690 Chr.3: 46578310-46578310 on Build GRCh38
rs936535214 Chr.3: 46578263-46578263 on Build GRCh38
rs551688560 Chr.3: 46578203-46578203 on Build GRCh38
rs369690027 Chr.3: 46578037-46578037 on Build GRCh38
rs754664545 Chr.3: 46577950-46577950 on Build GRCh38
rs780797973 Chr.3: 46577955-46577955 on Build GRCh38
rs375634868 Chr.3: 46577992-46577992 on Build GRCh38
rs868683577 Chr.3: 46577960-46577960 on Build GRCh38
rs561483662 Chr.3: 46578167-46578167 on Build GRCh38
rs554290986 Chr.3: 46577899-46577899 on Build GRCh38
rs748033020 Chr.3: 46577980-46577980 on Build GRCh38
rs183597975 Chr.3: 46578164-46578164 on Build GRCh38
rs192773746 Chr.3: 46578267-46578267 on Build GRCh38
rs562100641 Chr.3: 46578114-46578114 on Build GRCh38
rs745887888 Chr.3: 46578012-46578012 on Build GRCh38
rs757428100 Chr.3: 46578005-46578005 on Build GRCh38
rs56398705 Chr.3: 46578125-46578125 on Build GRCh38
rs1055228392 Chr.3: 46577975-46577975 on Build GRCh38
rs572946325 Chr.3: 46577912-46577912 on Build GRCh38
rs766362536 Chr.3: 46577988-46577988 on Build GRCh38
rs769129255 Chr.3: 46577951-46577951 on Build GRCh38
rs776116526 Chr.3: 46577952-46577952 on Build GRCh38
rs769549981 Chr.3: 46578240-46578240 on Build GRCh38
rs892411146 Chr.3: 46578268-46578268 on Build GRCh38
rs772808577 Chr.3: 46577971-46577971 on Build GRCh38
rs753989564 Chr.3: 46577998-46577998 on Build GRCh38
rs55955974 Chr.3: 46578064-46578064 on Build GRCh38
rs953981733 Chr.3: 46578086-46578086 on Build GRCh38
rs912477561 Chr.3: 46578121-46578121 on Build GRCh38
rs532002514 Chr.3: 46578180-46578180 on Build GRCh38
rs758528837 Chr.3: 46578018-46578018 on Build GRCh38
rs1025197681 Chr.3: 46577997-46577997 on Build GRCh38
rs1011251936 Chr.3: 46578316-46578316 on Build GRCh38
rs765004486 Chr.3: 46577955-46577955 on Build GRCh38
rs779108382 Chr.3: 46578008-46578008 on Build GRCh38
rs550498849 Chr.3: 46578184-46578184 on Build GRCh38
rs146804780 Chr.3: 46578073-46578073 on Build GRCh38
rs56197743 Chr.3: 46578283-46578283 on Build GRCh38
rs904576628 Chr.3: 46578235-46578235 on Build GRCh38
rs867814858 Chr.3: 46577875-46577875 on Build GRCh38
rs747589553 Chr.3: 46578039-46578039 on Build GRCh38
rs975156240 Chr.3: 46577989-46577989 on Build GRCh38
rs777004285 Chr.3: 46578056-46578056 on Build GRCh38
rs534028784 Chr.3: 46577917-46577917 on Build GRCh38
rs1044489782 Chr.3: 46578199-46578199 on Build GRCh38
rs569271425 Chr.3: 46578252-46578252 on Build GRCh38
rs898568073 Chr.3: 46577869-46577869 on Build GRCh38
rs754428178 Chr.3: 46577858-46577858 on Build GRCh38
rs560232746 Chr.3: 46578141-46578141 on Build GRCh38

View Full Product Details