Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 46581003-46581501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TDGF1
Entrez Gene ID
6997
Gene Name
teratocarcinoma-derived growth factor 1
Gene Aliases
CR, CRGF, CRIPTO
UniGene
Hs.385870
Gene Chromosome Location
Chr.3: 46574555-46582463 on Build GRCh38
Transcripts
NM_001174136.1 NM_003212.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7150594 c.506C>G p.S169C Chr.3: 46581189-46581189 on Build GRCh38
4497429 c.496C>T p.L166L Chr.3: 46581179-46581179 on Build GRCh38
5807262 c.470A>G p.H157R Chr.3: 46581153-46581153 on Build GRCh38
7265817 c.503C>A p.P168Q Chr.3: 46581186-46581186 on Build GRCh38
5864604 c.500C>A p.P167Q Chr.3: 46581183-46581183 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145651481 Chr.3: 46581130-46581130 on Build GRCh38
rs149148398 Chr.3: 46581458-46581458 on Build GRCh38
rs902856649 Chr.3: 46581036-46581036 on Build GRCh38
rs377424657 Chr.3: 46581187-46581187 on Build GRCh38
rs140599197 Chr.3: 46581277-46581277 on Build GRCh38
rs536686908 Chr.3: 46581049-46581049 on Build GRCh38
rs200034661 Chr.3: 46581093-46581093 on Build GRCh38
rs368974653 Chr.3: 46581254-46581254 on Build GRCh38
rs919476319 Chr.3: 46581116-46581116 on Build GRCh38
rs199680596 Chr.3: 46581112-46581112 on Build GRCh38
rs143388946 Chr.3: 46581195-46581195 on Build GRCh38
rs529958805 Chr.3: 46581413-46581413 on Build GRCh38
rs954593522 Chr.3: 46581050-46581050 on Build GRCh38
rs144324787 Chr.3: 46581372-46581372 on Build GRCh38
rs903586194 Chr.3: 46581349-46581349 on Build GRCh38
rs370270206 Chr.3: 46581239-46581239 on Build GRCh38
rs780889746 Chr.3: 46581091-46581092 on Build GRCh38
rs945032518 Chr.3: 46581283-46581283 on Build GRCh38
rs757319888 Chr.3: 46581138-46581138 on Build GRCh38
rs752295471 Chr.3: 46581231-46581232 on Build GRCh38
rs375142949 Chr.3: 46581185-46581185 on Build GRCh38
rs750477388 Chr.3: 46581157-46581157 on Build GRCh38
rs764704416 Chr.3: 46581334-46581334 on Build GRCh38
rs896114194 Chr.3: 46581085-46581085 on Build GRCh38
rs144592797 Chr.3: 46581174-46581174 on Build GRCh38
rs750468615 Chr.3: 46581260-46581260 on Build GRCh38
rs1009243412 Chr.3: 46581086-46581086 on Build GRCh38
rs112928637 Chr.3: 46581139-46581139 on Build GRCh38
rs998491086 Chr.3: 46581038-46581038 on Build GRCh38
rs140356933 Chr.3: 46581200-46581200 on Build GRCh38
rs540742671 Chr.3: 46581302-46581302 on Build GRCh38
rs146545269 Chr.3: 46581456-46581456 on Build GRCh38
rs181142338 Chr.3: 46581136-46581136 on Build GRCh38
rs1017312916 Chr.3: 46581384-46581384 on Build GRCh38
rs534737011 Chr.3: 46581111-46581111 on Build GRCh38
rs17852000 Chr.3: 46581193-46581193 on Build GRCh38
rs202071894 Chr.3: 46581252-46581252 on Build GRCh38
rs950687508 Chr.3: 46581457-46581457 on Build GRCh38
rs754469951 Chr.3: 46581085-46581086 on Build GRCh38
rs1017418979 Chr.3: 46581103-46581103 on Build GRCh38
rs370748283 Chr.3: 46581120-46581120 on Build GRCh38
rs150756875 Chr.3: 46581229-46581229 on Build GRCh38
rs372064575 Chr.3: 46581410-46581410 on Build GRCh38
rs773009525 Chr.3: 46581096-46581096 on Build GRCh38
rs909270528 Chr.3: 46581168-46581168 on Build GRCh38
rs963701291 Chr.3: 46581106-46581106 on Build GRCh38
rs761438345 Chr.3: 46581298-46581298 on Build GRCh38
rs879061016 Chr.3: 46581227-46581227 on Build GRCh38
rs552231663 Chr.3: 46581059-46581059 on Build GRCh38
rs372278806 Chr.3: 46581273-46581273 on Build GRCh38
rs62248660 Chr.3: 46581253-46581253 on Build GRCh38
rs747379324 Chr.3: 46581182-46581182 on Build GRCh38
rs1042076154 Chr.3: 46581316-46581316 on Build GRCh38
rs776472360 Chr.3: 46581087-46581087 on Build GRCh38
rs776213445 Chr.3: 46581186-46581186 on Build GRCh38
rs758458339 Chr.3: 46581263-46581263 on Build GRCh38
rs762888502 Chr.3: 46581098-46581098 on Build GRCh38
rs997341229 Chr.3: 46581418-46581418 on Build GRCh38
rs112343101 Chr.3: 46581280-46581280 on Build GRCh38
rs138586687 Chr.3: 46581158-46581158 on Build GRCh38
rs1016238695 Chr.3: 46581459-46581459 on Build GRCh38
rs759292430 Chr.3: 46581246-46581246 on Build GRCh38
rs1000598874 Chr.3: 46581382-46581382 on Build GRCh38
rs765477657 Chr.3: 46581257-46581257 on Build GRCh38
rs759269251 Chr.3: 46581121-46581121 on Build GRCh38
rs751117885 Chr.3: 46581084-46581085 on Build GRCh38
rs557273125 Chr.3: 46581194-46581194 on Build GRCh38
rs770850994 Chr.3: 46581226-46581226 on Build GRCh38
rs762398499 Chr.3: 46581222-46581222 on Build GRCh38
rs201293652 Chr.3: 46581250-46581250 on Build GRCh38
rs548492558 Chr.3: 46581432-46581432 on Build GRCh38

View Full Product Details