Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49001359-49001871 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
P4HTM
Entrez Gene ID
54681
Gene Name
prolyl 4-hydroxylase, transmembrane
Gene Aliases
EGLN4, HIFPH4, P4H-TM, PH-4, PH4, PHD4
UniGene
Hs.654944
Gene Chromosome Location
Chr.3: 48989908-49007149 on Build GRCh38
Transcripts
NM_177938.2 NM_177939.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4118348 c.508C>T p.R170C Chr.3: 49001509-49001509 on Build GRCh38
6863671 c.474C>T p.L158L Chr.3: 49001475-49001475 on Build GRCh38
3595143 c.498G>A p.K166K Chr.3: 49001499-49001499 on Build GRCh38
3595144 c.498G>A p.K166K Chr.3: 49001499-49001499 on Build GRCh38
5602008 c.622C>T p.R208C Chr.3: 49001623-49001623 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149345145 Chr.3: 49001568-49001568 on Build GRCh38
rs970317404 Chr.3: 49001709-49001709 on Build GRCh38
rs753161604 Chr.3: 49001546-49001546 on Build GRCh38
rs368504809 Chr.3: 49001445-49001445 on Build GRCh38
rs913690470 Chr.3: 49001761-49001761 on Build GRCh38
rs924950551 Chr.3: 49001719-49001719 on Build GRCh38
rs768160824 Chr.3: 49001509-49001509 on Build GRCh38
rs879239497 Chr.3: 49001699-49001699 on Build GRCh38
rs72934780 Chr.3: 49001677-49001677 on Build GRCh38
rs766174732 Chr.3: 49001638-49001638 on Build GRCh38
rs771797015 Chr.3: 49001489-49001489 on Build GRCh38
rs903835305 Chr.3: 49001751-49001751 on Build GRCh38
rs143747733 Chr.3: 49001490-49001490 on Build GRCh38
rs760444979 Chr.3: 49001502-49001502 on Build GRCh38
rs756516236 Chr.3: 49001550-49001550 on Build GRCh38
rs772731122 Chr.3: 49001623-49001623 on Build GRCh38
rs748444950 Chr.3: 49001584-49001584 on Build GRCh38
rs774451170 Chr.3: 49001471-49001471 on Build GRCh38
rs140407460 Chr.3: 49001594-49001594 on Build GRCh38
rs938105536 Chr.3: 49001423-49001423 on Build GRCh38
rs764380547 Chr.3: 49001448-49001448 on Build GRCh38
rs750045495 Chr.3: 49001544-49001544 on Build GRCh38
rs565035006 Chr.3: 49001439-49001439 on Build GRCh38
rs755308276 Chr.3: 49001430-49001430 on Build GRCh38
rs936432936 Chr.3: 49001524-49001524 on Build GRCh38
rs756134121 Chr.3: 49001733-49001733 on Build GRCh38
rs1056579759 Chr.3: 49001428-49001428 on Build GRCh38
rs148666784 Chr.3: 49001589-49001589 on Build GRCh38
rs758446091 Chr.3: 49001603-49001603 on Build GRCh38
rs139608844 Chr.3: 49001572-49001572 on Build GRCh38
rs752636462 Chr.3: 49001695-49001695 on Build GRCh38
rs772383492 Chr.3: 49001576-49001576 on Build GRCh38
rs756764915 Chr.3: 49001414-49001414 on Build GRCh38
rs567658392 Chr.3: 49001735-49001735 on Build GRCh38
rs979009736 Chr.3: 49001714-49001714 on Build GRCh38
rs764687838 Chr.3: 49001622-49001622 on Build GRCh38
rs776566750 Chr.3: 49001602-49001602 on Build GRCh38
rs201760384 Chr.3: 49001393-49001393 on Build GRCh38
rs111283754 Chr.3: 49001645-49001645 on Build GRCh38
rs146471142 Chr.3: 49001585-49001585 on Build GRCh38
rs747511805 Chr.3: 49001420-49001421 on Build GRCh38
rs200947739 Chr.3: 49001552-49001552 on Build GRCh38
rs776980169 Chr.3: 49001535-49001535 on Build GRCh38
rs774099043 Chr.3: 49001510-49001510 on Build GRCh38
rs1023121989 Chr.3: 49001704-49001704 on Build GRCh38
rs964937615 Chr.3: 49001573-49001573 on Build GRCh38
rs571477998 Chr.3: 49001840-49001840 on Build GRCh38
rs761982398 Chr.3: 49001545-49001545 on Build GRCh38
rs938551156 Chr.3: 49001693-49001693 on Build GRCh38
rs750073079 Chr.3: 49001426-49001426 on Build GRCh38
rs538826627 Chr.3: 49001845-49001845 on Build GRCh38
rs185898197 Chr.3: 49001802-49001802 on Build GRCh38
rs746037527 Chr.3: 49001485-49001485 on Build GRCh38
rs764744092 Chr.3: 49001542-49001542 on Build GRCh38
rs767487328 Chr.3: 49001656-49001656 on Build GRCh38
rs993861770 Chr.3: 49001735-49001736 on Build GRCh38
rs924950217 Chr.3: 49001516-49001516 on Build GRCh38
rs988211934 Chr.3: 49001747-49001747 on Build GRCh38
rs531790010 Chr.3: 49001771-49001771 on Build GRCh38
rs1017880780 Chr.3: 49001826-49001826 on Build GRCh38
rs771030949 Chr.3: 49001470-49001470 on Build GRCh38
rs756560920 Chr.3: 49001655-49001655 on Build GRCh38
rs778402583 Chr.3: 49001446-49001446 on Build GRCh38
rs753398663 Chr.3: 49001394-49001394 on Build GRCh38
rs749445943 Chr.3: 49001467-49001467 on Build GRCh38
rs762349081 Chr.3: 49001629-49001629 on Build GRCh38
rs149397788 Chr.3: 49001420-49001420 on Build GRCh38
rs372281689 Chr.3: 49001557-49001557 on Build GRCh38

View Full Product Details