Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 51994964-51995467 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPL29
Entrez Gene ID
6159
Gene Name
ribosomal protein L29
Gene Aliases
HIP, HUMRPL29, L29, RPL29P10, RPL29_3_370
UniGene
Hs.425125
Gene Chromosome Location
Chr.3: 51993628-51995942 on Build GRCh38
Transcripts
NM_000992.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1046809 c.53G>A p.R18K Chr.3: 51995091-51995091 on Build GRCh38
3993197 c.33C>T p.N11N Chr.3: 51995429-51995429 on Build GRCh38
2852519 c.74G>T p.R25L Chr.3: 51995070-51995070 on Build GRCh38
6991820 c.61A>G p.I21V Chr.3: 51995083-51995083 on Build GRCh38
1617828 c.87C>G p.Y29* Chr.3: 51995057-51995057 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753786404 Chr.3: 51995089-51995089 on Build GRCh38
rs1003495295 Chr.3: 51995176-51995176 on Build GRCh38
rs879025442 Chr.3: 51995227-51995227 on Build GRCh38
rs145181246 Chr.3: 51995072-51995072 on Build GRCh38
rs771448655 Chr.3: 51995154-51995154 on Build GRCh38
rs778256940 Chr.3: 51995031-51995031 on Build GRCh38
rs372452330 Chr.3: 51994990-51994990 on Build GRCh38
rs567703640 Chr.3: 51995103-51995103 on Build GRCh38
rs779115397 Chr.3: 51995125-51995125 on Build GRCh38
rs140909692 Chr.3: 51995032-51995032 on Build GRCh38
rs768227761 Chr.3: 51995052-51995052 on Build GRCh38
rs771764948 Chr.3: 51995318-51995318 on Build GRCh38
rs755072539 Chr.3: 51994999-51994999 on Build GRCh38
rs761349784 Chr.3: 51995385-51995385 on Build GRCh38
rs376969283 Chr.3: 51995383-51995383 on Build GRCh38
rs768048389 Chr.3: 51995378-51995378 on Build GRCh38
rs141394482 Chr.3: 51995054-51995054 on Build GRCh38
rs771601931 Chr.3: 51995045-51995045 on Build GRCh38
rs1010598904 Chr.3: 51995274-51995274 on Build GRCh38
rs778943286 Chr.3: 51995002-51995002 on Build GRCh38
rs910708049 Chr.3: 51995412-51995412 on Build GRCh38
rs776544186 Chr.3: 51995404-51995404 on Build GRCh38
rs781742693 Chr.3: 51995158-51995158 on Build GRCh38
rs747341551 Chr.3: 51995153-51995153 on Build GRCh38
rs553663601 Chr.3: 51995392-51995392 on Build GRCh38
rs1037816266 Chr.3: 51995044-51995044 on Build GRCh38
rs895005636 Chr.3: 51995229-51995229 on Build GRCh38
rs966451679 Chr.3: 51995295-51995295 on Build GRCh38
rs746384999 Chr.3: 51995374-51995374 on Build GRCh38
rs773459190 Chr.3: 51995422-51995423 on Build GRCh38
rs760575973 Chr.3: 51995078-51995078 on Build GRCh38
rs772148318 Chr.3: 51995263-51995263 on Build GRCh38
rs938589823 Chr.3: 51995358-51995358 on Build GRCh38
rs1047723569 Chr.3: 51995138-51995138 on Build GRCh38
rs190552580 Chr.3: 51995360-51995360 on Build GRCh38
rs760404241 Chr.3: 51995397-51995397 on Build GRCh38
rs774951161 Chr.3: 51995326-51995326 on Build GRCh38
rs767205632 Chr.3: 51995070-51995070 on Build GRCh38
rs376766326 Chr.3: 51995344-51995344 on Build GRCh38
rs867681492 Chr.3: 51995387-51995387 on Build GRCh38
rs776989278 Chr.3: 51995389-51995390 on Build GRCh38
rs771802969 Chr.3: 51995151-51995151 on Build GRCh38
rs748405199 Chr.3: 51995010-51995010 on Build GRCh38
rs777964040 Chr.3: 51995152-51995152 on Build GRCh38
rs977299277 Chr.3: 51995308-51995308 on Build GRCh38
rs910758974 Chr.3: 51995026-51995026 on Build GRCh38
rs549938327 Chr.3: 51995283-51995283 on Build GRCh38
rs759525225 Chr.3: 51995419-51995419 on Build GRCh38
rs1015564601 Chr.3: 51995174-51995174 on Build GRCh38
rs375396108 Chr.3: 51995050-51995050 on Build GRCh38
rs762182187 Chr.3: 51995399-51995400 on Build GRCh38
rs371328431 Chr.3: 51995051-51995051 on Build GRCh38
rs953128149 Chr.3: 51995221-51995221 on Build GRCh38
rs373799953 Chr.3: 51995144-51995144 on Build GRCh38
rs752857868 Chr.3: 51995444-51995444 on Build GRCh38
rs796621837 Chr.3: 51995219-51995219 on Build GRCh38
rs990329114 Chr.3: 51995223-51995223 on Build GRCh38
rs895199441 Chr.3: 51995068-51995068 on Build GRCh38
rs766164893 Chr.3: 51995402-51995402 on Build GRCh38
rs549511263 Chr.3: 51995057-51995057 on Build GRCh38
rs533280345 Chr.3: 51995336-51995336 on Build GRCh38
rs999558088 Chr.3: 51995309-51995309 on Build GRCh38
rs538329973 Chr.3: 51995276-51995276 on Build GRCh38
rs200863244 Chr.3: 51995395-51995395 on Build GRCh38
rs967586213 Chr.3: 51995248-51995248 on Build GRCh38
rs371232875 Chr.3: 51995122-51995122 on Build GRCh38
rs146299911 Chr.3: 51995020-51995020 on Build GRCh38
rs766255228 Chr.3: 51995085-51995085 on Build GRCh38
rs997102523 Chr.3: 51995191-51995191 on Build GRCh38
rs752846984 Chr.3: 51995141-51995141 on Build GRCh38
rs369232552 Chr.3: 51995426-51995426 on Build GRCh38
rs1028647458 Chr.3: 51995215-51995215 on Build GRCh38
rs771797637 Chr.3: 51995389-51995389 on Build GRCh38
rs571967831 Chr.3: 51995441-51995441 on Build GRCh38
rs765349642 Chr.3: 51995411-51995411 on Build GRCh38
rs773971440 Chr.3: 51995380-51995380 on Build GRCh38
rs745649849 Chr.3: 51995009-51995010 on Build GRCh38
rs906406687 Chr.3: 51995169-51995169 on Build GRCh38

View Full Product Details