Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 53287052-53287553 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCP1A
Entrez Gene ID
55802
Gene Name
decapping mRNA 1A
Gene Aliases
HSA275986, Nbla00360, SMAD4IP1, SMIF
UniGene
Hs.476353
Gene Chromosome Location
Chr.3: 53283423-53347627 on Build GRCh38
Transcripts
NM_001290204.1 NM_001290205.1 NM_001290206.1 NM_001290207.1 NM_018403.6

SNP Detail

SNP ID Chromosome Location
rs35103194 Chr.3: 53287425-53287425 on Build GRCh38
rs189655368 Chr.3: 53287193-53287193 on Build GRCh38
rs1008876699 Chr.3: 53287226-53287226 on Build GRCh38
rs550293483 Chr.3: 53287086-53287086 on Build GRCh38
rs1000890063 Chr.3: 53287169-53287169 on Build GRCh38
rs140898371 Chr.3: 53287232-53287232 on Build GRCh38
rs748908106 Chr.3: 53287237-53287237 on Build GRCh38
rs34879291 Chr.3: 53287451-53287451 on Build GRCh38
rs984465492 Chr.3: 53287370-53287370 on Build GRCh38
rs116121608 Chr.3: 53287414-53287414 on Build GRCh38
rs35716152 Chr.3: 53287369-53287369 on Build GRCh38
rs946225009 Chr.3: 53287094-53287094 on Build GRCh38
rs534965669 Chr.3: 53287253-53287253 on Build GRCh38
rs768249276 Chr.3: 53287239-53287239 on Build GRCh38
rs11709050 Chr.3: 53287511-53287511 on Build GRCh38
rs75351315 Chr.3: 53287515-53287515 on Build GRCh38
rs151150072 Chr.3: 53287343-53287343 on Build GRCh38
rs374509545 Chr.3: 53287377-53287377 on Build GRCh38
rs962089734 Chr.3: 53287332-53287332 on Build GRCh38
rs554878842 Chr.3: 53287410-53287410 on Build GRCh38
rs35336193 Chr.3: 53287404-53287404 on Build GRCh38
rs994007306 Chr.3: 53287344-53287344 on Build GRCh38
rs566334845 Chr.3: 53287251-53287251 on Build GRCh38
rs35455336 Chr.3: 53287391-53287391 on Build GRCh38
rs952509080 Chr.3: 53287369-53287369 on Build GRCh38
rs1025525234 Chr.3: 53287345-53287345 on Build GRCh38
rs916998219 Chr.3: 53287490-53287490 on Build GRCh38
rs904696886 Chr.3: 53287161-53287161 on Build GRCh38
rs781887022 Chr.3: 53287342-53287342 on Build GRCh38
rs375028190 Chr.3: 53287456-53287456 on Build GRCh38
rs373454213 Chr.3: 53287370-53287370 on Build GRCh38
rs889140712 Chr.3: 53287222-53287222 on Build GRCh38
rs1041828985 Chr.3: 53287117-53287117 on Build GRCh38
rs11713081 Chr.3: 53287371-53287371 on Build GRCh38

View Full Product Details