Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109280697-109281203 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSRC1
Entrez Gene ID
84722
Gene Name
proline and serine rich coiled-coil 1
Gene Aliases
DDA3, FP3214
UniGene
Hs.405925
Gene Chromosome Location
Chr.1: 109279554-109283182 on Build GRCh38
Transcripts
NM_001005290.3 NM_001032291.2 NM_032636.7 XM_005271283.2 XM_011542306.2 XM_017002560.1 XM_017002561.1 XM_017002562.1 XM_017002563.1 XM_017002564.1 XM_017002565.1 XM_017002566.1 XM_017002567.1 XM_017002568.1 XM_017002569.1 XM_017002570.1 XM_017002571.1 XM_017002572.1 XM_017002573.1 XM_017002574.1 XM_017002575.1 XM_017002576.1 XM_017002577.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6453677 c.726C>T p.S242S Chr.1: 109280854-109280854 on Build GRCh38
6459090 c.665C>T p.A222V Chr.1: 109280915-109280915 on Build GRCh38
6440008 c.704_705CC>TT p.T235I Chr.1: 109280875-109280876 on Build GRCh38
6459089 c.856C>T p.P286S Chr.1: 109280915-109280915 on Build GRCh38
3996728 c.819T>C p.S273S Chr.1: 109280952-109280952 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs34863121 Chr.1: 109280836-109280836 on Build GRCh38
rs201288741 Chr.1: 109280977-109280977 on Build GRCh38
rs377629228 Chr.1: 109280837-109280837 on Build GRCh38
rs566855388 Chr.1: 109281101-109281101 on Build GRCh38
rs759346587 Chr.1: 109280981-109280981 on Build GRCh38
rs916716878 Chr.1: 109280724-109280724 on Build GRCh38
rs767191589 Chr.1: 109280774-109280774 on Build GRCh38
rs780411346 Chr.1: 109280853-109280853 on Build GRCh38
rs769227372 Chr.1: 109280858-109280858 on Build GRCh38
rs746035630 Chr.1: 109281153-109281153 on Build GRCh38
rs753766878 Chr.1: 109280805-109280805 on Build GRCh38
rs766944653 Chr.1: 109281172-109281172 on Build GRCh38
rs762813062 Chr.1: 109280990-109280990 on Build GRCh38
rs879157378 Chr.1: 109281154-109281154 on Build GRCh38
rs115080838 Chr.1: 109281174-109281174 on Build GRCh38
rs750252914 Chr.1: 109280735-109280735 on Build GRCh38
rs768941386 Chr.1: 109280946-109280946 on Build GRCh38
rs778494394 Chr.1: 109280746-109280746 on Build GRCh38
rs370247596 Chr.1: 109280888-109280888 on Build GRCh38
rs116389032 Chr.1: 109280835-109280835 on Build GRCh38
rs779467392 Chr.1: 109280948-109280948 on Build GRCh38
rs573223784 Chr.1: 109280758-109280758 on Build GRCh38
rs756907701 Chr.1: 109280928-109280928 on Build GRCh38
rs200443495 Chr.1: 109280906-109280906 on Build GRCh38
rs76057315 Chr.1: 109280915-109280915 on Build GRCh38
rs760988332 Chr.1: 109280886-109280886 on Build GRCh38
rs774521035 Chr.1: 109280768-109280768 on Build GRCh38
rs143295778 Chr.1: 109281110-109281110 on Build GRCh38
rs562216765 Chr.1: 109280751-109280751 on Build GRCh38
rs749926297 Chr.1: 109280918-109280919 on Build GRCh38
rs201557600 Chr.1: 109280795-109280795 on Build GRCh38
rs567496428 Chr.1: 109281023-109281023 on Build GRCh38
rs751535048 Chr.1: 109281056-109281056 on Build GRCh38
rs558698544 Chr.1: 109281133-109281133 on Build GRCh38
rs138862732 Chr.1: 109281142-109281142 on Build GRCh38
rs761891333 Chr.1: 109280778-109280778 on Build GRCh38
rs547156607 Chr.1: 109281098-109281098 on Build GRCh38
rs569207773 Chr.1: 109281148-109281148 on Build GRCh38
rs779333680 Chr.1: 109280783-109280783 on Build GRCh38
rs920274376 Chr.1: 109281001-109281001 on Build GRCh38
rs747692414 Chr.1: 109280747-109280747 on Build GRCh38
rs115047241 Chr.1: 109280887-109280887 on Build GRCh38
rs528132469 Chr.1: 109280931-109280931 on Build GRCh38
rs753572009 Chr.1: 109281177-109281177 on Build GRCh38
rs144022094 Chr.1: 109280796-109280796 on Build GRCh38
rs988503564 Chr.1: 109281074-109281074 on Build GRCh38
rs373207876 Chr.1: 109281173-109281173 on Build GRCh38
rs372778262 Chr.1: 109280763-109280763 on Build GRCh38
rs369453365 Chr.1: 109280738-109280738 on Build GRCh38
rs377253826 Chr.1: 109281169-109281169 on Build GRCh38
rs75417653 Chr.1: 109280952-109280952 on Build GRCh38
rs369340326 Chr.1: 109280754-109280754 on Build GRCh38
rs747843135 Chr.1: 109280897-109280897 on Build GRCh38
rs199962633 Chr.1: 109281176-109281176 on Build GRCh38
rs371358138 Chr.1: 109281109-109281109 on Build GRCh38
rs777044777 Chr.1: 109280978-109280978 on Build GRCh38
rs370025855 Chr.1: 109281017-109281017 on Build GRCh38
rs751091374 Chr.1: 109281007-109281007 on Build GRCh38
rs554828160 Chr.1: 109281181-109281181 on Build GRCh38
rs757958046 Chr.1: 109280990-109280991 on Build GRCh38
rs994143034 Chr.1: 109280878-109280878 on Build GRCh38
rs770130922 Chr.1: 109280852-109280852 on Build GRCh38
rs747067621 Chr.1: 109281053-109281053 on Build GRCh38
rs762318401 Chr.1: 109280992-109280992 on Build GRCh38
rs183826417 Chr.1: 109280866-109280866 on Build GRCh38
rs145365921 Chr.1: 109280929-109280929 on Build GRCh38
rs372725080 Chr.1: 109281162-109281162 on Build GRCh38
rs768504733 Chr.1: 109281168-109281168 on Build GRCh38
rs769940825 Chr.1: 109281161-109281161 on Build GRCh38
rs891555317 Chr.1: 109280794-109280794 on Build GRCh38
rs755722949 Chr.1: 109281127-109281127 on Build GRCh38
rs372190199 Chr.1: 109280970-109280970 on Build GRCh38
rs373785514 Chr.1: 109281124-109281124 on Build GRCh38
rs761749353 Chr.1: 109281170-109281170 on Build GRCh38
rs374362431 Chr.1: 109281024-109281024 on Build GRCh38
rs1023839607 Chr.1: 109280937-109280937 on Build GRCh38
rs377659117 Chr.1: 109281097-109281097 on Build GRCh38
rs375515364 Chr.1: 109281126-109281126 on Build GRCh38
rs199706816 Chr.1: 109280973-109280973 on Build GRCh38
rs765011270 Chr.1: 109280986-109280986 on Build GRCh38
rs1009537118 Chr.1: 109281171-109281171 on Build GRCh38
rs200801704 Chr.1: 109281045-109281045 on Build GRCh38
rs746731747 Chr.1: 109280846-109280846 on Build GRCh38
rs756809218 Chr.1: 109281009-109281009 on Build GRCh38
rs760943124 Chr.1: 109280974-109280974 on Build GRCh38
rs374787678 Chr.1: 109280767-109280767 on Build GRCh38
rs773468310 Chr.1: 109280872-109280872 on Build GRCh38
rs753029210 Chr.1: 109281138-109281138 on Build GRCh38
rs778396520 Chr.1: 109281147-109281147 on Build GRCh38
rs370847517 Chr.1: 109281113-109281113 on Build GRCh38
rs536764912 Chr.1: 109280737-109280737 on Build GRCh38
rs771178654 Chr.1: 109281078-109281078 on Build GRCh38
rs761919628 Chr.1: 109280867-109280867 on Build GRCh38
rs749755144 Chr.1: 109280854-109280854 on Build GRCh38
rs781437878 Chr.1: 109281151-109281151 on Build GRCh38
rs754975302 Chr.1: 109280832-109280832 on Build GRCh38
rs758831671 Chr.1: 109281178-109281178 on Build GRCh38
rs567432597 Chr.1: 109280730-109280730 on Build GRCh38
rs780789835 Chr.1: 109281010-109281010 on Build GRCh38
rs749005057 Chr.1: 109281165-109281165 on Build GRCh38
rs35358724 Chr.1: 109280799-109280800 on Build GRCh38
rs920387969 Chr.1: 109280788-109280788 on Build GRCh38
rs754518659 Chr.1: 109280741-109280741 on Build GRCh38
rs759501505 Chr.1: 109280895-109280895 on Build GRCh38
rs550864315 Chr.1: 109281015-109281015 on Build GRCh38
rs758856426 Chr.1: 109281033-109281033 on Build GRCh38
rs752620108 Chr.1: 109280905-109280905 on Build GRCh38
rs1047480378 Chr.1: 109281018-109281018 on Build GRCh38
rs766277944 Chr.1: 109281043-109281043 on Build GRCh38
rs776991427 Chr.1: 109281089-109281089 on Build GRCh38
rs370720646 Chr.1: 109280861-109280861 on Build GRCh38
rs765202106 Chr.1: 109280898-109280898 on Build GRCh38
rs374878696 Chr.1: 109280987-109280987 on Build GRCh38
rs368372959 Chr.1: 109280750-109280750 on Build GRCh38

View Full Product Details