Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 94063002-94063501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHFR2
Entrez Gene ID
200895
Gene Name
dihydrofolate reductase 2
Gene Aliases
DHFRL1, DHFRP4
UniGene
Hs.718516
Gene Chromosome Location
Chr.3: 94057922-94063656 on Build GRCh38
Transcripts
NM_001195643.1 XM_011512537.2 NM_022072.3

Gene Symbol
NSUN3
Entrez Gene ID
63899
Gene Name
NOP2/Sun RNA methyltransferase family member 3
Gene Aliases
MST077, MSTP077
UniGene
Hs.448003
Gene Chromosome Location
Chr.3: 94063011-94126786 on Build GRCh38
Transcripts
NM_001195643.1 XM_011512537.2 NM_022072.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3597957 c.1A>G p.M1V Chr.3: 94063127-94063127 on Build GRCh38
5954815 c.6G>A p.L2L Chr.3: 94063132-94063132 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs896131082 Chr.3: 94063029-94063029 on Build GRCh38
rs200722600 Chr.3: 94063154-94063154 on Build GRCh38
rs777740002 Chr.3: 94063146-94063146 on Build GRCh38
rs760255610 Chr.3: 94063121-94063121 on Build GRCh38
rs369301118 Chr.3: 94063189-94063189 on Build GRCh38
rs955370611 Chr.3: 94063214-94063214 on Build GRCh38
rs546505122 Chr.3: 94063127-94063127 on Build GRCh38
rs751949268 Chr.3: 94063088-94063088 on Build GRCh38
rs1030094358 Chr.3: 94063211-94063211 on Build GRCh38
rs749434221 Chr.3: 94063138-94063138 on Build GRCh38
rs878925525 Chr.3: 94063240-94063240 on Build GRCh38
rs918721101 Chr.3: 94063288-94063288 on Build GRCh38
rs1036161142 Chr.3: 94063217-94063217 on Build GRCh38
rs769492967 Chr.3: 94063166-94063166 on Build GRCh38
rs764753173 Chr.3: 94063082-94063082 on Build GRCh38
rs750557588 Chr.3: 94063120-94063120 on Build GRCh38
rs974247945 Chr.3: 94063272-94063272 on Build GRCh38
rs113542305 Chr.3: 94063248-94063248 on Build GRCh38
rs770595392 Chr.3: 94063160-94063160 on Build GRCh38
rs112562393 Chr.3: 94063077-94063077 on Build GRCh38
rs1045125040 Chr.3: 94063381-94063381 on Build GRCh38
rs758321795 Chr.3: 94063145-94063145 on Build GRCh38
rs947770084 Chr.3: 94063367-94063367 on Build GRCh38
rs1051648097 Chr.3: 94063253-94063253 on Build GRCh38
rs994237613 Chr.3: 94063249-94063249 on Build GRCh38
rs781643163 Chr.3: 94063105-94063105 on Build GRCh38
rs538273385 Chr.3: 94063445-94063445 on Build GRCh38
rs373976221 Chr.3: 94063073-94063073 on Build GRCh38
rs992335226 Chr.3: 94063051-94063051 on Build GRCh38
rs548771238 Chr.3: 94063190-94063190 on Build GRCh38
rs975986336 Chr.3: 94063210-94063210 on Build GRCh38
rs558400388 Chr.3: 94063454-94063454 on Build GRCh38
rs1006886237 Chr.3: 94063279-94063279 on Build GRCh38
rs1000535919 Chr.3: 94063407-94063407 on Build GRCh38
rs529792533 Chr.3: 94063224-94063224 on Build GRCh38
rs1044983194 Chr.3: 94063169-94063169 on Build GRCh38
rs752194788 Chr.3: 94063052-94063053 on Build GRCh38
rs563224021 Chr.3: 94063112-94063112 on Build GRCh38
rs755641915 Chr.3: 94063297-94063297 on Build GRCh38
rs967753612 Chr.3: 94063081-94063081 on Build GRCh38
rs565302810 Chr.3: 94063311-94063311 on Build GRCh38
rs903645565 Chr.3: 94063392-94063392 on Build GRCh38
rs1001075510 Chr.3: 94063396-94063396 on Build GRCh38
rs909166833 Chr.3: 94063182-94063182 on Build GRCh38
rs374944959 Chr.3: 94063167-94063167 on Build GRCh38
rs917681779 Chr.3: 94063460-94063460 on Build GRCh38
rs896718569 Chr.3: 94063357-94063357 on Build GRCh38
rs941832304 Chr.3: 94063191-94063191 on Build GRCh38
rs749724895 Chr.3: 94063170-94063170 on Build GRCh38
rs917562009 Chr.3: 94063143-94063143 on Build GRCh38
rs776507869 Chr.3: 94063161-94063161 on Build GRCh38
rs891111969 Chr.3: 94063257-94063257 on Build GRCh38
rs567203072 Chr.3: 94063200-94063200 on Build GRCh38
rs957105416 Chr.3: 94063411-94063411 on Build GRCh38
rs1031104978 Chr.3: 94063400-94063400 on Build GRCh38
rs551393730 Chr.3: 94063305-94063305 on Build GRCh38
rs756369375 Chr.3: 94063123-94063123 on Build GRCh38
rs985849541 Chr.3: 94063237-94063239 on Build GRCh38
rs376685855 Chr.3: 94063233-94063233 on Build GRCh38
rs968238651 Chr.3: 94063352-94063352 on Build GRCh38
rs941421243 Chr.3: 94063271-94063271 on Build GRCh38
rs566112877 Chr.3: 94063347-94063347 on Build GRCh38
rs200084381 Chr.3: 94063163-94063163 on Build GRCh38
rs930241221 Chr.3: 94063303-94063303 on Build GRCh38

View Full Product Details