Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 112345192-112345737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD200
Entrez Gene ID
4345
Gene Name
CD200 molecule
Gene Aliases
MOX1, MOX2, MRC, OX-2
UniGene
Hs.79015
Gene Chromosome Location
Chr.3: 112332224-112362812 on Build GRCh38
Transcripts
NM_001004196.3 NM_001318826.1 NM_001318828.1 NM_001318830.1 NM_005944.6 XM_005247481.4 XM_005247482.2 XM_005247485.2 XM_005247486.2 XM_005247488.2 XM_017006452.1 XM_017006453.1 XM_017006454.1 XM_017006455.1 XM_017006456.1 XM_017006457.1 XM_017006458.1 XM_017006459.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7116712 c.470C>T p.S157L Chr.3: 112345262-112345262 on Build GRCh38
339447 c.412C>T p.L138L Chr.3: 112345204-112345204 on Build GRCh38
3974077 c.444C>T p.F148F Chr.3: 112345236-112345236 on Build GRCh38
5511925 c.476C>T p.T159M Chr.3: 112345268-112345268 on Build GRCh38
3118355 c.408C>T p.I136I Chr.3: 112345200-112345200 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1029430368 Chr.3: 112345466-112345466 on Build GRCh38
rs41270439 Chr.3: 112345357-112345357 on Build GRCh38
rs948862862 Chr.3: 112345355-112345355 on Build GRCh38
rs951267047 Chr.3: 112345395-112345395 on Build GRCh38
rs941620793 Chr.3: 112345345-112345345 on Build GRCh38
rs767343650 Chr.3: 112345329-112345329 on Build GRCh38
rs78923604 Chr.3: 112345629-112345629 on Build GRCh38
rs529327173 Chr.3: 112345653-112345653 on Build GRCh38
rs767314171 Chr.3: 112345600-112345600 on Build GRCh38
rs776832299 Chr.3: 112345288-112345288 on Build GRCh38
rs974356333 Chr.3: 112345535-112345535 on Build GRCh38
rs376525029 Chr.3: 112345218-112345218 on Build GRCh38
rs1053562551 Chr.3: 112345619-112345619 on Build GRCh38
rs60497880 Chr.3: 112345419-112345419 on Build GRCh38
rs989743681 Chr.3: 112345278-112345278 on Build GRCh38
rs199651350 Chr.3: 112345287-112345287 on Build GRCh38
rs955092843 Chr.3: 112345472-112345472 on Build GRCh38
rs200134698 Chr.3: 112345324-112345324 on Build GRCh38
rs750281125 Chr.3: 112345265-112345265 on Build GRCh38
rs549002713 Chr.3: 112345675-112345675 on Build GRCh38
rs1045813494 Chr.3: 112345356-112345356 on Build GRCh38
rs747091243 Chr.3: 112345286-112345286 on Build GRCh38
rs1045863813 Chr.3: 112345575-112345575 on Build GRCh38
rs148497135 Chr.3: 112345649-112345649 on Build GRCh38
rs780968132 Chr.3: 112345247-112345247 on Build GRCh38
rs763680234 Chr.3: 112345315-112345315 on Build GRCh38
rs377372870 Chr.3: 112345583-112345583 on Build GRCh38
rs1016777280 Chr.3: 112345695-112345695 on Build GRCh38
rs79203724 Chr.3: 112345269-112345269 on Build GRCh38
rs1043666165 Chr.3: 112345639-112345639 on Build GRCh38
rs375873909 Chr.3: 112345231-112345231 on Build GRCh38
rs1025489789 Chr.3: 112345384-112345384 on Build GRCh38
rs777667143 Chr.3: 112345284-112345284 on Build GRCh38
rs573610876 Chr.3: 112345431-112345431 on Build GRCh38
rs762534304 Chr.3: 112345216-112345216 on Build GRCh38
rs895772138 Chr.3: 112345354-112345354 on Build GRCh38
rs201035128 Chr.3: 112345386-112345386 on Build GRCh38
rs909567806 Chr.3: 112345227-112345227 on Build GRCh38
rs773374389 Chr.3: 112345331-112345331 on Build GRCh38
rs946833155 Chr.3: 112345518-112345518 on Build GRCh38
rs117277710 Chr.3: 112345488-112345488 on Build GRCh38
rs11921546 Chr.3: 112345416-112345416 on Build GRCh38
rs1022125191 Chr.3: 112345280-112345280 on Build GRCh38
rs1016661839 Chr.3: 112345409-112345409 on Build GRCh38
rs531127167 Chr.3: 112345512-112345512 on Build GRCh38
rs886956506 Chr.3: 112345682-112345682 on Build GRCh38
rs963814629 Chr.3: 112345713-112345713 on Build GRCh38
rs771330143 Chr.3: 112345405-112345405 on Build GRCh38
rs34710949 Chr.3: 112345240-112345240 on Build GRCh38
rs112085090 Chr.3: 112345282-112345282 on Build GRCh38
rs1038978855 Chr.3: 112345350-112345350 on Build GRCh38
rs183888589 Chr.3: 112345606-112345606 on Build GRCh38
rs773778920 Chr.3: 112345318-112345318 on Build GRCh38
rs988767042 Chr.3: 112345505-112345505 on Build GRCh38
rs564535248 Chr.3: 112345623-112345623 on Build GRCh38
rs998841836 Chr.3: 112345376-112345376 on Build GRCh38
rs118005027 Chr.3: 112345401-112345401 on Build GRCh38
rs533484933 Chr.3: 112345624-112345624 on Build GRCh38
rs113545453 Chr.3: 112345352-112345352 on Build GRCh38
rs1047383997 Chr.3: 112345662-112345665 on Build GRCh38
rs577378883 Chr.3: 112345560-112345560 on Build GRCh38
rs867172395 Chr.3: 112345241-112345242 on Build GRCh38
rs774267779 Chr.3: 112345233-112345233 on Build GRCh38
rs948904414 Chr.3: 112345573-112345573 on Build GRCh38
rs750086735 Chr.3: 112345340-112345340 on Build GRCh38
rs763325053 Chr.3: 112345314-112345314 on Build GRCh38
rs759956477 Chr.3: 112345299-112345299 on Build GRCh38
rs761829311 Chr.3: 112345581-112345581 on Build GRCh38
rs1029481243 Chr.3: 112345685-112345685 on Build GRCh38
rs113185557 Chr.3: 112345428-112345428 on Build GRCh38
rs144847706 Chr.3: 112345558-112345558 on Build GRCh38
rs113483136 Chr.3: 112345479-112345479 on Build GRCh38
rs368810090 Chr.3: 112345268-112345268 on Build GRCh38
rs916126276 Chr.3: 112345539-112345539 on Build GRCh38
rs79269828 Chr.3: 112345281-112345281 on Build GRCh38
rs375864607 Chr.3: 112345311-112345311 on Build GRCh38

View Full Product Details