Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 121911441-121911967 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC15A2
Entrez Gene ID
6565
Gene Name
solute carrier family 15 member 2
Gene Aliases
PEPT2
UniGene
Hs.518089
Gene Chromosome Location
Chr.3: 121894324-121944187 on Build GRCh38
Transcripts
NM_001145998.1 NM_021082.3 XM_005247722.3 XM_006713736.3 XM_017007074.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1693280 c.349C>T p.L117F Chr.3: 121911587-121911587 on Build GRCh38
108019 c.353C>T p.S118F Chr.3: 121911591-121911591 on Build GRCh38
1418219 c.362A>G p.Y121C Chr.3: 121911600-121911600 on Build GRCh38
3121057 c.428+1G>A p.? Chr.3: 121911667-121911667 on Build GRCh38
4935835 c.415C>A p.Q139K Chr.3: 121911653-121911653 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs896881989 Chr.3: 121911600-121911600 on Build GRCh38
rs536388645 Chr.3: 121911575-121911575 on Build GRCh38
rs949239057 Chr.3: 121911571-121911571 on Build GRCh38
rs756533683 Chr.3: 121911547-121911547 on Build GRCh38
rs1020896021 Chr.3: 121911768-121911768 on Build GRCh38
rs1048148268 Chr.3: 121911625-121911625 on Build GRCh38
rs1037720498 Chr.3: 121911532-121911532 on Build GRCh38
rs770725883 Chr.3: 121911608-121911608 on Build GRCh38
rs767664905 Chr.3: 121911645-121911645 on Build GRCh38
rs949348479 Chr.3: 121911885-121911885 on Build GRCh38
rs138471341 Chr.3: 121911591-121911591 on Build GRCh38
rs775669951 Chr.3: 121911714-121911714 on Build GRCh38
rs979683863 Chr.3: 121911831-121911831 on Build GRCh38
rs369991905 Chr.3: 121911643-121911643 on Build GRCh38
rs371513641 Chr.3: 121911666-121911666 on Build GRCh38
rs549056117 Chr.3: 121911830-121911830 on Build GRCh38
rs767956544 Chr.3: 121911533-121911533 on Build GRCh38
rs772342164 Chr.3: 121911706-121911707 on Build GRCh38
rs577892849 Chr.3: 121911740-121911740 on Build GRCh38
rs913830124 Chr.3: 121911799-121911799 on Build GRCh38
rs6438687 Chr.3: 121911481-121911481 on Build GRCh38
rs115742448 Chr.3: 121911766-121911766 on Build GRCh38
rs540890984 Chr.3: 121911652-121911652 on Build GRCh38
rs754271930 Chr.3: 121911664-121911664 on Build GRCh38
rs145777064 Chr.3: 121911785-121911785 on Build GRCh38
rs777365545 Chr.3: 121911708-121911708 on Build GRCh38
rs778534312 Chr.3: 121911583-121911583 on Build GRCh38
rs186149211 Chr.3: 121911915-121911915 on Build GRCh38
rs542107292 Chr.3: 121911554-121911554 on Build GRCh38
rs778589273 Chr.3: 121911696-121911696 on Build GRCh38
rs200983585 Chr.3: 121911691-121911691 on Build GRCh38
rs992074671 Chr.3: 121911788-121911788 on Build GRCh38
rs780486500 Chr.3: 121911550-121911550 on Build GRCh38
rs937343599 Chr.3: 121911502-121911502 on Build GRCh38
rs917846830 Chr.3: 121911529-121911529 on Build GRCh38
rs530773286 Chr.3: 121911925-121911925 on Build GRCh38
rs992860401 Chr.3: 121911811-121911811 on Build GRCh38
rs747718082 Chr.3: 121911585-121911585 on Build GRCh38
rs752316795 Chr.3: 121911702-121911702 on Build GRCh38
rs760659543 Chr.3: 121911470-121911470 on Build GRCh38
rs371506732 Chr.3: 121911467-121911467 on Build GRCh38
rs771590932 Chr.3: 121911594-121911594 on Build GRCh38
rs766809516 Chr.3: 121911660-121911660 on Build GRCh38
rs761333212 Chr.3: 121911749-121911749 on Build GRCh38
rs746999057 Chr.3: 121911603-121911603 on Build GRCh38
rs528655814 Chr.3: 121911927-121911927 on Build GRCh38
rs750907563 Chr.3: 121911539-121911539 on Build GRCh38
rs958660202 Chr.3: 121911800-121911800 on Build GRCh38
rs189488640 Chr.3: 121911769-121911769 on Build GRCh38
rs990695107 Chr.3: 121911516-121911516 on Build GRCh38
rs181504404 Chr.3: 121911898-121911898 on Build GRCh38
rs199897644 Chr.3: 121911640-121911640 on Build GRCh38
rs757860397 Chr.3: 121911663-121911663 on Build GRCh38
rs776570546 Chr.3: 121911613-121911613 on Build GRCh38
rs1034436162 Chr.3: 121911792-121911792 on Build GRCh38
rs375861983 Chr.3: 121911602-121911602 on Build GRCh38
rs764466745 Chr.3: 121911626-121911626 on Build GRCh38
rs201991806 Chr.3: 121911647-121911647 on Build GRCh38
rs749599908 Chr.3: 121911563-121911563 on Build GRCh38
rs1037310022 Chr.3: 121911587-121911587 on Build GRCh38
rs757949184 Chr.3: 121911703-121911703 on Build GRCh38

View Full Product Details