Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 126105691-126106184 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH1L1
Entrez Gene ID
10840
Gene Name
aldehyde dehydrogenase 1 family member L1
Gene Aliases
10-FTHFDH, 10-fTHF, FDH, FTHFD
UniGene
Hs.434435
Gene Chromosome Location
Chr.3: 126103561-126181526 on Build GRCh38
Transcripts
NM_001270364.1 NM_001270365.1 NM_012190.3 NR_072979.1 XM_006713481.2 XM_011512355.1 XM_017005613.1 XM_017005614.1 NR_046602.1

Gene Symbol
ALDH1L1-AS1
Entrez Gene ID
100874204
Gene Name
ALDH1L1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.742339
Gene Chromosome Location
Chr.3: 126103640-126108069 on Build GRCh38
Transcripts
NM_001270364.1 NM_001270365.1 NM_012190.3 NR_072979.1 XM_006713481.2 XM_011512355.1 XM_017005613.1 XM_017005614.1 NR_046602.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
339449 c.2586G>T p.T862T Chr.3: 126105793-126105793 on Build GRCh38
1685419 c.2619A>G p.G873G Chr.3: 126105760-126105760 on Build GRCh38
3673828 c.2547C>T p.V849V Chr.3: 126105832-126105832 on Build GRCh38
4482643 c.2597C>T p.T866I Chr.3: 126105782-126105782 on Build GRCh38
5368257 c.2585C>T p.T862M Chr.3: 126105794-126105794 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761642800 Chr.3: 126105891-126105891 on Build GRCh38
rs980214591 Chr.3: 126106102-126106102 on Build GRCh38
rs773196663 Chr.3: 126105898-126105898 on Build GRCh38
rs751304415 Chr.3: 126105787-126105787 on Build GRCh38
rs768885925 Chr.3: 126105889-126105889 on Build GRCh38
rs189265455 Chr.3: 126106045-126106045 on Build GRCh38
rs538821586 Chr.3: 126105890-126105890 on Build GRCh38
rs898481388 Chr.3: 126105785-126105785 on Build GRCh38
rs561248921 Chr.3: 126106149-126106149 on Build GRCh38
rs114832953 Chr.3: 126106158-126106158 on Build GRCh38
rs142086991 Chr.3: 126105752-126105752 on Build GRCh38
rs752368429 Chr.3: 126105914-126105914 on Build GRCh38
rs370612209 Chr.3: 126105942-126105942 on Build GRCh38
rs530057896 Chr.3: 126105829-126105829 on Build GRCh38
rs764315946 Chr.3: 126105778-126105794 on Build GRCh38
rs550253428 Chr.3: 126106070-126106070 on Build GRCh38
rs766017092 Chr.3: 126106055-126106055 on Build GRCh38
rs150564471 Chr.3: 126106150-126106150 on Build GRCh38
rs767390342 Chr.3: 126105793-126105793 on Build GRCh38
rs960789769 Chr.3: 126105945-126105945 on Build GRCh38
rs763813220 Chr.3: 126105782-126105782 on Build GRCh38
rs1037556651 Chr.3: 126106008-126106008 on Build GRCh38
rs779608112 Chr.3: 126105796-126105796 on Build GRCh38
rs755656855 Chr.3: 126105794-126105794 on Build GRCh38
rs545195717 Chr.3: 126106072-126106072 on Build GRCh38
rs746372949 Chr.3: 126105855-126105855 on Build GRCh38
rs775642396 Chr.3: 126105780-126105780 on Build GRCh38
rs113845761 Chr.3: 126105770-126105770 on Build GRCh38
rs753490987 Chr.3: 126105822-126105822 on Build GRCh38
rs968374394 Chr.3: 126106088-126106088 on Build GRCh38
rs755783175 Chr.3: 126105751-126105751 on Build GRCh38
rs748360743 Chr.3: 126105971-126105971 on Build GRCh38
rs200290145 Chr.3: 126105771-126105771 on Build GRCh38
rs760592282 Chr.3: 126105902-126105902 on Build GRCh38
rs979854934 Chr.3: 126105927-126105927 on Build GRCh38
rs561296762 Chr.3: 126105781-126105781 on Build GRCh38
rs757762713 Chr.3: 126105923-126105923 on Build GRCh38
rs772384247 Chr.3: 126105763-126105763 on Build GRCh38
rs191970426 Chr.3: 126105772-126105772 on Build GRCh38
rs778930779 Chr.3: 126105978-126105994 on Build GRCh38
rs759169238 Chr.3: 126106057-126106057 on Build GRCh38
rs565092674 Chr.3: 126106087-126106087 on Build GRCh38
rs938458385 Chr.3: 126106139-126106139 on Build GRCh38
rs886259124 Chr.3: 126105995-126105995 on Build GRCh38
rs537898479 Chr.3: 126105955-126105955 on Build GRCh38
rs376780648 Chr.3: 126105913-126105913 on Build GRCh38
rs757393751 Chr.3: 126105966-126105967 on Build GRCh38
rs747482271 Chr.3: 126105768-126105768 on Build GRCh38
rs939653053 Chr.3: 126105809-126105809 on Build GRCh38
rs754045034 Chr.3: 126105785-126105787 on Build GRCh38
rs141656827 Chr.3: 126105912-126105912 on Build GRCh38
rs749342590 Chr.3: 126105943-126105943 on Build GRCh38
rs763094048 Chr.3: 126105881-126105881 on Build GRCh38
rs980122951 Chr.3: 126106030-126106030 on Build GRCh38
rs565026541 Chr.3: 126105742-126105742 on Build GRCh38
rs886394320 Chr.3: 126105777-126105777 on Build GRCh38
rs774672237 Chr.3: 126105959-126105959 on Build GRCh38
rs376404957 Chr.3: 126105778-126105778 on Build GRCh38
rs771908952 Chr.3: 126105972-126105972 on Build GRCh38
rs139413481 Chr.3: 126105845-126105845 on Build GRCh38
rs372302697 Chr.3: 126106046-126106046 on Build GRCh38
rs989076577 Chr.3: 126106076-126106076 on Build GRCh38
rs781680826 Chr.3: 126105928-126105928 on Build GRCh38
rs371106041 Chr.3: 126105896-126105896 on Build GRCh38
rs72965956 Chr.3: 126105876-126105876 on Build GRCh38
rs746393449 Chr.3: 126105929-126105929 on Build GRCh38
rs940632725 Chr.3: 126106007-126106007 on Build GRCh38
rs745778125 Chr.3: 126106022-126106022 on Build GRCh38
rs772932999 Chr.3: 126105974-126105974 on Build GRCh38
rs988112359 Chr.3: 126105950-126105950 on Build GRCh38
rs4646754 Chr.3: 126106056-126106056 on Build GRCh38
rs375074619 Chr.3: 126105903-126105903 on Build GRCh38
rs931236937 Chr.3: 126106106-126106106 on Build GRCh38
rs575445311 Chr.3: 126106113-126106113 on Build GRCh38
rs552442699 Chr.3: 126105907-126105907 on Build GRCh38
rs554383640 Chr.3: 126105994-126105994 on Build GRCh38
rs778002659 Chr.3: 126105841-126105841 on Build GRCh38
rs756626583 Chr.3: 126105941-126105941 on Build GRCh38
rs544268504 Chr.3: 126106127-126106127 on Build GRCh38
rs748936377 Chr.3: 126105762-126105762 on Build GRCh38
rs778185806 Chr.3: 126105766-126105766 on Build GRCh38
rs1019775598 Chr.3: 126105861-126105861 on Build GRCh38
rs771401495 Chr.3: 126105848-126105848 on Build GRCh38
rs749897803 Chr.3: 126105743-126105743 on Build GRCh38
rs905781851 Chr.3: 126106132-126106133 on Build GRCh38
rs770058477 Chr.3: 126105870-126105870 on Build GRCh38
rs773003853 Chr.3: 126106029-126106029 on Build GRCh38
rs202021917 Chr.3: 126105854-126105854 on Build GRCh38

View Full Product Details